Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 138714312-138714846 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF13
Entrez Gene ID
2258
Gene Name
fibroblast growth factor 13
Gene Aliases
FGF-13, FGF2, FHF-2, FHF2
UniGene
Hs.6540
Gene Chromosome Location
Chr.X: 138631573-139222889 on Build GRCh38
Transcripts
NM_001139498.1 NM_001139500.1 NM_001139501.1 NM_001139502.1 NM_033642.2 XM_005262399.1 NR_038405.1

Gene Symbol
FGF13-AS1
Entrez Gene ID
100129662
Gene Name
FGF13 antisense RNA 1
Gene Aliases
-
UniGene
Hs.571747
Gene Chromosome Location
Chr.X: 138712107-138716605 on Build GRCh38
Transcripts
NM_001139498.1 NM_001139500.1 NM_001139501.1 NM_001139502.1 NM_033642.2 XM_005262399.1 NR_038405.1

SNP Detail

SNP ID Chromosome Location
rs923517845 Chr.X: 138714640-138714653 on Build GRCh38
rs973223295 Chr.X: 138714596-138714596 on Build GRCh38
rs1052433384 Chr.X: 138714661-138714661 on Build GRCh38
rs866883539 Chr.X: 138714436-138714436 on Build GRCh38
rs963492528 Chr.X: 138714577-138714577 on Build GRCh38
rs769510549 Chr.X: 138714417-138714417 on Build GRCh38
rs773641064 Chr.X: 138714624-138714624 on Build GRCh38
rs893586651 Chr.X: 138714383-138714383 on Build GRCh38
rs537063087 Chr.X: 138714655-138714655 on Build GRCh38
rs900654578 Chr.X: 138714657-138714657 on Build GRCh38
rs761540470 Chr.X: 138714567-138714567 on Build GRCh38
rs763551426 Chr.X: 138714633-138714634 on Build GRCh38
rs781107823 Chr.X: 138714372-138714372 on Build GRCh38
rs746960311 Chr.X: 138714812-138714812 on Build GRCh38
rs890422810 Chr.X: 138714526-138714526 on Build GRCh38
rs776490267 Chr.X: 138714471-138714471 on Build GRCh38
rs987849887 Chr.X: 138714647-138714647 on Build GRCh38
rs775830236 Chr.X: 138714795-138714795 on Build GRCh38
rs189269086 Chr.X: 138714792-138714792 on Build GRCh38
rs969214892 Chr.X: 138714616-138714616 on Build GRCh38
rs899169856 Chr.X: 138714714-138714714 on Build GRCh38
rs184952127 Chr.X: 138714743-138714743 on Build GRCh38
rs145707053 Chr.X: 138714692-138714692 on Build GRCh38
rs1015402817 Chr.X: 138714567-138714567 on Build GRCh38
rs943520617 Chr.X: 138714553-138714553 on Build GRCh38
rs932181811 Chr.X: 138714660-138714660 on Build GRCh38
rs1030600296 Chr.X: 138714455-138714455 on Build GRCh38
rs979504513 Chr.X: 138714501-138714501 on Build GRCh38
rs1004912017 Chr.X: 138714560-138714560 on Build GRCh38
rs935033562 Chr.X: 138714371-138714371 on Build GRCh38
rs762504981 Chr.X: 138714711-138714711 on Build GRCh38
rs1011683279 Chr.X: 138714384-138714384 on Build GRCh38
rs982415967 Chr.X: 138714621-138714621 on Build GRCh38
rs934980066 Chr.X: 138714626-138714626 on Build GRCh38
rs910692534 Chr.X: 138714506-138714506 on Build GRCh38
rs773002801 Chr.X: 138714654-138714654 on Build GRCh38
rs774903265 Chr.X: 138714649-138714649 on Build GRCh38
rs770291921 Chr.X: 138714609-138714609 on Build GRCh38
rs768178842 Chr.X: 138714452-138714452 on Build GRCh38
rs140333976 Chr.X: 138714418-138714418 on Build GRCh38
rs747743073 Chr.X: 138714403-138714403 on Build GRCh38
rs1023746520 Chr.X: 138714767-138714767 on Build GRCh38
rs1039430229 Chr.X: 138714656-138714656 on Build GRCh38
rs766750903 Chr.X: 138714646-138714646 on Build GRCh38
rs758188883 Chr.X: 138714715-138714715 on Build GRCh38
rs191630464 Chr.X: 138714663-138714663 on Build GRCh38
rs771057311 Chr.X: 138714662-138714663 on Build GRCh38
rs767582261 Chr.X: 138714661-138714662 on Build GRCh38
rs754590395 Chr.X: 138714822-138714822 on Build GRCh38
rs891673489 Chr.X: 138714666-138714666 on Build GRCh38
rs988538289 Chr.X: 138714379-138714379 on Build GRCh38

View Full Product Details