Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 138714719-138715220 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FGF13
Entrez Gene ID
2258
Gene Name
fibroblast growth factor 13
Gene Aliases
FGF-13, FGF2, FHF-2, FHF2
UniGene
Hs.6540
Gene Chromosome Location
Chr.X: 138631573-139222889 on Build GRCh38
Transcripts
NM_001139498.1 NM_001139500.1 NM_001139501.1 NM_001139502.1 NM_033642.2 XM_005262399.1 NR_038405.1

Gene Symbol
FGF13-AS1
Entrez Gene ID
100129662
Gene Name
FGF13 antisense RNA 1
Gene Aliases
-
UniGene
Hs.571747
Gene Chromosome Location
Chr.X: 138712107-138716605 on Build GRCh38
Transcripts
NM_001139498.1 NM_001139500.1 NM_001139501.1 NM_001139502.1 NM_033642.2 XM_005262399.1 NR_038405.1

SNP Detail

SNP ID Chromosome Location
rs940917500 Chr.X: 138715185-138715185 on Build GRCh38
rs774789335 Chr.X: 138715151-138715151 on Build GRCh38
rs773737435 Chr.X: 138715066-138715066 on Build GRCh38
rs890693469 Chr.X: 138715130-138715130 on Build GRCh38
rs930763633 Chr.X: 138714944-138714944 on Build GRCh38
rs754178054 Chr.X: 138715193-138715193 on Build GRCh38
rs921040968 Chr.X: 138715110-138715110 on Build GRCh38
rs767522992 Chr.X: 138714974-138714974 on Build GRCh38
rs746960311 Chr.X: 138714812-138714812 on Build GRCh38
rs747803038 Chr.X: 138714952-138714953 on Build GRCh38
rs1023746520 Chr.X: 138714767-138714767 on Build GRCh38
rs775830236 Chr.X: 138714795-138714795 on Build GRCh38
rs189269086 Chr.X: 138714792-138714792 on Build GRCh38
rs975103737 Chr.X: 138715093-138715093 on Build GRCh38
rs956689427 Chr.X: 138714868-138714868 on Build GRCh38
rs988205286 Chr.X: 138714894-138714894 on Build GRCh38
rs1051975239 Chr.X: 138715123-138715123 on Build GRCh38
rs771298938 Chr.X: 138715129-138715129 on Build GRCh38
rs923474251 Chr.X: 138715021-138715021 on Build GRCh38
rs769526922 Chr.X: 138714991-138714991 on Build GRCh38
rs946403331 Chr.X: 138715052-138715052 on Build GRCh38
rs184952127 Chr.X: 138714743-138714743 on Build GRCh38
rs764357212 Chr.X: 138714861-138714861 on Build GRCh38
rs865909068 Chr.X: 138715022-138715022 on Build GRCh38
rs749666295 Chr.X: 138715118-138715118 on Build GRCh38
rs914875429 Chr.X: 138714921-138714921 on Build GRCh38
rs950996450 Chr.X: 138715082-138715082 on Build GRCh38
rs754590395 Chr.X: 138714822-138714822 on Build GRCh38
rs983715299 Chr.X: 138715103-138715103 on Build GRCh38
rs780788156 Chr.X: 138714847-138714847 on Build GRCh38

View Full Product Details