Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 6968626-6969192 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DEFA10P
Entrez Gene ID
449493
Gene Name
defensin alpha 10, pseudogene
Gene Aliases
-
UniGene
Hs.552564
Gene Chromosome Location
Chr.8: 6968141-6969113 on Build GRCh38
Transcripts
NR_029386.1

SNP Detail

SNP ID Chromosome Location
rs560881745 Chr.8: 6968967-6968967 on Build GRCh38
rs1043868216 Chr.8: 6968781-6968781 on Build GRCh38
rs183360357 Chr.8: 6969103-6969103 on Build GRCh38
rs759545311 Chr.8: 6968765-6968765 on Build GRCh38
rs1045403361 Chr.8: 6968841-6968841 on Build GRCh38
rs991364963 Chr.8: 6969003-6969003 on Build GRCh38
rs550831258 Chr.8: 6968808-6968808 on Build GRCh38
rs998064000 Chr.8: 6968726-6968740 on Build GRCh38
rs935904995 Chr.8: 6968784-6968784 on Build GRCh38
rs562957373 Chr.8: 6969054-6969054 on Build GRCh38
rs116472946 Chr.8: 6968789-6968789 on Build GRCh38
rs115702637 Chr.8: 6968862-6968862 on Build GRCh38
rs886822713 Chr.8: 6968842-6968842 on Build GRCh38
rs910574598 Chr.8: 6968916-6968916 on Build GRCh38
rs898689065 Chr.8: 6968865-6968865 on Build GRCh38
rs955386145 Chr.8: 6968692-6968692 on Build GRCh38
rs185356923 Chr.8: 6968831-6968831 on Build GRCh38
rs201673847 Chr.8: 6968813-6968813 on Build GRCh38
rs986111091 Chr.8: 6968739-6968739 on Build GRCh38
rs200246650 Chr.8: 6968843-6968843 on Build GRCh38
rs892676186 Chr.8: 6968769-6968769 on Build GRCh38
rs199626845 Chr.8: 6969011-6969011 on Build GRCh38
rs202169189 Chr.8: 6968821-6968821 on Build GRCh38
rs71525779 Chr.8: 6968748-6968748 on Build GRCh38
rs944986328 Chr.8: 6968751-6968751 on Build GRCh38
rs531859988 Chr.8: 6969055-6969055 on Build GRCh38
rs914149153 Chr.8: 6968929-6968929 on Build GRCh38
rs1049929759 Chr.8: 6969085-6969085 on Build GRCh38
rs945530655 Chr.8: 6968779-6968779 on Build GRCh38
rs977629573 Chr.8: 6968734-6968734 on Build GRCh38
rs71511144 Chr.8: 6968696-6968735 on Build GRCh38
rs2951842 Chr.8: 6968999-6968999 on Build GRCh38
rs3739385 Chr.8: 6968693-6968693 on Build GRCh38
rs1027928334 Chr.8: 6969089-6969089 on Build GRCh38
rs879593679 Chr.8: 6968746-6968746 on Build GRCh38
rs753323914 Chr.8: 6968925-6968925 on Build GRCh38
rs986163364 Chr.8: 6968907-6968907 on Build GRCh38
rs902544062 Chr.8: 6969113-6969113 on Build GRCh38
rs532598396 Chr.8: 6968767-6968767 on Build GRCh38
rs1027464266 Chr.8: 6968899-6968899 on Build GRCh38
rs1014219738 Chr.8: 6968879-6968879 on Build GRCh38
rs1017636815 Chr.8: 6969117-6969117 on Build GRCh38
rs1033977736 Chr.8: 6968655-6968655 on Build GRCh38
rs115010656 Chr.8: 6968890-6968890 on Build GRCh38
rs368576333 Chr.8: 6968868-6968868 on Build GRCh38
rs140219298 Chr.8: 6968926-6968926 on Build GRCh38
rs374573258 Chr.8: 6969098-6969098 on Build GRCh38
rs554764419 Chr.8: 6968924-6968924 on Build GRCh38
rs571196508 Chr.8: 6969084-6969084 on Build GRCh38
rs35495028 Chr.8: 6968884-6968884 on Build GRCh38
rs915572188 Chr.8: 6968799-6968799 on Build GRCh38
rs908219497 Chr.8: 6969044-6969044 on Build GRCh38
rs765668061 Chr.8: 6968696-6968696 on Build GRCh38
rs763530404 Chr.8: 6968893-6968893 on Build GRCh38
rs550770462 Chr.8: 6969073-6969073 on Build GRCh38
rs1024220628 Chr.8: 6968882-6968882 on Build GRCh38
rs958630609 Chr.8: 6968680-6968680 on Build GRCh38
rs190655638 Chr.8: 6969018-6969018 on Build GRCh38
rs1024173417 Chr.8: 6968684-6968684 on Build GRCh38
rs778541324 Chr.8: 6969119-6969119 on Build GRCh38
rs200374870 Chr.8: 6968793-6968793 on Build GRCh38
rs894180736 Chr.8: 6969030-6969030 on Build GRCh38
rs945611274 Chr.8: 6968969-6968969 on Build GRCh38
rs981483759 Chr.8: 6969025-6969025 on Build GRCh38
rs920682957 Chr.8: 6968709-6968709 on Build GRCh38
rs758681574 Chr.8: 6969053-6969053 on Build GRCh38
rs747778730 Chr.8: 6969142-6969142 on Build GRCh38
rs371996241 Chr.8: 6969083-6969083 on Build GRCh38
rs754444424 Chr.8: 6968940-6968940 on Build GRCh38
rs200129611 Chr.8: 6968958-6968959 on Build GRCh38
rs575029641 Chr.8: 6968891-6968891 on Build GRCh38
rs17078435 Chr.8: 6968768-6968768 on Build GRCh38
rs530341034 Chr.8: 6968704-6968704 on Build GRCh38
rs566332926 Chr.8: 6968870-6968870 on Build GRCh38
rs376366269 Chr.8: 6969037-6969037 on Build GRCh38
rs567535259 Chr.8: 6968812-6968812 on Build GRCh38
rs569422936 Chr.8: 6968782-6968782 on Build GRCh38
rs370379459 Chr.8: 6968846-6968846 on Build GRCh38
rs774693248 Chr.8: 6968871-6968871 on Build GRCh38
rs913881103 Chr.8: 6969168-6969168 on Build GRCh38
rs544364613 Chr.8: 6968904-6968904 on Build GRCh38
rs976058993 Chr.8: 6969161-6969161 on Build GRCh38
rs201826576 Chr.8: 6968860-6968860 on Build GRCh38
rs763214575 Chr.8: 6968792-6968792 on Build GRCh38
rs116156102 Chr.8: 6968886-6968886 on Build GRCh38
rs200616058 Chr.8: 6969043-6969043 on Build GRCh38
rs925446420 Chr.8: 6969008-6969008 on Build GRCh38
rs372952273 Chr.8: 6969120-6969120 on Build GRCh38
rs2702875 Chr.8: 6968722-6968722 on Build GRCh38
rs549378736 Chr.8: 6968683-6968683 on Build GRCh38
rs146865952 Chr.8: 6969042-6969042 on Build GRCh38
rs751956501 Chr.8: 6968848-6968848 on Build GRCh38
rs540579857 Chr.8: 6968930-6968930 on Build GRCh38
rs997119272 Chr.8: 6968667-6968667 on Build GRCh38

View Full Product Details