Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 7997082-7997627 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM66E
Entrez Gene ID
100132103
Gene Name
family with sequence similarity 66 member E
Gene Aliases
-
UniGene
Hs.694680
Gene Chromosome Location
Chr.8: 7955013-8008755 on Build GRCh38
Transcripts
NR_027424.1

SNP Detail

SNP ID Chromosome Location
rs201680181 Chr.8: 7997481-7997481 on Build GRCh38
rs982920870 Chr.8: 7997576-7997576 on Build GRCh38
rs565639441 Chr.8: 7997392-7997392 on Build GRCh38
rs910089579 Chr.8: 7997584-7997584 on Build GRCh38
rs199583630 Chr.8: 7997449-7997449 on Build GRCh38
rs577784752 Chr.8: 7997582-7997582 on Build GRCh38
rs200376913 Chr.8: 7997476-7997476 on Build GRCh38
rs534670907 Chr.8: 7997402-7997402 on Build GRCh38
rs199748836 Chr.8: 7997517-7997517 on Build GRCh38
rs557943843 Chr.8: 7997419-7997419 on Build GRCh38

View Full Product Details