Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.8: 39474984-39475532 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM3A
Entrez Gene ID
1587
Gene Name
ADAM metallopeptidase domain 3A (pseudogene)
Gene Aliases
ADAM3, CYRN1, tMDCI
UniGene
Hs.98848
Gene Chromosome Location
Chr.8: 39451045-39522989 on Build GRCh38
Transcripts
NR_001569.3 NR_024106.2 NR_024107.2 NR_073423.1

SNP Detail

SNP ID Chromosome Location
rs762716622 Chr.8: 39475466-39475466 on Build GRCh38
rs780444344 Chr.8: 39475377-39475377 on Build GRCh38
rs190673383 Chr.8: 39475148-39475148 on Build GRCh38
rs752909782 Chr.8: 39475061-39475061 on Build GRCh38
rs183262337 Chr.8: 39475179-39475179 on Build GRCh38
rs140887317 Chr.8: 39475445-39475445 on Build GRCh38
rs898771496 Chr.8: 39475028-39475028 on Build GRCh38
rs377522298 Chr.8: 39475359-39475359 on Build GRCh38
rs755224274 Chr.8: 39475384-39475385 on Build GRCh38
rs187977130 Chr.8: 39475245-39475245 on Build GRCh38
rs199887672 Chr.8: 39475376-39475377 on Build GRCh38
rs193129408 Chr.8: 39475253-39475253 on Build GRCh38
rs573701791 Chr.8: 39475424-39475424 on Build GRCh38
rs1002922487 Chr.8: 39475097-39475097 on Build GRCh38
rs887071553 Chr.8: 39475080-39475080 on Build GRCh38
rs73616437 Chr.8: 39475221-39475221 on Build GRCh38
rs1036158068 Chr.8: 39475501-39475501 on Build GRCh38
rs62510462 Chr.8: 39475389-39475389 on Build GRCh38
rs7840611 Chr.8: 39475365-39475365 on Build GRCh38
rs879754002 Chr.8: 39475190-39475190 on Build GRCh38
rs756459010 Chr.8: 39475268-39475268 on Build GRCh38
rs775738905 Chr.8: 39475384-39475384 on Build GRCh38
rs6997906 Chr.8: 39475051-39475051 on Build GRCh38
rs148021497 Chr.8: 39475423-39475423 on Build GRCh38
rs143575224 Chr.8: 39475330-39475330 on Build GRCh38
rs922193907 Chr.8: 39475050-39475050 on Build GRCh38
rs373273892 Chr.8: 39475376-39475376 on Build GRCh38
rs112427411 Chr.8: 39475509-39475509 on Build GRCh38
rs567691386 Chr.8: 39475092-39475092 on Build GRCh38

View Full Product Details