Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 81915266-81915809 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
SPATA31D5P
Entrez Gene ID
347127
Gene Name
SPATA31 subfamily D member 5, pseudogene
Gene Aliases
FAM75D5, FAM75D5P
UniGene
Hs.215235
Gene Chromosome Location
Chr.9: 81913437-81919927 on Build GRCh38
Transcripts
NR_026851.1

SNP Detail

SNP ID Chromosome Location
rs776880249 Chr.9: 81915642-81915642 on Build GRCh38
rs575529096 Chr.9: 81915313-81915313 on Build GRCh38
rs148643857 Chr.9: 81915295-81915295 on Build GRCh38
rs1053742975 Chr.9: 81915318-81915318 on Build GRCh38
rs752023785 Chr.9: 81915439-81915439 on Build GRCh38
rs879044748 Chr.9: 81915336-81915336 on Build GRCh38
rs751743931 Chr.9: 81915655-81915655 on Build GRCh38
rs766400368 Chr.9: 81915705-81915705 on Build GRCh38
rs952507796 Chr.9: 81915409-81915409 on Build GRCh38
rs565451825 Chr.9: 81915422-81915422 on Build GRCh38
rs779740992 Chr.9: 81915683-81915683 on Build GRCh38
rs760482246 Chr.9: 81915653-81915653 on Build GRCh38
rs41277923 Chr.9: 81915481-81915481 on Build GRCh38
rs773529388 Chr.9: 81915698-81915698 on Build GRCh38
rs763296819 Chr.9: 81915419-81915419 on Build GRCh38
rs762356383 Chr.9: 81915644-81915644 on Build GRCh38
rs780761912 Chr.9: 81915759-81915759 on Build GRCh38
rs1043242176 Chr.9: 81915299-81915299 on Build GRCh38
rs576614147 Chr.9: 81915527-81915527 on Build GRCh38
rs999033956 Chr.9: 81915317-81915317 on Build GRCh38
rs1048075426 Chr.9: 81915661-81915661 on Build GRCh38
rs751199388 Chr.9: 81915493-81915494 on Build GRCh38
rs764751720 Chr.9: 81915427-81915427 on Build GRCh38
rs756308814 Chr.9: 81915619-81915619 on Build GRCh38
rs1015451228 Chr.9: 81915574-81915574 on Build GRCh38
rs780228207 Chr.9: 81915438-81915438 on Build GRCh38
rs551294860 Chr.9: 81915679-81915679 on Build GRCh38
rs766119712 Chr.9: 81915692-81915692 on Build GRCh38
rs561216576 Chr.9: 81915342-81915342 on Build GRCh38
rs765826324 Chr.9: 81915533-81915533 on Build GRCh38
rs749445927 Chr.9: 81915502-81915502 on Build GRCh38
rs532748157 Chr.9: 81915674-81915674 on Build GRCh38
rs111520975 Chr.9: 81915561-81915561 on Build GRCh38
rs745919664 Chr.9: 81915593-81915593 on Build GRCh38
rs568879206 Chr.9: 81915462-81915462 on Build GRCh38
rs762584759 Chr.9: 81915465-81915465 on Build GRCh38
rs770115800 Chr.9: 81915407-81915407 on Build GRCh38
rs770405496 Chr.9: 81915584-81915584 on Build GRCh38
rs774013105 Chr.9: 81915542-81915542 on Build GRCh38
rs778455631 Chr.9: 81915627-81915627 on Build GRCh38
rs780261598 Chr.9: 81915684-81915684 on Build GRCh38
rs888171866 Chr.9: 81915681-81915681 on Build GRCh38
rs771836997 Chr.9: 81915633-81915633 on Build GRCh38
rs749775510 Chr.9: 81915789-81915789 on Build GRCh38
rs142129625 Chr.9: 81915404-81915404 on Build GRCh38
rs113953702 Chr.9: 81915425-81915425 on Build GRCh38
rs764158764 Chr.9: 81915424-81915424 on Build GRCh38
rs758431132 Chr.9: 81915747-81915747 on Build GRCh38
rs770317595 Chr.9: 81915695-81915695 on Build GRCh38
rs747773761 Chr.9: 81915352-81915352 on Build GRCh38
rs766090941 Chr.9: 81915470-81915470 on Build GRCh38
rs757390502 Chr.9: 81915510-81915510 on Build GRCh38
rs778003266 Chr.9: 81915446-81915446 on Build GRCh38
rs763485343 Chr.9: 81915703-81915703 on Build GRCh38
rs543154980 Chr.9: 81915335-81915335 on Build GRCh38
rs755404280 Chr.9: 81915440-81915440 on Build GRCh38
rs771818210 Chr.9: 81915379-81915379 on Build GRCh38
rs561222260 Chr.9: 81915616-81915616 on Build GRCh38
rs754328580 Chr.9: 81915596-81915596 on Build GRCh38
rs749430908 Chr.9: 81915331-81915331 on Build GRCh38
rs565505555 Chr.9: 81915670-81915670 on Build GRCh38
rs765083746 Chr.9: 81915735-81915735 on Build GRCh38
rs535859898 Chr.9: 81915464-81915464 on Build GRCh38
rs756899445 Chr.9: 81915349-81915349 on Build GRCh38
rs769101346 Chr.9: 81915638-81915638 on Build GRCh38
rs764895235 Chr.9: 81915378-81915378 on Build GRCh38
rs890733845 Chr.9: 81915338-81915338 on Build GRCh38
rs761363014 Chr.9: 81915426-81915426 on Build GRCh38
rs540698411 Chr.9: 81915665-81915665 on Build GRCh38
rs762495611 Chr.9: 81915530-81915530 on Build GRCh38
rs761771998 Chr.9: 81915458-81915458 on Build GRCh38
rs764583378 Chr.9: 81915492-81915493 on Build GRCh38
rs904724560 Chr.9: 81915302-81915302 on Build GRCh38
rs778578855 Chr.9: 81915360-81915360 on Build GRCh38
rs764738567 Chr.9: 81915553-81915553 on Build GRCh38
rs780502751 Chr.9: 81915392-81915392 on Build GRCh38
rs748684616 Chr.9: 81915445-81915445 on Build GRCh38
rs751706319 Chr.9: 81915611-81915611 on Build GRCh38
rs770018130 Chr.9: 81915765-81915765 on Build GRCh38
rs888111165 Chr.9: 81915334-81915334 on Build GRCh38
rs749202753 Chr.9: 81915618-81915618 on Build GRCh38
rs768457145 Chr.9: 81915456-81915456 on Build GRCh38
rs758054172 Chr.9: 81915434-81915434 on Build GRCh38
rs878982799 Chr.9: 81915614-81915614 on Build GRCh38
rs754669731 Chr.9: 81915755-81915755 on Build GRCh38
rs191988835 Chr.9: 81915587-81915587 on Build GRCh38
rs114426311 Chr.9: 81915583-81915583 on Build GRCh38
rs759725537 Chr.9: 81915707-81915707 on Build GRCh38
rs757311982 Chr.9: 81915560-81915560 on Build GRCh38
rs756847592 Chr.9: 81915614-81915615 on Build GRCh38
rs148326094 Chr.9: 81915720-81915722 on Build GRCh38
rs748007109 Chr.9: 81915575-81915575 on Build GRCh38
rs12345231 Chr.9: 81915451-81915451 on Build GRCh38
rs748023470 Chr.9: 81915523-81915523 on Build GRCh38
rs769223370 Chr.9: 81915400-81915400 on Build GRCh38
rs746554663 Chr.9: 81915753-81915753 on Build GRCh38
rs760407386 Chr.9: 81915636-81915636 on Build GRCh38
rs530371850 Chr.9: 81915748-81915748 on Build GRCh38
rs776418015 Chr.9: 81915516-81915516 on Build GRCh38
rs558344720 Chr.9: 81915513-81915513 on Build GRCh38
rs751844884 Chr.9: 81915473-81915473 on Build GRCh38
rs773745882 Chr.9: 81915585-81915585 on Build GRCh38
rs758612550 Chr.9: 81915607-81915607 on Build GRCh38
rs772709169 Chr.9: 81915528-81915528 on Build GRCh38
rs12345266 Chr.9: 81915497-81915497 on Build GRCh38
rs751797182 Chr.9: 81915613-81915613 on Build GRCh38
rs761178656 Chr.9: 81915594-81915594 on Build GRCh38
rs750514477 Chr.9: 81915600-81915600 on Build GRCh38
rs774465586 Chr.9: 81915706-81915706 on Build GRCh38
rs753945846 Chr.9: 81915555-81915555 on Build GRCh38
rs746609716 Chr.9: 81915689-81915689 on Build GRCh38
rs764664338 Chr.9: 81915461-81915461 on Build GRCh38
rs776584180 Chr.9: 81915457-81915457 on Build GRCh38
rs745437063 Chr.9: 81915631-81915631 on Build GRCh38
rs528558233 Chr.9: 81915399-81915399 on Build GRCh38
rs756943919 Chr.9: 81915732-81915732 on Build GRCh38
rs768460499 Chr.9: 81915515-81915515 on Build GRCh38
rs750040583 Chr.9: 81915432-81915432 on Build GRCh38
rs573398951 Chr.9: 81915651-81915651 on Build GRCh38
rs778773841 Chr.9: 81915676-81915676 on Build GRCh38
rs746102305 Chr.9: 81915514-81915514 on Build GRCh38
rs538169896 Chr.9: 81915552-81915552 on Build GRCh38
rs756701930 Chr.9: 81915489-81915489 on Build GRCh38
rs183317604 Chr.9: 81915723-81915723 on Build GRCh38
rs777523437 Chr.9: 81915656-81915656 on Build GRCh38
rs777020378 Chr.9: 81915693-81915693 on Build GRCh38
rs200441822 Chr.9: 81915403-81915403 on Build GRCh38
rs767863889 Chr.9: 81915547-81915547 on Build GRCh38
rs775225132 Chr.9: 81915635-81915635 on Build GRCh38
rs1004323299 Chr.9: 81915340-81915340 on Build GRCh38
rs775143248 Chr.9: 81915453-81915453 on Build GRCh38
rs867030616 Chr.9: 81915406-81915406 on Build GRCh38
rs753186116 Chr.9: 81915487-81915487 on Build GRCh38
rs770824181 Chr.9: 81915623-81915623 on Build GRCh38
rs1041108274 Chr.9: 81915521-81915521 on Build GRCh38
rs747844645 Chr.9: 81915762-81915762 on Build GRCh38
rs915982428 Chr.9: 81915292-81915292 on Build GRCh38
rs781745430 Chr.9: 81915444-81915444 on Build GRCh38
rs758564925 Chr.9: 81915563-81915563 on Build GRCh38
rs750286121 Chr.9: 81915746-81915746 on Build GRCh38
rs759113454 Chr.9: 81915546-81915546 on Build GRCh38
rs753616426 Chr.9: 81915673-81915673 on Build GRCh38
rs750507503 Chr.9: 81915652-81915652 on Build GRCh38
rs773516557 Chr.9: 81915410-81915410 on Build GRCh38
rs777289328 Chr.9: 81915421-81915421 on Build GRCh38
rs542667244 Chr.9: 81915595-81915595 on Build GRCh38
rs773650680 Chr.9: 81915774-81915774 on Build GRCh38
rs746861877 Chr.9: 81915454-81915454 on Build GRCh38
rs199691053 Chr.9: 81915474-81915474 on Build GRCh38
rs767700240 Chr.9: 81915712-81915712 on Build GRCh38

View Full Product Details