Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 87773295-87773842 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTSL3P
Entrez Gene ID
392360
Gene Name
cathepsin L family member 3, pseudogene
Gene Aliases
CTSL3, HCTSL-s
UniGene
Hs.690450
Gene Chromosome Location
Chr.9: 87772915-87786884 on Build GRCh38
Transcripts
NR_027917.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6116686 c.377C>T p.P126L Chr.9: 87773596-87773596 on Build GRCh38
6637830 c.407C>A p.P136H Chr.9: 87773626-87773626 on Build GRCh38
4419440 c.236G>A p.S79N Chr.9: 87773455-87773455 on Build GRCh38
5619483 c.310G>T p.A104S Chr.9: 87773529-87773529 on Build GRCh38
7162388 c.393G>A p.W131* Chr.9: 87773612-87773612 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs777532670 Chr.9: 87773610-87773610 on Build GRCh38
rs780639148 Chr.9: 87773604-87773604 on Build GRCh38
rs556650553 Chr.9: 87773359-87773359 on Build GRCh38
rs116991499 Chr.9: 87773711-87773711 on Build GRCh38
rs377521277 Chr.9: 87773818-87773818 on Build GRCh38
rs532125948 Chr.9: 87773589-87773589 on Build GRCh38
rs562980208 Chr.9: 87773597-87773597 on Build GRCh38
rs746897463 Chr.9: 87773499-87773499 on Build GRCh38
rs139706141 Chr.9: 87773625-87773625 on Build GRCh38
rs185179522 Chr.9: 87773452-87773452 on Build GRCh38
rs1026200810 Chr.9: 87773337-87773337 on Build GRCh38
rs759664428 Chr.9: 87773435-87773435 on Build GRCh38
rs772703694 Chr.9: 87773526-87773526 on Build GRCh38
rs143676137 Chr.9: 87773510-87773510 on Build GRCh38
rs964749388 Chr.9: 87773700-87773700 on Build GRCh38
rs149566005 Chr.9: 87773666-87773666 on Build GRCh38
rs764407906 Chr.9: 87773584-87773584 on Build GRCh38
rs796501947 Chr.9: 87773489-87773490 on Build GRCh38
rs746171376 Chr.9: 87773618-87773618 on Build GRCh38
rs767639591 Chr.9: 87773554-87773554 on Build GRCh38
rs933144502 Chr.9: 87773474-87773474 on Build GRCh38
rs540099375 Chr.9: 87773557-87773557 on Build GRCh38
rs11141967 Chr.9: 87773586-87773586 on Build GRCh38
rs933203910 Chr.9: 87773477-87773477 on Build GRCh38
rs977958795 Chr.9: 87773481-87773481 on Build GRCh38
rs936494714 Chr.9: 87773745-87773745 on Build GRCh38
rs746197404 Chr.9: 87773587-87773587 on Build GRCh38
rs546889225 Chr.9: 87773561-87773561 on Build GRCh38
rs544108619 Chr.9: 87773332-87773332 on Build GRCh38
rs7853691 Chr.9: 87773455-87773455 on Build GRCh38
rs577480968 Chr.9: 87773323-87773323 on Build GRCh38
rs148843343 Chr.9: 87773453-87773453 on Build GRCh38
rs750236294 Chr.9: 87773456-87773456 on Build GRCh38
rs766945485 Chr.9: 87773420-87773420 on Build GRCh38
rs773640268 Chr.9: 87773416-87773416 on Build GRCh38
rs76207551 Chr.9: 87773701-87773701 on Build GRCh38
rs375477232 Chr.9: 87773696-87773696 on Build GRCh38
rs761928253 Chr.9: 87773626-87773626 on Build GRCh38
rs764868788 Chr.9: 87773410-87773412 on Build GRCh38
rs555129058 Chr.9: 87773590-87773590 on Build GRCh38
rs771045827 Chr.9: 87773655-87773656 on Build GRCh38
rs7853247 Chr.9: 87773355-87773355 on Build GRCh38
rs965010209 Chr.9: 87773398-87773398 on Build GRCh38
rs760030025 Chr.9: 87773545-87773545 on Build GRCh38
rs879268014 Chr.9: 87773720-87773720 on Build GRCh38
rs138191329 Chr.9: 87773596-87773596 on Build GRCh38
rs78381498 Chr.9: 87773502-87773502 on Build GRCh38
rs762599964 Chr.9: 87773478-87773478 on Build GRCh38
rs773326904 Chr.9: 87773810-87773810 on Build GRCh38
rs754782123 Chr.9: 87773648-87773648 on Build GRCh38
rs760678118 Chr.9: 87773573-87773573 on Build GRCh38
rs989212688 Chr.9: 87773770-87773770 on Build GRCh38
rs371964987 Chr.9: 87773601-87773601 on Build GRCh38
rs769576125 Chr.9: 87773511-87773511 on Build GRCh38
rs761907444 Chr.9: 87773796-87773796 on Build GRCh38
rs983313914 Chr.9: 87773714-87773715 on Build GRCh38
rs771179926 Chr.9: 87773534-87773534 on Build GRCh38
rs758088012 Chr.9: 87773478-87773478 on Build GRCh38
rs763402315 Chr.9: 87773527-87773527 on Build GRCh38
rs747694952 Chr.9: 87773599-87773599 on Build GRCh38
rs144380408 Chr.9: 87773783-87773783 on Build GRCh38
rs751964172 Chr.9: 87773427-87773427 on Build GRCh38
rs750519217 Chr.9: 87773489-87773490 on Build GRCh38
rs767588791 Chr.9: 87773651-87773651 on Build GRCh38
rs1037798752 Chr.9: 87773518-87773518 on Build GRCh38
rs376285822 Chr.9: 87773630-87773630 on Build GRCh38
rs771390679 Chr.9: 87773614-87773614 on Build GRCh38
rs141361280 Chr.9: 87773619-87773619 on Build GRCh38
rs571060820 Chr.9: 87773817-87773817 on Build GRCh38
rs923730338 Chr.9: 87773490-87773490 on Build GRCh38
rs376833660 Chr.9: 87773492-87773492 on Build GRCh38
rs772294118 Chr.9: 87773806-87773806 on Build GRCh38
rs751407414 Chr.9: 87773634-87773634 on Build GRCh38
rs1018321769 Chr.9: 87773418-87773418 on Build GRCh38
rs753784622 Chr.9: 87773399-87773399 on Build GRCh38
rs878860216 Chr.9: 87773635-87773635 on Build GRCh38
rs752662474 Chr.9: 87773662-87773662 on Build GRCh38
rs780837365 Chr.9: 87773505-87773505 on Build GRCh38
rs774964998 Chr.9: 87773544-87773544 on Build GRCh38
rs371652926 Chr.9: 87773417-87773417 on Build GRCh38
rs10735592 Chr.9: 87773624-87773624 on Build GRCh38
rs767551464 Chr.9: 87773443-87773443 on Build GRCh38

View Full Product Details