Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.9: 121454641-121455194 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GGTA1P
Entrez Gene ID
2681
Gene Name
glycoprotein, alpha-galactosyltransferase 1 pseudogene
Gene Aliases
GGTA, GGTA1, GLYT2, a1/3GTP
UniGene
Hs.97469
Gene Chromosome Location
Chr.9: 121455041-121500027 on Build GRCh38
Transcripts
NR_003191.1 NR_045211.1

SNP Detail

SNP ID Chromosome Location
rs944366413 Chr.9: 121454968-121454968 on Build GRCh38
rs1014250817 Chr.9: 121455126-121455126 on Build GRCh38
rs1052762854 Chr.9: 121454674-121454674 on Build GRCh38
rs560249309 Chr.9: 121454831-121454831 on Build GRCh38
rs527975805 Chr.9: 121454739-121454739 on Build GRCh38
rs760112805 Chr.9: 121454971-121454971 on Build GRCh38
rs892587362 Chr.9: 121454959-121454959 on Build GRCh38
rs140684846 Chr.9: 121454949-121454949 on Build GRCh38
rs569246743 Chr.9: 121454970-121454970 on Build GRCh38
rs368824787 Chr.9: 121455027-121455030 on Build GRCh38
rs192799497 Chr.9: 121454828-121454828 on Build GRCh38
rs1042727366 Chr.9: 121454904-121454904 on Build GRCh38
rs199936104 Chr.9: 121455016-121455019 on Build GRCh38
rs961769515 Chr.9: 121454801-121454801 on Build GRCh38
rs183080538 Chr.9: 121454860-121454860 on Build GRCh38
rs1016340888 Chr.9: 121454750-121454750 on Build GRCh38
rs534181172 Chr.9: 121454693-121454693 on Build GRCh38
rs1015556045 Chr.9: 121454756-121454756 on Build GRCh38
rs1002194641 Chr.9: 121455137-121455137 on Build GRCh38
rs551593655 Chr.9: 121455014-121455014 on Build GRCh38
rs188408237 Chr.9: 121455085-121455085 on Build GRCh38
rs534145003 Chr.9: 121455115-121455115 on Build GRCh38
rs1040091104 Chr.9: 121454738-121454738 on Build GRCh38
rs972188905 Chr.9: 121454786-121454786 on Build GRCh38
rs528413258 Chr.9: 121454946-121454946 on Build GRCh38
rs892646625 Chr.9: 121454687-121454687 on Build GRCh38
rs1024407384 Chr.9: 121455153-121455153 on Build GRCh38
rs888678436 Chr.9: 121454742-121454742 on Build GRCh38
rs532107624 Chr.9: 121454977-121454977 on Build GRCh38
rs549266657 Chr.9: 121454922-121454922 on Build GRCh38
rs564324030 Chr.9: 121454945-121454945 on Build GRCh38
rs970106523 Chr.9: 121455155-121455155 on Build GRCh38
rs912973266 Chr.9: 121454695-121454695 on Build GRCh38
rs933171580 Chr.9: 121454851-121454851 on Build GRCh38
rs113633580 Chr.9: 121454727-121454727 on Build GRCh38
rs144158349 Chr.9: 121454740-121454740 on Build GRCh38
rs919062034 Chr.9: 121454829-121454829 on Build GRCh38
rs775086173 Chr.9: 121455045-121455045 on Build GRCh38
rs550991553 Chr.9: 121454969-121454969 on Build GRCh38
rs4527952 Chr.9: 121454755-121454755 on Build GRCh38
rs533415176 Chr.9: 121455010-121455010 on Build GRCh38
rs1013759979 Chr.9: 121455055-121455055 on Build GRCh38
rs10818539 Chr.9: 121454735-121454735 on Build GRCh38
rs979902473 Chr.9: 121454906-121454906 on Build GRCh38
rs1014383331 Chr.9: 121454814-121454814 on Build GRCh38
rs773513297 Chr.9: 121454741-121454741 on Build GRCh38
rs1005768368 Chr.9: 121455058-121455058 on Build GRCh38
rs1025226241 Chr.9: 121454737-121454737 on Build GRCh38
rs555655505 Chr.9: 121454728-121454728 on Build GRCh38
rs768141168 Chr.9: 121455032-121455032 on Build GRCh38
rs144597869 Chr.9: 121455117-121455117 on Build GRCh38
rs925879900 Chr.9: 121454907-121454907 on Build GRCh38
rs756291433 Chr.9: 121454878-121454878 on Build GRCh38

View Full Product Details