Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.X: 15675565-15676113 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CA5BP1
Entrez Gene ID
340591
Gene Name
carbonic anhydrase 5B pseudogene 1
Gene Aliases
CA5BL, CA5BP, PRO2325
UniGene
Hs.732244
Gene Chromosome Location
Chr.X: 15674916-15703351 on Build GRCh38
Transcripts
NR_026551.1 XM_017029680.1

Gene Symbol
TMEM27
Entrez Gene ID
57393
Gene Name
transmembrane protein 27
Gene Aliases
NX-17, NX17
UniGene
Hs.129614
Gene Chromosome Location
Chr.X: 15627316-15675624 on Build GRCh38
Transcripts
NR_026551.1 XM_017029680.1

SNP Detail

SNP ID Chromosome Location
rs775351095 Chr.X: 15675923-15675923 on Build GRCh38
rs12012790 Chr.X: 15675988-15675988 on Build GRCh38
rs190788254 Chr.X: 15675750-15675750 on Build GRCh38
rs939252810 Chr.X: 15675795-15675795 on Build GRCh38
rs774830625 Chr.X: 15675751-15675751 on Build GRCh38
rs911630826 Chr.X: 15675883-15675883 on Build GRCh38
rs999523233 Chr.X: 15675849-15675849 on Build GRCh38
rs759182437 Chr.X: 15675736-15675736 on Build GRCh38
rs984293837 Chr.X: 15675761-15675761 on Build GRCh38
rs181705304 Chr.X: 15675774-15675774 on Build GRCh38
rs768797223 Chr.X: 15675603-15675603 on Build GRCh38
rs946577737 Chr.X: 15676041-15676041 on Build GRCh38
rs752474319 Chr.X: 15676063-15676063 on Build GRCh38
rs766424661 Chr.X: 15675851-15675851 on Build GRCh38
rs966265960 Chr.X: 15675719-15675719 on Build GRCh38
rs927905220 Chr.X: 15675792-15675792 on Build GRCh38
rs952334219 Chr.X: 15675932-15675932 on Build GRCh38
rs972437930 Chr.X: 15675963-15675963 on Build GRCh38
rs972382956 Chr.X: 15675783-15675783 on Build GRCh38
rs773987377 Chr.X: 15675819-15675819 on Build GRCh38
rs1004216122 Chr.X: 15676017-15676017 on Build GRCh38
rs1043565956 Chr.X: 15676043-15676043 on Build GRCh38
rs1045263195 Chr.X: 15675676-15675676 on Build GRCh38
rs1055019692 Chr.X: 15675803-15675803 on Build GRCh38
rs768125180 Chr.X: 15675808-15675808 on Build GRCh38
rs915165301 Chr.X: 15675939-15675939 on Build GRCh38
rs984962220 Chr.X: 15675937-15675937 on Build GRCh38
rs916489923 Chr.X: 15676004-15676004 on Build GRCh38
rs938976433 Chr.X: 15675976-15675976 on Build GRCh38
rs5934263 Chr.X: 15675734-15675734 on Build GRCh38
rs368752988 Chr.X: 15675766-15675766 on Build GRCh38
rs760187457 Chr.X: 15675972-15675972 on Build GRCh38
rs893709564 Chr.X: 15675654-15675654 on Build GRCh38
rs1056595525 Chr.X: 15675983-15675983 on Build GRCh38
rs997294 Chr.X: 15676066-15676066 on Build GRCh38
rs997482248 Chr.X: 15676069-15676071 on Build GRCh38
rs781568809 Chr.X: 15675729-15675729 on Build GRCh38
rs1011421220 Chr.X: 15675588-15675588 on Build GRCh38
rs963246209 Chr.X: 15675769-15675769 on Build GRCh38
rs905284519 Chr.X: 15675722-15675722 on Build GRCh38
rs754037565 Chr.X: 15675875-15675875 on Build GRCh38
rs935034827 Chr.X: 15675891-15675891 on Build GRCh38
rs961113251 Chr.X: 15675957-15675957 on Build GRCh38
rs746492947 Chr.X: 15675648-15675648 on Build GRCh38
rs770403557 Chr.X: 15675673-15675673 on Build GRCh38
rs1010779694 Chr.X: 15675666-15675666 on Build GRCh38
rs184629340 Chr.X: 15675817-15675817 on Build GRCh38
rs1035083721 Chr.X: 15675748-15675748 on Build GRCh38
rs777688651 Chr.X: 15676083-15676083 on Build GRCh38
rs1029121951 Chr.X: 15675741-15675741 on Build GRCh38
rs901820438 Chr.X: 15675864-15675864 on Build GRCh38
rs1044919628 Chr.X: 15675949-15675949 on Build GRCh38
rs750597832 Chr.X: 15675812-15675812 on Build GRCh38
rs1017312282 Chr.X: 15676020-15676020 on Build GRCh38
rs1019192405 Chr.X: 15675679-15675679 on Build GRCh38
rs894990736 Chr.X: 15675804-15675804 on Build GRCh38
rs999470986 Chr.X: 15675675-15675675 on Build GRCh38
rs772766394 Chr.X: 15675597-15675598 on Build GRCh38
rs755194608 Chr.X: 15675942-15675942 on Build GRCh38

View Full Product Details