Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 16492420-16493009 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CROCCP3
Entrez Gene ID
114819
Gene Name
ciliary rootlet coiled-coil, rootletin pseudogene 3
Gene Aliases
CROCCL2, dJ798A10.2
UniGene
Hs.597881
Gene Chromosome Location
Chr.1: 16467436-16492701 on Build GRCh38
Transcripts
NR_023386.1

SNP Detail

SNP ID Chromosome Location
rs540501740 Chr.1: 16492486-16492486 on Build GRCh38
rs377399610 Chr.1: 16492611-16492611 on Build GRCh38
rs903226823 Chr.1: 16492884-16492884 on Build GRCh38
rs149041440 Chr.1: 16492627-16492627 on Build GRCh38
rs769370747 Chr.1: 16492958-16492959 on Build GRCh38
rs370350350 Chr.1: 16492908-16492908 on Build GRCh38
rs143072971 Chr.1: 16492804-16492804 on Build GRCh38
rs922470801 Chr.1: 16492782-16492782 on Build GRCh38
rs369519439 Chr.1: 16492649-16492649 on Build GRCh38
rs976545151 Chr.1: 16492781-16492781 on Build GRCh38
rs531279621 Chr.1: 16492719-16492719 on Build GRCh38
rs753195704 Chr.1: 16492521-16492521 on Build GRCh38
rs908257734 Chr.1: 16492771-16492771 on Build GRCh38
rs774731028 Chr.1: 16492938-16492938 on Build GRCh38
rs571249825 Chr.1: 16492738-16492738 on Build GRCh38
rs537932675 Chr.1: 16492970-16492970 on Build GRCh38
rs556054058 Chr.1: 16492879-16492879 on Build GRCh38
rs182476062 Chr.1: 16492861-16492861 on Build GRCh38
rs932560171 Chr.1: 16492505-16492505 on Build GRCh38
rs1050415976 Chr.1: 16492513-16492513 on Build GRCh38
rs1043075318 Chr.1: 16492883-16492883 on Build GRCh38
rs768979445 Chr.1: 16492729-16492729 on Build GRCh38
rs547713442 Chr.1: 16492779-16492779 on Build GRCh38
rs749519477 Chr.1: 16492595-16492596 on Build GRCh38
rs36084231 Chr.1: 16492596-16492596 on Build GRCh38
rs1040894525 Chr.1: 16492454-16492454 on Build GRCh38
rs939823657 Chr.1: 16492445-16492445 on Build GRCh38
rs896015636 Chr.1: 16492926-16492926 on Build GRCh38
rs1057220172 Chr.1: 16492912-16492912 on Build GRCh38
rs551470887 Chr.1: 16492723-16492723 on Build GRCh38
rs952505025 Chr.1: 16492756-16492756 on Build GRCh38
rs549271582 Chr.1: 16492678-16492678 on Build GRCh38
rs755751350 Chr.1: 16492876-16492876 on Build GRCh38
rs910538035 Chr.1: 16492878-16492878 on Build GRCh38
rs756815551 Chr.1: 16492560-16492560 on Build GRCh38
rs1049703540 Chr.1: 16492833-16492833 on Build GRCh38
rs901058573 Chr.1: 16492488-16492488 on Build GRCh38
rs138769290 Chr.1: 16492885-16492885 on Build GRCh38
rs374219619 Chr.1: 16492703-16492703 on Build GRCh38
rs376547125 Chr.1: 16492770-16492770 on Build GRCh38
rs960277543 Chr.1: 16492657-16492657 on Build GRCh38
rs1022151683 Chr.1: 16492524-16492524 on Build GRCh38

View Full Product Details