Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 45499405-45499949 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC163P
Entrez Gene ID
126661
Gene Name
coiled-coil domain containing 163, pseudogene
Gene Aliases
C1orf231
UniGene
Hs.461300
Gene Chromosome Location
Chr.1: 45493926-45500079 on Build GRCh38
Transcripts
NR_033296.1 NR_033297.1 NR_033298.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
4772347 c.20G>T p.W7L Chr.1: 45499590-45499590 on Build GRCh38
4772346 c.20G>T p.W7L Chr.1: 45499590-45499590 on Build GRCh38
6278410 c.112C>T p.L38F Chr.1: 45499410-45499410 on Build GRCh38
6278409 c.112C>T p.L38F Chr.1: 45499410-45499410 on Build GRCh38
7359838 c.70C>T p.R24W Chr.1: 45499540-45499540 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs753070176 Chr.1: 45499579-45499579 on Build GRCh38
rs12568145 Chr.1: 45499648-45499648 on Build GRCh38
rs541561405 Chr.1: 45499675-45499675 on Build GRCh38
rs552146790 Chr.1: 45499858-45499858 on Build GRCh38
rs369501552 Chr.1: 45499453-45499453 on Build GRCh38
rs781437428 Chr.1: 45499464-45499464 on Build GRCh38
rs868240442 Chr.1: 45499427-45499427 on Build GRCh38
rs756425361 Chr.1: 45499510-45499510 on Build GRCh38
rs560617593 Chr.1: 45499601-45499601 on Build GRCh38
rs769383879 Chr.1: 45499628-45499628 on Build GRCh38
rs760511046 Chr.1: 45499539-45499539 on Build GRCh38
rs772327250 Chr.1: 45499806-45499806 on Build GRCh38
rs182868907 Chr.1: 45499692-45499692 on Build GRCh38
rs368386391 Chr.1: 45499615-45499615 on Build GRCh38
rs1026716897 Chr.1: 45499818-45499818 on Build GRCh38
rs775151482 Chr.1: 45499536-45499536 on Build GRCh38
rs543819205 Chr.1: 45499708-45499708 on Build GRCh38
rs973847607 Chr.1: 45499501-45499501 on Build GRCh38
rs754083314 Chr.1: 45499587-45499587 on Build GRCh38
rs778052667 Chr.1: 45499498-45499498 on Build GRCh38
rs746824188 Chr.1: 45499621-45499621 on Build GRCh38
rs759978591 Chr.1: 45499562-45499562 on Build GRCh38
rs760258542 Chr.1: 45499439-45499439 on Build GRCh38
rs749439631 Chr.1: 45499500-45499500 on Build GRCh38
rs776241361 Chr.1: 45499552-45499552 on Build GRCh38
rs930349408 Chr.1: 45499646-45499646 on Build GRCh38
rs974687736 Chr.1: 45499482-45499482 on Build GRCh38
rs764573291 Chr.1: 45499652-45499652 on Build GRCh38
rs7546268 Chr.1: 45499668-45499668 on Build GRCh38
rs561334431 Chr.1: 45499676-45499676 on Build GRCh38
rs772717347 Chr.1: 45499632-45499632 on Build GRCh38
rs969506534 Chr.1: 45499495-45499495 on Build GRCh38
rs553802345 Chr.1: 45499761-45499761 on Build GRCh38
rs983692811 Chr.1: 45499549-45499549 on Build GRCh38
rs768004897 Chr.1: 45499564-45499564 on Build GRCh38
rs754195638 Chr.1: 45499653-45499653 on Build GRCh38
rs546826001 Chr.1: 45499822-45499822 on Build GRCh38
rs1049283297 Chr.1: 45499647-45499647 on Build GRCh38
rs554525203 Chr.1: 45499671-45499671 on Build GRCh38
rs559118770 Chr.1: 45499663-45499663 on Build GRCh38
rs1040737349 Chr.1: 45499592-45499592 on Build GRCh38
rs767138648 Chr.1: 45499432-45499432 on Build GRCh38
rs534420690 Chr.1: 45499488-45499488 on Build GRCh38
rs761350578 Chr.1: 45499553-45499553 on Build GRCh38
rs201193008 Chr.1: 45499540-45499540 on Build GRCh38
rs532234765 Chr.1: 45499428-45499428 on Build GRCh38
rs375188001 Chr.1: 45499452-45499452 on Build GRCh38
rs966305099 Chr.1: 45499883-45499883 on Build GRCh38
rs979238647 Chr.1: 45499516-45499516 on Build GRCh38
rs753408882 Chr.1: 45499442-45499442 on Build GRCh38
rs777844678 Chr.1: 45499850-45499850 on Build GRCh38
rs776044639 Chr.1: 45499642-45499642 on Build GRCh38
rs369887445 Chr.1: 45499583-45499583 on Build GRCh38
rs771987719 Chr.1: 45499532-45499532 on Build GRCh38
rs755473389 Chr.1: 45499455-45499455 on Build GRCh38
rs996464800 Chr.1: 45499730-45499730 on Build GRCh38
rs12568144 Chr.1: 45499635-45499635 on Build GRCh38
rs567739155 Chr.1: 45499558-45499558 on Build GRCh38
rs376618650 Chr.1: 45499840-45499840 on Build GRCh38
rs200271659 Chr.1: 45499516-45499516 on Build GRCh38
rs758333387 Chr.1: 45499612-45499612 on Build GRCh38
rs373270668 Chr.1: 45499913-45499913 on Build GRCh38
rs991797852 Chr.1: 45499484-45499484 on Build GRCh38
rs200044008 Chr.1: 45499430-45499430 on Build GRCh38
rs565678054 Chr.1: 45499434-45499434 on Build GRCh38
rs763735331 Chr.1: 45499441-45499441 on Build GRCh38
rs1013262048 Chr.1: 45499700-45499700 on Build GRCh38
rs976353286 Chr.1: 45499893-45499893 on Build GRCh38
rs376450068 Chr.1: 45499627-45499627 on Build GRCh38
rs745876909 Chr.1: 45499611-45499611 on Build GRCh38
rs548479868 Chr.1: 45499474-45499474 on Build GRCh38
rs762520120 Chr.1: 45499634-45499634 on Build GRCh38
rs952232278 Chr.1: 45499824-45499824 on Build GRCh38
rs754164416 Chr.1: 45499561-45499561 on Build GRCh38
rs1023762253 Chr.1: 45499725-45499725 on Build GRCh38
rs1018748582 Chr.1: 45499695-45499695 on Build GRCh38
rs965915807 Chr.1: 45499473-45499473 on Build GRCh38
rs934824028 Chr.1: 45499559-45499559 on Build GRCh38
rs999333226 Chr.1: 45499655-45499655 on Build GRCh38
rs371947887 Chr.1: 45499521-45499521 on Build GRCh38
rs762454996 Chr.1: 45499615-45499625 on Build GRCh38
rs756488005 Chr.1: 45499580-45499580 on Build GRCh38
rs969075107 Chr.1: 45499777-45499777 on Build GRCh38
rs536294878 Chr.1: 45499754-45499754 on Build GRCh38
rs188374041 Chr.1: 45499805-45499805 on Build GRCh38
rs556610850 Chr.1: 45499625-45499625 on Build GRCh38
rs772664925 Chr.1: 45499575-45499582 on Build GRCh38
rs779146566 Chr.1: 45499609-45499609 on Build GRCh38

View Full Product Details