Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 63549478-63550030 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEU2L
Entrez Gene ID
79469
Gene Name
deleted in lymphocytic leukemia 2-like
Gene Aliases
BCMSUNL
UniGene
Hs.659291
Gene Chromosome Location
Chr.1: 63548980-63550636 on Build GRCh38
Transcripts
NR_002771.1 NM_032437.3 XM_005271274.3 XM_006710976.3 XM_006710977.1 XM_011542301.2 XM_017002548.1

Gene Symbol
EFCAB7
Entrez Gene ID
84455
Gene Name
EF-hand calcium binding domain 7
Gene Aliases
-
UniGene
Hs.652324
Gene Chromosome Location
Chr.1: 63523301-63585400 on Build GRCh38
Transcripts
NR_002771.1 NM_032437.3 XM_005271274.3 XM_006710976.3 XM_006710977.1 XM_011542301.2 XM_017002548.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3948611 c.182G>T p.W61L Chr.1: 63549515-63549515 on Build GRCh38
3997627 c.213G>A p.S71S Chr.1: 63549546-63549546 on Build GRCh38
5078916 c.233_234delAA p.K80fs*>3 Chr.1: 63549566-63549567 on Build GRCh38
5091509 c.233delA p.M81fs*1 Chr.1: 63549566-63549566 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs6675580 Chr.1: 63549671-63549671 on Build GRCh38
rs892742933 Chr.1: 63549701-63549701 on Build GRCh38
rs976638404 Chr.1: 63549551-63549551 on Build GRCh38
rs78135692 Chr.1: 63549905-63549905 on Build GRCh38
rs796912651 Chr.1: 63549534-63549534 on Build GRCh38
rs529569611 Chr.1: 63549741-63549741 on Build GRCh38
rs1018037541 Chr.1: 63549818-63549818 on Build GRCh38
rs755051971 Chr.1: 63549565-63549566 on Build GRCh38
rs746322454 Chr.1: 63549782-63549782 on Build GRCh38
rs750061714 Chr.1: 63549626-63549626 on Build GRCh38
rs369947835 Chr.1: 63549740-63549741 on Build GRCh38
rs984060666 Chr.1: 63549949-63549949 on Build GRCh38
rs761231050 Chr.1: 63549618-63549618 on Build GRCh38
rs951313853 Chr.1: 63549936-63549938 on Build GRCh38
rs969266459 Chr.1: 63549968-63549968 on Build GRCh38
rs550340857 Chr.1: 63549770-63549770 on Build GRCh38
rs1007029182 Chr.1: 63549811-63549811 on Build GRCh38
rs115715373 Chr.1: 63549902-63549902 on Build GRCh38
rs200068339 Chr.1: 63549786-63549786 on Build GRCh38
rs918538003 Chr.1: 63549558-63549558 on Build GRCh38
rs217485 Chr.1: 63549546-63549546 on Build GRCh38
rs36003192 Chr.1: 63549774-63549774 on Build GRCh38
rs751438623 Chr.1: 63549566-63549566 on Build GRCh38
rs182386513 Chr.1: 63549699-63549699 on Build GRCh38
rs548616172 Chr.1: 63549531-63549531 on Build GRCh38
rs965031667 Chr.1: 63549873-63549873 on Build GRCh38
rs909666250 Chr.1: 63549954-63549954 on Build GRCh38
rs1044249745 Chr.1: 63549732-63549733 on Build GRCh38
rs762850448 Chr.1: 63549545-63549545 on Build GRCh38
rs561063124 Chr.1: 63549922-63549924 on Build GRCh38
rs745590747 Chr.1: 63549948-63549948 on Build GRCh38
rs566283653 Chr.1: 63549765-63549765 on Build GRCh38
rs1021356129 Chr.1: 63549521-63549521 on Build GRCh38
rs751058731 Chr.1: 63549593-63549593 on Build GRCh38
rs114906600 Chr.1: 63549728-63549728 on Build GRCh38
rs757292804 Chr.1: 63549625-63549625 on Build GRCh38
rs1025566434 Chr.1: 63549912-63549912 on Build GRCh38
rs568851020 Chr.1: 63549876-63549876 on Build GRCh38
rs948098210 Chr.1: 63549601-63549601 on Build GRCh38

View Full Product Details