Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 63550226-63550610 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DLEU2L
Entrez Gene ID
79469
Gene Name
deleted in lymphocytic leukemia 2-like
Gene Aliases
BCMSUNL
UniGene
Hs.659291
Gene Chromosome Location
Chr.1: 63548980-63550636 on Build GRCh38
Transcripts
NR_002771.1 NM_032437.3 XM_005271274.3 XM_006710976.3 XM_006710977.1 XM_011542301.2 XM_017002548.1

Gene Symbol
EFCAB7
Entrez Gene ID
84455
Gene Name
EF-hand calcium binding domain 7
Gene Aliases
-
UniGene
Hs.652324
Gene Chromosome Location
Chr.1: 63523301-63585400 on Build GRCh38
Transcripts
NR_002771.1 NM_032437.3 XM_005271274.3 XM_006710976.3 XM_006710977.1 XM_011542301.2 XM_017002548.1

SNP Detail

SNP ID Chromosome Location
rs377595998 Chr.1: 63550266-63550266 on Build GRCh38
rs980078811 Chr.1: 63550503-63550503 on Build GRCh38
rs761388295 Chr.1: 63550361-63550361 on Build GRCh38
rs969614826 Chr.1: 63550293-63550293 on Build GRCh38
rs34598062 Chr.1: 63550281-63550281 on Build GRCh38
rs772629848 Chr.1: 63550441-63550441 on Build GRCh38
rs572997070 Chr.1: 63550306-63550306 on Build GRCh38
rs538761848 Chr.1: 63550501-63550501 on Build GRCh38
rs545750036 Chr.1: 63550285-63550285 on Build GRCh38
rs571576164 Chr.1: 63550386-63550386 on Build GRCh38
rs1035064988 Chr.1: 63550372-63550372 on Build GRCh38
rs762952736 Chr.1: 63550548-63550548 on Build GRCh38
rs772118889 Chr.1: 63550385-63550390 on Build GRCh38
rs540454482 Chr.1: 63550458-63550458 on Build GRCh38
rs910827741 Chr.1: 63550554-63550554 on Build GRCh38
rs556153967 Chr.1: 63550270-63550270 on Build GRCh38
rs943528711 Chr.1: 63550558-63550558 on Build GRCh38
rs913880296 Chr.1: 63550495-63550495 on Build GRCh38
rs564280968 Chr.1: 63550300-63550300 on Build GRCh38
rs763357810 Chr.1: 63550409-63550409 on Build GRCh38

View Full Product Details