Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 110489263-110489816 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYMP
Entrez Gene ID
643160
Gene Name
chymosin pseudogene
Gene Aliases
-
UniGene
Hs.532720
Gene Chromosome Location
Chr.1: 110480766-110491269 on Build GRCh38
Transcripts
NR_003599.2 NR_108042.2

Gene Symbol
LOC440602
Entrez Gene ID
440602
Gene Name
uncharacterized LOC440602
Gene Aliases
-
UniGene
Hs.708945
Gene Chromosome Location
Chr.1: 110487680-110490258 on Build GRCh38
Transcripts
NR_003599.2 NR_108042.2

SNP Detail

SNP ID Chromosome Location
rs1001639905 Chr.1: 110489311-110489311 on Build GRCh38
rs894221143 Chr.1: 110489495-110489495 on Build GRCh38
rs755034607 Chr.1: 110489693-110489693 on Build GRCh38
rs927918611 Chr.1: 110489668-110489668 on Build GRCh38
rs902803099 Chr.1: 110489622-110489622 on Build GRCh38
rs1055645345 Chr.1: 110489596-110489596 on Build GRCh38
rs991401192 Chr.1: 110489686-110489686 on Build GRCh38
rs762498678 Chr.1: 110489414-110489414 on Build GRCh38
rs745660752 Chr.1: 110489480-110489480 on Build GRCh38
rs184979745 Chr.1: 110489540-110489540 on Build GRCh38
rs766288827 Chr.1: 110489688-110489688 on Build GRCh38
rs386634560 Chr.1: 110489514-110489515 on Build GRCh38
rs934376462 Chr.1: 110489729-110489729 on Build GRCh38
rs982109390 Chr.1: 110489662-110489662 on Build GRCh38
rs914360225 Chr.1: 110489704-110489704 on Build GRCh38
rs2271886 Chr.1: 110489488-110489488 on Build GRCh38
rs114449079 Chr.1: 110489333-110489333 on Build GRCh38
rs1857516 Chr.1: 110489419-110489419 on Build GRCh38
rs902918989 Chr.1: 110489618-110489618 on Build GRCh38
rs189290503 Chr.1: 110489589-110489589 on Build GRCh38
rs906983955 Chr.1: 110489454-110489454 on Build GRCh38
rs139729374 Chr.1: 110489617-110489617 on Build GRCh38
rs906938566 Chr.1: 110489306-110489306 on Build GRCh38
rs966529527 Chr.1: 110489533-110489533 on Build GRCh38
rs143756293 Chr.1: 110489439-110489439 on Build GRCh38
rs181941241 Chr.1: 110489515-110489515 on Build GRCh38
rs962274915 Chr.1: 110489678-110489679 on Build GRCh38
rs527562154 Chr.1: 110489334-110489334 on Build GRCh38
rs907009458 Chr.1: 110489312-110489312 on Build GRCh38
rs1048387272 Chr.1: 110489292-110489292 on Build GRCh38
rs1011292914 Chr.1: 110489496-110489496 on Build GRCh38
rs959960611 Chr.1: 110489682-110489682 on Build GRCh38
rs564183821 Chr.1: 110489607-110489607 on Build GRCh38
rs998512944 Chr.1: 110489623-110489623 on Build GRCh38
rs116028041 Chr.1: 110489643-110489643 on Build GRCh38
rs1048941159 Chr.1: 110489429-110489429 on Build GRCh38
rs189133749 Chr.1: 110489374-110489374 on Build GRCh38
rs931374408 Chr.1: 110489425-110489425 on Build GRCh38
rs888686940 Chr.1: 110489756-110489756 on Build GRCh38
rs571977200 Chr.1: 110489487-110489487 on Build GRCh38
rs912749388 Chr.1: 110489367-110489367 on Build GRCh38
rs911994565 Chr.1: 110489370-110489370 on Build GRCh38
rs537397317 Chr.1: 110489755-110489755 on Build GRCh38
rs370959233 Chr.1: 110489394-110489394 on Build GRCh38
rs1029062669 Chr.1: 110489650-110489650 on Build GRCh38
rs368376095 Chr.1: 110489440-110489440 on Build GRCh38
rs77684967 Chr.1: 110489798-110489798 on Build GRCh38
rs557111600 Chr.1: 110489762-110489762 on Build GRCh38
rs940613370 Chr.1: 110489584-110489584 on Build GRCh38
rs928574686 Chr.1: 110489590-110489590 on Build GRCh38
rs1055474519 Chr.1: 110489479-110489479 on Build GRCh38
rs559307830 Chr.1: 110489548-110489548 on Build GRCh38
rs371091947 Chr.1: 110489698-110489698 on Build GRCh38
rs902826829 Chr.1: 110489505-110489505 on Build GRCh38
rs780855564 Chr.1: 110489308-110489308 on Build GRCh38
rs1003602507 Chr.1: 110489775-110489775 on Build GRCh38
rs549997788 Chr.1: 110489632-110489632 on Build GRCh38
rs150055285 Chr.1: 110489689-110489689 on Build GRCh38
rs760781118 Chr.1: 110489592-110489592 on Build GRCh38
rs968045593 Chr.1: 110489364-110489364 on Build GRCh38
rs181246354 Chr.1: 110489656-110489656 on Build GRCh38
rs907777369 Chr.1: 110489435-110489435 on Build GRCh38
rs539211582 Chr.1: 110489398-110489398 on Build GRCh38
rs114302734 Chr.1: 110489774-110489774 on Build GRCh38
rs548958556 Chr.1: 110489299-110489299 on Build GRCh38
rs17629863 Chr.1: 110489534-110489534 on Build GRCh38
rs900823016 Chr.1: 110489741-110489741 on Build GRCh38
rs767123033 Chr.1: 110489724-110489724 on Build GRCh38
rs894282973 Chr.1: 110489600-110489600 on Build GRCh38
rs887692755 Chr.1: 110489297-110489297 on Build GRCh38
rs887862309 Chr.1: 110489796-110489796 on Build GRCh38
rs1022485476 Chr.1: 110489341-110489341 on Build GRCh38
rs908600176 Chr.1: 110489581-110489581 on Build GRCh38
rs1857515 Chr.1: 110489352-110489352 on Build GRCh38
rs1015709896 Chr.1: 110489658-110489658 on Build GRCh38
rs940063183 Chr.1: 110489586-110489586 on Build GRCh38
rs146843769 Chr.1: 110489555-110489555 on Build GRCh38
rs555377881 Chr.1: 110489460-110489460 on Build GRCh38
rs542741654 Chr.1: 110489545-110489545 on Build GRCh38
rs966147110 Chr.1: 110489372-110489372 on Build GRCh38
rs984197729 Chr.1: 110489562-110489562 on Build GRCh38
rs997025053 Chr.1: 110489633-110489633 on Build GRCh38
rs1034011504 Chr.1: 110489332-110489332 on Build GRCh38
rs573931023 Chr.1: 110489763-110489763 on Build GRCh38
rs907831549 Chr.1: 110489556-110489556 on Build GRCh38
rs116300006 Chr.1: 110489442-110489442 on Build GRCh38
rs1019238177 Chr.1: 110489625-110489625 on Build GRCh38
rs815327 Chr.1: 110489514-110489514 on Build GRCh38
rs1011700215 Chr.1: 110489351-110489351 on Build GRCh38
rs921898439 Chr.1: 110489416-110489416 on Build GRCh38
rs145426032 Chr.1: 110489690-110489690 on Build GRCh38
rs952932806 Chr.1: 110489653-110489653 on Build GRCh38
rs1036302807 Chr.1: 110489443-110489443 on Build GRCh38
rs977616155 Chr.1: 110489365-110489365 on Build GRCh38
rs754481914 Chr.1: 110489303-110489303 on Build GRCh38
rs902815295 Chr.1: 110489743-110489743 on Build GRCh38
rs762758968 Chr.1: 110489661-110489661 on Build GRCh38
rs1040699041 Chr.1: 110489730-110489730 on Build GRCh38
rs533532596 Chr.1: 110489500-110489500 on Build GRCh38
rs571840342 Chr.1: 110489753-110489753 on Build GRCh38
rs892875497 Chr.1: 110489603-110489603 on Build GRCh38
rs958472680 Chr.1: 110489344-110489344 on Build GRCh38
rs375410464 Chr.1: 110489672-110489672 on Build GRCh38

View Full Product Details