Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 121564304-121564856 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
EMBP1
Entrez Gene ID
647121
Gene Name
embigin pseudogene 1
Gene Aliases
-
UniGene
Hs.697682
Gene Chromosome Location
Chr.1: 121519112-121571888 on Build GRCh38
Transcripts
NR_003955.1

SNP Detail

SNP ID Chromosome Location
rs782388960 Chr.1: 121564762-121564762 on Build GRCh38
rs182397619 Chr.1: 121564810-121564810 on Build GRCh38
rs782359355 Chr.1: 121564829-121564829 on Build GRCh38
rs782738574 Chr.1: 121564807-121564807 on Build GRCh38
rs199602448 Chr.1: 121564674-121564674 on Build GRCh38
rs781904131 Chr.1: 121564813-121564813 on Build GRCh38
rs782807393 Chr.1: 121564777-121564777 on Build GRCh38
rs782580331 Chr.1: 121564812-121564812 on Build GRCh38
rs782039898 Chr.1: 121564734-121564734 on Build GRCh38
rs781835437 Chr.1: 121564410-121564410 on Build GRCh38
rs782308830 Chr.1: 121564733-121564733 on Build GRCh38
rs782724681 Chr.1: 121564383-121564383 on Build GRCh38
rs782044963 Chr.1: 121564773-121564773 on Build GRCh38
rs782194447 Chr.1: 121564712-121564712 on Build GRCh38
rs782601942 Chr.1: 121564695-121564695 on Build GRCh38
rs149461902 Chr.1: 121564420-121564420 on Build GRCh38
rs782753393 Chr.1: 121564796-121564796 on Build GRCh38
rs115966809 Chr.1: 121564769-121564769 on Build GRCh38
rs781987179 Chr.1: 121564768-121564768 on Build GRCh38
rs782815557 Chr.1: 121564623-121564623 on Build GRCh38
rs782246005 Chr.1: 121564824-121564824 on Build GRCh38
rs782722158 Chr.1: 121564667-121564667 on Build GRCh38
rs781830161 Chr.1: 121564804-121564804 on Build GRCh38
rs781883520 Chr.1: 121564625-121564625 on Build GRCh38
rs115455483 Chr.1: 121564425-121564425 on Build GRCh38
rs782343053 Chr.1: 121564612-121564612 on Build GRCh38
rs782646270 Chr.1: 121564821-121564821 on Build GRCh38
rs587619780 Chr.1: 121564480-121564480 on Build GRCh38
rs587681709 Chr.1: 121564686-121564686 on Build GRCh38
rs782774892 Chr.1: 121564737-121564737 on Build GRCh38
rs782127192 Chr.1: 121564637-121564637 on Build GRCh38
rs7534860 Chr.1: 121564370-121564370 on Build GRCh38
rs189301463 Chr.1: 121564659-121564659 on Build GRCh38
rs758508261 Chr.1: 121564437-121564438 on Build GRCh38
rs782285278 Chr.1: 121564753-121564753 on Build GRCh38
rs587691704 Chr.1: 121564622-121564622 on Build GRCh38
rs782503113 Chr.1: 121564688-121564688 on Build GRCh38
rs186147240 Chr.1: 121564426-121564426 on Build GRCh38
rs374487848 Chr.1: 121564621-121564621 on Build GRCh38
rs782548730 Chr.1: 121564816-121564816 on Build GRCh38
rs777675014 Chr.1: 121564504-121564504 on Build GRCh38
rs781838418 Chr.1: 121564643-121564643 on Build GRCh38
rs781880376 Chr.1: 121564784-121564784 on Build GRCh38
rs781803348 Chr.1: 121564830-121564830 on Build GRCh38
rs782753735 Chr.1: 121564638-121564638 on Build GRCh38
rs782508384 Chr.1: 121564791-121564791 on Build GRCh38
rs782548927 Chr.1: 121564684-121564684 on Build GRCh38
rs587733439 Chr.1: 121564583-121564583 on Build GRCh38
rs587693586 Chr.1: 121564578-121564578 on Build GRCh38
rs782017351 Chr.1: 121564635-121564635 on Build GRCh38
rs782232255 Chr.1: 121564606-121564606 on Build GRCh38
rs781928666 Chr.1: 121564772-121564772 on Build GRCh38

View Full Product Details