Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 147179728-147180259 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CHD1L
Entrez Gene ID
9557
Gene Name
chromodomain helicase DNA binding protein 1 like
Gene Aliases
ALC1, CHDL
UniGene
Hs.191164
Gene Chromosome Location
Chr.1: 147172804-147295766 on Build GRCh38
Transcripts
XM_017002858.1 NR_002305.1

Gene Symbol
PDIA3P1
Entrez Gene ID
171423
Gene Name
protein disulfide isomerase family A member 3 pseudogene 1
Gene Aliases
ERp60, GRP58P, PDIA3P
UniGene
Hs.658566
Gene Chromosome Location
Chr.1: 147177851-147179949 on Build GRCh38
Transcripts
XM_017002858.1 NR_002305.1

SNP Detail

SNP ID Chromosome Location
rs1027100138 Chr.1: 147179849-147179849 on Build GRCh38
rs202066614 Chr.1: 147179961-147179962 on Build GRCh38
rs368932204 Chr.1: 147179907-147179907 on Build GRCh38
rs952900565 Chr.1: 147180082-147180082 on Build GRCh38
rs10900323 Chr.1: 147180096-147180096 on Build GRCh38
rs200162460 Chr.1: 147179952-147179953 on Build GRCh38
rs782345627 Chr.1: 147180111-147180111 on Build GRCh38
rs782287346 Chr.1: 147179995-147179995 on Build GRCh38
rs782323699 Chr.1: 147179953-147179953 on Build GRCh38
rs970685454 Chr.1: 147180144-147180144 on Build GRCh38
rs74388450 Chr.1: 147179966-147179966 on Build GRCh38
rs141256302 Chr.1: 147179901-147179901 on Build GRCh38
rs1049456458 Chr.1: 147180206-147180206 on Build GRCh38
rs981372587 Chr.1: 147180149-147180149 on Build GRCh38
rs587680405 Chr.1: 147179968-147179968 on Build GRCh38
rs782674229 Chr.1: 147179754-147179754 on Build GRCh38
rs994727174 Chr.1: 147179801-147179801 on Build GRCh38
rs755894207 Chr.1: 147180182-147180182 on Build GRCh38
rs72708530 Chr.1: 147180122-147180122 on Build GRCh38
rs928515634 Chr.1: 147180162-147180162 on Build GRCh38
rs947127439 Chr.1: 147180201-147180201 on Build GRCh38
rs587597598 Chr.1: 147179769-147179769 on Build GRCh38
rs914365700 Chr.1: 147180185-147180185 on Build GRCh38
rs115000329 Chr.1: 147179875-147179875 on Build GRCh38
rs757270219 Chr.1: 147179993-147179993 on Build GRCh38
rs587615719 Chr.1: 147180078-147180078 on Build GRCh38
rs1053352677 Chr.1: 147180180-147180180 on Build GRCh38
rs71678783 Chr.1: 147179952-147179952 on Build GRCh38
rs115428662 Chr.1: 147179872-147179872 on Build GRCh38
rs587734345 Chr.1: 147180057-147180057 on Build GRCh38
rs934539135 Chr.1: 147180176-147180176 on Build GRCh38

View Full Product Details