Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 143881964-143882519 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FCGR1CP
Entrez Gene ID
100132417
Gene Name
Fc fragment of IgG receptor Ic, pseudogene
Gene Aliases
CD64c, FCGR1C, FCRIC, IGFR1, IGFRC
UniGene
Hs.635062
Gene Chromosome Location
Chr.1: 143874743-143883733 on Build GRCh38
Transcripts
NR_027484.2 XM_006711719.3

Gene Symbol
LOC100996713
Entrez Gene ID
100996713
Gene Name
uncharacterized LOC100996713
Gene Aliases
-
UniGene
Gene Chromosome Location
Chr.1: 143878414-143885954 on Build GRCh38
Transcripts
NR_027484.2 XM_006711719.3

SNP Detail

SNP ID Chromosome Location
rs587683624 Chr.1: 143882332-143882332 on Build GRCh38
rs587662529 Chr.1: 143882018-143882018 on Build GRCh38
rs587701733 Chr.1: 143882441-143882441 on Build GRCh38
rs148842033 Chr.1: 143882320-143882320 on Build GRCh38
rs587650184 Chr.1: 143882085-143882085 on Build GRCh38
rs199765895 Chr.1: 143882096-143882096 on Build GRCh38
rs587720669 Chr.1: 143882367-143882367 on Build GRCh38
rs587698859 Chr.1: 143882446-143882446 on Build GRCh38
rs113243187 Chr.1: 143882007-143882007 on Build GRCh38
rs587636588 Chr.1: 143882341-143882341 on Build GRCh38
rs149076838 Chr.1: 143882166-143882166 on Build GRCh38
rs587707273 Chr.1: 143882162-143882162 on Build GRCh38
rs587650451 Chr.1: 143882414-143882414 on Build GRCh38
rs61805133 Chr.1: 143882411-143882411 on Build GRCh38
rs777095626 Chr.1: 143882052-143882052 on Build GRCh38
rs56098172 Chr.1: 143882091-143882091 on Build GRCh38
rs587729925 Chr.1: 143882472-143882472 on Build GRCh38
rs184101012 Chr.1: 143882023-143882023 on Build GRCh38
rs587763739 Chr.1: 143882333-143882333 on Build GRCh38
rs587627642 Chr.1: 143882489-143882489 on Build GRCh38
rs587695636 Chr.1: 143882203-143882203 on Build GRCh38
rs71622632 Chr.1: 143882017-143882017 on Build GRCh38
rs188360074 Chr.1: 143882089-143882089 on Build GRCh38
rs71622631 Chr.1: 143881988-143881988 on Build GRCh38
rs771389266 Chr.1: 143882013-143882013 on Build GRCh38
rs587600972 Chr.1: 143882442-143882442 on Build GRCh38
rs587602982 Chr.1: 143882010-143882010 on Build GRCh38
rs587688844 Chr.1: 143882496-143882496 on Build GRCh38
rs111756820 Chr.1: 143882184-143882184 on Build GRCh38
rs376971982 Chr.1: 143882200-143882200 on Build GRCh38
rs587757209 Chr.1: 143882261-143882261 on Build GRCh38
rs587668027 Chr.1: 143881998-143881998 on Build GRCh38

View Full Product Details