Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 171339992-171340535 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FMO4
Entrez Gene ID
2329
Gene Name
flavin containing monooxygenase 4
Gene Aliases
FMO2
UniGene
Hs.386502
Gene Chromosome Location
Chr.1: 171314131-171342084 on Build GRCh38
Transcripts
NM_002022.2 XM_005245045.3 XM_005245046.3 XM_005245048.3 XM_006711243.3 XM_011509347.2 XM_011509348.2 NR_002719.2

Gene Symbol
TOP1P1
Entrez Gene ID
7151
Gene Name
topoisomerase (DNA) I pseudogene 1
Gene Aliases
-
UniGene
Hs.661092
Gene Chromosome Location
Chr.1: 171338896-171341326 on Build GRCh38
Transcripts
NM_002022.2 XM_005245045.3 XM_005245046.3 XM_005245048.3 XM_006711243.3 XM_011509347.2 XM_011509348.2 NR_002719.2

SNP Detail

SNP ID Chromosome Location
rs1016400209 Chr.1: 171340404-171340406 on Build GRCh38
rs553308289 Chr.1: 171340439-171340439 on Build GRCh38
rs575736907 Chr.1: 171340143-171340143 on Build GRCh38
rs532423574 Chr.1: 171340250-171340250 on Build GRCh38
rs751966232 Chr.1: 171340056-171340056 on Build GRCh38
rs542305799 Chr.1: 171340061-171340061 on Build GRCh38
rs958918761 Chr.1: 171340316-171340316 on Build GRCh38
rs936568253 Chr.1: 171340484-171340484 on Build GRCh38
rs749605097 Chr.1: 171340447-171340447 on Build GRCh38
rs1041796396 Chr.1: 171340249-171340249 on Build GRCh38
rs1050552434 Chr.1: 171340116-171340116 on Build GRCh38
rs945210795 Chr.1: 171340514-171340514 on Build GRCh38
rs745986023 Chr.1: 171340511-171340511 on Build GRCh38
rs1010243991 Chr.1: 171340188-171340188 on Build GRCh38
rs540997332 Chr.1: 171340405-171340405 on Build GRCh38
rs936620708 Chr.1: 171340144-171340144 on Build GRCh38
rs567312436 Chr.1: 171340025-171340025 on Build GRCh38
rs45447795 Chr.1: 171340083-171340083 on Build GRCh38
rs572350980 Chr.1: 171340043-171340043 on Build GRCh38
rs780639935 Chr.1: 171340457-171340458 on Build GRCh38
rs111790068 Chr.1: 171340408-171340408 on Build GRCh38
rs1006499357 Chr.1: 171340400-171340400 on Build GRCh38
rs529871676 Chr.1: 171340464-171340464 on Build GRCh38
rs1053520998 Chr.1: 171340154-171340154 on Build GRCh38
rs562515231 Chr.1: 171340451-171340451 on Build GRCh38
rs146261362 Chr.1: 171340171-171340171 on Build GRCh38
rs1030078242 Chr.1: 171340440-171340440 on Build GRCh38
rs777918398 Chr.1: 171340262-171340262 on Build GRCh38
rs532902589 Chr.1: 171340164-171340164 on Build GRCh38
rs764840854 Chr.1: 171340044-171340044 on Build GRCh38
rs756531910 Chr.1: 171340218-171340218 on Build GRCh38
rs55809312 Chr.1: 171340109-171340109 on Build GRCh38
rs989364649 Chr.1: 171340505-171340505 on Build GRCh38
rs12737633 Chr.1: 171340480-171340480 on Build GRCh38

View Full Product Details