Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.1: 228651816-228652362 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DUSP5P1
Entrez Gene ID
574029
Gene Name
dual specificity phosphatase 5 pseudogene 1
Gene Aliases
DUSP5P
UniGene
Hs.647777
Gene Chromosome Location
Chr.1: 228644910-228652412 on Build GRCh38
Transcripts
NR_002834.1 NR_037962.1

Gene Symbol
RHOU
Entrez Gene ID
58480
Gene Name
ras homolog family member U
Gene Aliases
ARHU, CDC42L1, G28K, WRCH1, hG28K
UniGene
Hs.647774
Gene Chromosome Location
Chr.1: 228644647-228746669 on Build GRCh38
Transcripts
NR_002834.1 NR_037962.1

SNP Detail

SNP ID Chromosome Location
rs953982322 Chr.1: 228652313-228652313 on Build GRCh38
rs151190513 Chr.1: 228652213-228652213 on Build GRCh38
rs932100634 Chr.1: 228651839-228651839 on Build GRCh38
rs146517740 Chr.1: 228651908-228651908 on Build GRCh38
rs7536593 Chr.1: 228651850-228651850 on Build GRCh38
rs183633332 Chr.1: 228652251-228652251 on Build GRCh38
rs531969445 Chr.1: 228651961-228651961 on Build GRCh38
rs751637429 Chr.1: 228652064-228652064 on Build GRCh38
rs183112836 Chr.1: 228651893-228651893 on Build GRCh38
rs573767685 Chr.1: 228652281-228652281 on Build GRCh38
rs187682156 Chr.1: 228652197-228652197 on Build GRCh38
rs143391300 Chr.1: 228652201-228652201 on Build GRCh38
rs144172873 Chr.1: 228651871-228651871 on Build GRCh38
rs945966498 Chr.1: 228652267-228652267 on Build GRCh38
rs547642593 Chr.1: 228652180-228652180 on Build GRCh38
rs1009443909 Chr.1: 228651976-228651976 on Build GRCh38
rs527910086 Chr.1: 228652060-228652060 on Build GRCh38
rs567832398 Chr.1: 228652335-228652335 on Build GRCh38
rs530636061 Chr.1: 228652177-228652177 on Build GRCh38
rs1036516864 Chr.1: 228652148-228652148 on Build GRCh38
rs11810178 Chr.1: 228652219-228652219 on Build GRCh38
rs1048170720 Chr.1: 228651969-228651969 on Build GRCh38
rs971016511 Chr.1: 228652179-228652179 on Build GRCh38
rs571564309 Chr.1: 228652014-228652014 on Build GRCh38
rs191726831 Chr.1: 228652226-228652226 on Build GRCh38
rs1050918956 Chr.1: 228651842-228651842 on Build GRCh38
rs902480419 Chr.1: 228652093-228652093 on Build GRCh38
rs919390582 Chr.1: 228652163-228652163 on Build GRCh38
rs865927936 Chr.1: 228652040-228652040 on Build GRCh38
rs562807355 Chr.1: 228651899-228651899 on Build GRCh38
rs10916349 Chr.1: 228651991-228651991 on Build GRCh38
rs549927413 Chr.1: 228652183-228652183 on Build GRCh38
rs901109830 Chr.1: 228652022-228652022 on Build GRCh38
rs139949097 Chr.1: 228651936-228651936 on Build GRCh38
rs757389247 Chr.1: 228652081-228652081 on Build GRCh38
rs1034733696 Chr.1: 228651935-228651935 on Build GRCh38
rs190926605 Chr.1: 228651874-228651874 on Build GRCh38
rs912015205 Chr.1: 228652088-228652088 on Build GRCh38
rs930928303 Chr.1: 228652166-228652166 on Build GRCh38
rs1032378823 Chr.1: 228652135-228652135 on Build GRCh38
rs868630163 Chr.1: 228652038-228652038 on Build GRCh38
rs749115728 Chr.1: 228651987-228651987 on Build GRCh38
rs547915044 Chr.1: 228652102-228652102 on Build GRCh38
rs115659962 Chr.1: 228652178-228652178 on Build GRCh38
rs188166631 Chr.1: 228652317-228652317 on Build GRCh38
rs979731509 Chr.1: 228652034-228652034 on Build GRCh38
rs371946160 Chr.1: 228652308-228652308 on Build GRCh38
rs754685575 Chr.1: 228652208-228652208 on Build GRCh38
rs972941083 Chr.1: 228652188-228652188 on Build GRCh38
rs918228161 Chr.1: 228651922-228651923 on Build GRCh38
rs1010081545 Chr.1: 228652041-228652041 on Build GRCh38
rs571262695 Chr.1: 228652220-228652220 on Build GRCh38
rs896261357 Chr.1: 228652207-228652207 on Build GRCh38
rs540283308 Chr.1: 228651866-228651866 on Build GRCh38
rs771816718 Chr.1: 228652276-228652276 on Build GRCh38

View Full Product Details