Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 73719003-73719552 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BMS1P4
Entrez Gene ID
729096
Gene Name
BMS1, ribosome biogenesis factor pseudogene 4
Gene Aliases
BMS1LP4
UniGene
Hs.709171
Gene Chromosome Location
Chr.10: 73699151-73730514 on Build GRCh38
Transcripts
NR_026592.2

SNP Detail

SNP ID Chromosome Location
rs952154733 Chr.10: 73719070-73719070 on Build GRCh38
rs535639545 Chr.10: 73719506-73719506 on Build GRCh38
rs183585555 Chr.10: 73719489-73719489 on Build GRCh38
rs962850280 Chr.10: 73719258-73719258 on Build GRCh38
rs774958744 Chr.10: 73719496-73719496 on Build GRCh38
rs576584915 Chr.10: 73719035-73719035 on Build GRCh38
rs750518850 Chr.10: 73719500-73719501 on Build GRCh38
rs369420895 Chr.10: 73719282-73719282 on Build GRCh38
rs972910490 Chr.10: 73719275-73719275 on Build GRCh38
rs6480709 Chr.10: 73719367-73719367 on Build GRCh38
rs1009996129 Chr.10: 73719357-73719357 on Build GRCh38
rs1054202785 Chr.10: 73719326-73719326 on Build GRCh38
rs532835564 Chr.10: 73719177-73719177 on Build GRCh38
rs554733055 Chr.10: 73719516-73719516 on Build GRCh38
rs562931848 Chr.10: 73719231-73719231 on Build GRCh38
rs990908997 Chr.10: 73719229-73719229 on Build GRCh38
rs907046430 Chr.10: 73719382-73719382 on Build GRCh38
rs1012703254 Chr.10: 73719498-73719498 on Build GRCh38
rs566582353 Chr.10: 73719294-73719294 on Build GRCh38
rs368130201 Chr.10: 73719296-73719296 on Build GRCh38
rs2127365 Chr.10: 73719066-73719066 on Build GRCh38
rs547931008 Chr.10: 73719283-73719283 on Build GRCh38
rs1034711248 Chr.10: 73719430-73719430 on Build GRCh38
rs1002766299 Chr.10: 73719417-73719417 on Build GRCh38
rs1020203889 Chr.10: 73719121-73719121 on Build GRCh38
rs374188271 Chr.10: 73719165-73719165 on Build GRCh38
rs537743063 Chr.10: 73719372-73719372 on Build GRCh38
rs934220415 Chr.10: 73719281-73719281 on Build GRCh38
rs191698042 Chr.10: 73719485-73719485 on Build GRCh38
rs774167919 Chr.10: 73719139-73719139 on Build GRCh38
rs1003203702 Chr.10: 73719128-73719128 on Build GRCh38
rs551739504 Chr.10: 73719354-73719354 on Build GRCh38
rs540649139 Chr.10: 73719124-73719124 on Build GRCh38
rs551342927 Chr.10: 73719218-73719218 on Build GRCh38
rs573128916 Chr.10: 73719120-73719120 on Build GRCh38
rs942383678 Chr.10: 73719325-73719325 on Build GRCh38
rs918764209 Chr.10: 73719280-73719280 on Build GRCh38
rs892955742 Chr.10: 73719065-73719065 on Build GRCh38
rs892922908 Chr.10: 73719342-73719342 on Build GRCh38

View Full Product Details