Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.10: 130108170-130108716 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE7P
Entrez Gene ID
119437
Gene Name
CTAGE family member 7, pseudogene
Gene Aliases
CTAGE7, CTAGEP, bA500G10.2, rcCTAGE5
UniGene
Hs.620802
Gene Chromosome Location
Chr.10: 130106009-130108838 on Build GRCh38
Transcripts
NR_044994.1 NR_034125.1

Gene Symbol
LINC00959
Entrez Gene ID
387723
Gene Name
long intergenic non-protein coding RNA 959
Gene Aliases
-
UniGene
Hs.127394
Gene Chromosome Location
Chr.10: 130063896-130110817 on Build GRCh38
Transcripts
NR_044994.1 NR_034125.1

SNP Detail

SNP ID Chromosome Location
rs562881852 Chr.10: 130108295-130108295 on Build GRCh38
rs755071717 Chr.10: 130108280-130108280 on Build GRCh38
rs61874369 Chr.10: 130108378-130108378 on Build GRCh38
rs773329919 Chr.10: 130108344-130108344 on Build GRCh38
rs779126505 Chr.10: 130108348-130108348 on Build GRCh38
rs764262934 Chr.10: 130108403-130108403 on Build GRCh38
rs746298189 Chr.10: 130108424-130108424 on Build GRCh38
rs11017081 Chr.10: 130108279-130108279 on Build GRCh38
rs558883574 Chr.10: 130108542-130108542 on Build GRCh38
rs1013394325 Chr.10: 130108607-130108607 on Build GRCh38
rs758553802 Chr.10: 130108351-130108351 on Build GRCh38
rs768797786 Chr.10: 130108219-130108219 on Build GRCh38
rs191768647 Chr.10: 130108676-130108676 on Build GRCh38
rs748414975 Chr.10: 130108223-130108223 on Build GRCh38
rs762631933 Chr.10: 130108264-130108264 on Build GRCh38
rs773963211 Chr.10: 130108484-130108484 on Build GRCh38
rs374581228 Chr.10: 130108475-130108475 on Build GRCh38
rs758101723 Chr.10: 130108196-130108196 on Build GRCh38
rs771157342 Chr.10: 130108355-130108355 on Build GRCh38
rs757546899 Chr.10: 130108406-130108406 on Build GRCh38
rs747265689 Chr.10: 130108296-130108296 on Build GRCh38
rs112214906 Chr.10: 130108601-130108601 on Build GRCh38
rs773007220 Chr.10: 130108498-130108498 on Build GRCh38
rs560098907 Chr.10: 130108281-130108281 on Build GRCh38
rs779082614 Chr.10: 130108196-130108196 on Build GRCh38
rs752895661 Chr.10: 130108349-130108349 on Build GRCh38
rs759608610 Chr.10: 130108433-130108433 on Build GRCh38
rs754717296 Chr.10: 130108402-130108402 on Build GRCh38
rs572895238 Chr.10: 130108212-130108212 on Build GRCh38
rs770224293 Chr.10: 130108681-130108681 on Build GRCh38
rs780179803 Chr.10: 130108210-130108210 on Build GRCh38
rs532397181 Chr.10: 130108289-130108289 on Build GRCh38
rs771337766 Chr.10: 130108300-130108300 on Build GRCh38
rs1040636639 Chr.10: 130108538-130108538 on Build GRCh38
rs573955134 Chr.10: 130108344-130108344 on Build GRCh38
rs760689576 Chr.10: 130108396-130108396 on Build GRCh38
rs763854573 Chr.10: 130108273-130108273 on Build GRCh38
rs756172622 Chr.10: 130108342-130108342 on Build GRCh38
rs751238892 Chr.10: 130108320-130108320 on Build GRCh38
rs777196163 Chr.10: 130108236-130108236 on Build GRCh38
rs755386571 Chr.10: 130108193-130108193 on Build GRCh38
rs533742682 Chr.10: 130108642-130108642 on Build GRCh38
rs377722933 Chr.10: 130108251-130108251 on Build GRCh38
rs771828132 Chr.10: 130108488-130108488 on Build GRCh38
rs771126669 Chr.10: 130108353-130108354 on Build GRCh38
rs748526916 Chr.10: 130108572-130108572 on Build GRCh38
rs773120686 Chr.10: 130108394-130108394 on Build GRCh38
rs763670599 Chr.10: 130108318-130108318 on Build GRCh38
rs746351111 Chr.10: 130108361-130108361 on Build GRCh38
rs776900165 Chr.10: 130108376-130108379 on Build GRCh38
rs768179475 Chr.10: 130108380-130108380 on Build GRCh38
rs769261318 Chr.10: 130108307-130108307 on Build GRCh38
rs760484886 Chr.10: 130108500-130108500 on Build GRCh38
rs756484942 Chr.10: 130108436-130108436 on Build GRCh38
rs182265512 Chr.10: 130108371-130108371 on Build GRCh38
rs189223394 Chr.10: 130108249-130108249 on Build GRCh38
rs762580508 Chr.10: 130108311-130108311 on Build GRCh38
rs546712661 Chr.10: 130108423-130108423 on Build GRCh38
rs575508006 Chr.10: 130108237-130108237 on Build GRCh38
rs748689408 Chr.10: 130108250-130108250 on Build GRCh38
rs747060321 Chr.10: 130108343-130108344 on Build GRCh38
rs575659184 Chr.10: 130108654-130108655 on Build GRCh38
rs914181765 Chr.10: 130108539-130108539 on Build GRCh38
rs761569866 Chr.10: 130108389-130108389 on Build GRCh38
rs148869275 Chr.10: 130108321-130108323 on Build GRCh38
rs759584966 Chr.10: 130108516-130108516 on Build GRCh38
rs781649219 Chr.10: 130108356-130108356 on Build GRCh38
rs138337257 Chr.10: 130108643-130108643 on Build GRCh38
rs780986635 Chr.10: 130108439-130108439 on Build GRCh38
rs538921464 Chr.10: 130108233-130108233 on Build GRCh38
rs766295248 Chr.10: 130108501-130108501 on Build GRCh38
rs768021879 Chr.10: 130108461-130108461 on Build GRCh38
rs767197611 Chr.10: 130108392-130108392 on Build GRCh38
rs767378743 Chr.10: 130108338-130108338 on Build GRCh38
rs764801406 Chr.10: 130108200-130108200 on Build GRCh38
rs753343771 Chr.10: 130108192-130108192 on Build GRCh38
rs754650136 Chr.10: 130108199-130108199 on Build GRCh38
rs548444045 Chr.10: 130108374-130108374 on Build GRCh38
rs761468295 Chr.10: 130108325-130108325 on Build GRCh38
rs774667559 Chr.10: 130108220-130108220 on Build GRCh38
rs751368261 Chr.10: 130108274-130108274 on Build GRCh38
rs750300235 Chr.10: 130108340-130108340 on Build GRCh38
rs552448018 Chr.10: 130108294-130108294 on Build GRCh38
rs185909231 Chr.10: 130108243-130108243 on Build GRCh38
rs780491183 Chr.10: 130108438-130108438 on Build GRCh38
rs566456522 Chr.10: 130108485-130108485 on Build GRCh38
rs757189285 Chr.10: 130108206-130108206 on Build GRCh38
rs551388874 Chr.10: 130108278-130108278 on Build GRCh38
rs775029685 Chr.10: 130108308-130108308 on Build GRCh38
rs571280802 Chr.10: 130108306-130108306 on Build GRCh38
rs1028070714 Chr.10: 130108677-130108677 on Build GRCh38
rs187316073 Chr.10: 130108510-130108510 on Build GRCh38
rs773837911 Chr.10: 130108387-130108387 on Build GRCh38
rs552668092 Chr.10: 130108201-130108201 on Build GRCh38
rs776951837 Chr.10: 130108305-130108305 on Build GRCh38
rs574793713 Chr.10: 130108276-130108276 on Build GRCh38
rs535936044 Chr.10: 130108404-130108404 on Build GRCh38
rs766348335 Chr.10: 130108398-130108398 on Build GRCh38
rs777795852 Chr.10: 130108354-130108354 on Build GRCh38
rs1043799060 Chr.10: 130108553-130108553 on Build GRCh38
rs769660598 Chr.10: 130108518-130108521 on Build GRCh38
rs903569461 Chr.10: 130108559-130108559 on Build GRCh38
rs1015440919 Chr.10: 130108611-130108616 on Build GRCh38
rs558922358 Chr.10: 130108234-130108234 on Build GRCh38

View Full Product Details