Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8133694-8134236 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CLEC4A
Entrez Gene ID
50856
Gene Name
C-type lectin domain family 4 member A
Gene Aliases
CD367, CLECSF6, DCIR, DDB27, HDCGC13P, LLIR
UniGene
Hs.504657
Gene Chromosome Location
Chr.12: 8102932-8138607 on Build GRCh38
Transcripts
NM_016184.3 NM_194447.2 NM_194448.2 NM_194450.2 XM_011520684.2 XM_017019382.1 NR_036440.1

Gene Symbol
POU5F1P3
Entrez Gene ID
642559
Gene Name
POU class 5 homeobox 1 pseudogene 3
Gene Aliases
OCT4-pg3, OTF3L, POU5F1L, POU5FLC12
UniGene
Hs.646545
Gene Chromosome Location
Chr.12: 8133769-8134852 on Build GRCh38
Transcripts
NM_016184.3 NM_194447.2 NM_194448.2 NM_194450.2 XM_011520684.2 XM_017019382.1 NR_036440.1

SNP Detail

SNP ID Chromosome Location
rs993368844 Chr.12: 8134003-8134003 on Build GRCh38
rs947614244 Chr.12: 8133839-8133839 on Build GRCh38
rs764855473 Chr.12: 8134154-8134154 on Build GRCh38
rs758767714 Chr.12: 8134178-8134178 on Build GRCh38
rs111592684 Chr.12: 8134175-8134175 on Build GRCh38
rs954531808 Chr.12: 8134014-8134015 on Build GRCh38
rs367900744 Chr.12: 8133885-8133886 on Build GRCh38
rs571481913 Chr.12: 8134179-8134179 on Build GRCh38
rs894145917 Chr.12: 8133950-8133950 on Build GRCh38
rs977629927 Chr.12: 8134143-8134143 on Build GRCh38
rs193186392 Chr.12: 8134197-8134197 on Build GRCh38
rs1008749761 Chr.12: 8134020-8134020 on Build GRCh38
rs964103959 Chr.12: 8134107-8134107 on Build GRCh38
rs914567271 Chr.12: 8134133-8134133 on Build GRCh38
rs757732540 Chr.12: 8134215-8134215 on Build GRCh38
rs986871566 Chr.12: 8133798-8133798 on Build GRCh38
rs750068989 Chr.12: 8134116-8134116 on Build GRCh38
rs899128144 Chr.12: 8133982-8133982 on Build GRCh38
rs1026102787 Chr.12: 8133762-8133762 on Build GRCh38
rs374144237 Chr.12: 8134055-8134055 on Build GRCh38
rs111886055 Chr.12: 8134170-8134170 on Build GRCh38
rs773080046 Chr.12: 8133972-8133972 on Build GRCh38
rs780608976 Chr.12: 8134083-8134083 on Build GRCh38
rs751520748 Chr.12: 8134006-8134006 on Build GRCh38
rs1019723145 Chr.12: 8134022-8134022 on Build GRCh38
rs747521665 Chr.12: 8133736-8133736 on Build GRCh38
rs752557954 Chr.12: 8133939-8133939 on Build GRCh38
rs765873122 Chr.12: 8134050-8134050 on Build GRCh38
rs200565509 Chr.12: 8133831-8133831 on Build GRCh38
rs61642531 Chr.12: 8133889-8133890 on Build GRCh38
rs938432034 Chr.12: 8133900-8133900 on Build GRCh38
rs968966154 Chr.12: 8134136-8134136 on Build GRCh38
rs1037310819 Chr.12: 8133971-8133973 on Build GRCh38
rs1041940353 Chr.12: 8133871-8133871 on Build GRCh38
rs188301488 Chr.12: 8133742-8133742 on Build GRCh38
rs964056124 Chr.12: 8133806-8133806 on Build GRCh38
rs924792073 Chr.12: 8134160-8134160 on Build GRCh38
rs975395674 Chr.12: 8133807-8133807 on Build GRCh38
rs537265234 Chr.12: 8134191-8134191 on Build GRCh38
rs369213036 Chr.12: 8133761-8133761 on Build GRCh38
rs762746149 Chr.12: 8133970-8133970 on Build GRCh38
rs7315526 Chr.12: 8134166-8134166 on Build GRCh38
rs963252278 Chr.12: 8133737-8133737 on Build GRCh38
rs912558884 Chr.12: 8133800-8133800 on Build GRCh38
rs774185227 Chr.12: 8133930-8133931 on Build GRCh38
rs951582680 Chr.12: 8133764-8133764 on Build GRCh38
rs868508492 Chr.12: 8133817-8133817 on Build GRCh38

View Full Product Details