Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 8234154-8234708 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FAM86FP
Entrez Gene ID
653113
Gene Name
family with sequence similarity 86, member A pseudogene
Gene Aliases
-
UniGene
Hs.661749
Gene Chromosome Location
Chr.12: 8231049-8242946 on Build GRCh38
Transcripts
NR_024254.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
3872624 c.768C>T p.V256V Chr.12: 8234388-8234388 on Build GRCh38
3872623 c.797A>G p.Q266R Chr.12: 8234359-8234359 on Build GRCh38
3987237 c.731-5C>T p.? Chr.12: 8234430-8234430 on Build GRCh38
1128458 c.841C>A p.R281S Chr.12: 8234315-8234315 on Build GRCh38
6817532 c.838G>A p.V280I Chr.12: 8234318-8234318 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs774533033 Chr.12: 8234497-8234497 on Build GRCh38
rs200610009 Chr.12: 8234418-8234418 on Build GRCh38
rs9705655 Chr.12: 8234405-8234405 on Build GRCh38
rs10841037 Chr.12: 8234282-8234282 on Build GRCh38
rs891069199 Chr.12: 8234586-8234586 on Build GRCh38
rs868032365 Chr.12: 8234224-8234224 on Build GRCh38
rs200607723 Chr.12: 8234347-8234347 on Build GRCh38
rs76887484 Chr.12: 8234263-8234263 on Build GRCh38
rs376422111 Chr.12: 8234378-8234378 on Build GRCh38
rs200991102 Chr.12: 8234270-8234270 on Build GRCh38
rs2965690 Chr.12: 8234267-8234267 on Build GRCh38
rs1022417086 Chr.12: 8234640-8234640 on Build GRCh38
rs149095277 Chr.12: 8234327-8234327 on Build GRCh38
rs928230883 Chr.12: 8234468-8234468 on Build GRCh38
rs768394839 Chr.12: 8234387-8234387 on Build GRCh38
rs752996474 Chr.12: 8234184-8234184 on Build GRCh38
rs181263320 Chr.12: 8234514-8234514 on Build GRCh38
rs769290702 Chr.12: 8234364-8234364 on Build GRCh38
rs766158100 Chr.12: 8234516-8234516 on Build GRCh38
rs955298397 Chr.12: 8234357-8234357 on Build GRCh38
rs201178389 Chr.12: 8234283-8234283 on Build GRCh38
rs2968741 Chr.12: 8234261-8234261 on Build GRCh38
rs760679917 Chr.12: 8234423-8234423 on Build GRCh38
rs768448207 Chr.12: 8234598-8234598 on Build GRCh38
rs777609294 Chr.12: 8234318-8234318 on Build GRCh38
rs2968743 Chr.12: 8234315-8234315 on Build GRCh38
rs112976735 Chr.12: 8234625-8234625 on Build GRCh38
rs199498554 Chr.12: 8234437-8234437 on Build GRCh38
rs1018166364 Chr.12: 8234623-8234623 on Build GRCh38
rs185402894 Chr.12: 8234314-8234314 on Build GRCh38
rs190621314 Chr.12: 8234554-8234554 on Build GRCh38
rs1011386046 Chr.12: 8234634-8234635 on Build GRCh38
rs190791219 Chr.12: 8234430-8234430 on Build GRCh38
rs190700361 Chr.12: 8234330-8234330 on Build GRCh38
rs757962698 Chr.12: 8234578-8234578 on Build GRCh38
rs750133750 Chr.12: 8234368-8234368 on Build GRCh38
rs993818108 Chr.12: 8234324-8234324 on Build GRCh38
rs58469548 Chr.12: 8234641-8234642 on Build GRCh38
rs978105764 Chr.12: 8234443-8234443 on Build GRCh38
rs143285453 Chr.12: 8234673-8234673 on Build GRCh38
rs186013063 Chr.12: 8234424-8234424 on Build GRCh38
rs778672574 Chr.12: 8234618-8234618 on Build GRCh38
rs544688999 Chr.12: 8234210-8234210 on Build GRCh38
rs779418500 Chr.12: 8234377-8234377 on Build GRCh38
rs781134662 Chr.12: 8234352-8234352 on Build GRCh38
rs199980429 Chr.12: 8234241-8234241 on Build GRCh38
rs779657657 Chr.12: 8234556-8234556 on Build GRCh38
rs879790855 Chr.12: 8234400-8234400 on Build GRCh38
rs532817338 Chr.12: 8234302-8234302 on Build GRCh38
rs374528103 Chr.12: 8234604-8234604 on Build GRCh38
rs201336501 Chr.12: 8234239-8234239 on Build GRCh38
rs916749682 Chr.12: 8234512-8234512 on Build GRCh38
rs370639322 Chr.12: 8234390-8234390 on Build GRCh38
rs370752955 Chr.12: 8234529-8234529 on Build GRCh38
rs772427311 Chr.12: 8234660-8234660 on Build GRCh38
rs966802057 Chr.12: 8234652-8234652 on Build GRCh38
rs1038097413 Chr.12: 8234527-8234527 on Build GRCh38
rs781005461 Chr.12: 8234546-8234546 on Build GRCh38
rs769203566 Chr.12: 8234204-8234205 on Build GRCh38
rs199607880 Chr.12: 8234394-8234394 on Build GRCh38
rs368773660 Chr.12: 8234617-8234617 on Build GRCh38
rs2968742 Chr.12: 8234275-8234275 on Build GRCh38
rs746208705 Chr.12: 8234663-8234663 on Build GRCh38
rs939617458 Chr.12: 8234500-8234500 on Build GRCh38
rs759793004 Chr.12: 8234488-8234488 on Build GRCh38
rs185711519 Chr.12: 8234553-8234553 on Build GRCh38
rs75997195 Chr.12: 8234359-8234359 on Build GRCh38
rs12302716 Chr.12: 8234301-8234301 on Build GRCh38

View Full Product Details