Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 9233282-9233834 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
A2MP1
Entrez Gene ID
3
Gene Name
alpha-2-macroglobulin pseudogene 1
Gene Aliases
A2MP
UniGene
Hs.569125
Gene Chromosome Location
Chr.12: 9228533-9234207 on Build GRCh38
Transcripts
NR_040112.1

SNP Detail

SNP ID Chromosome Location
rs1018690650 Chr.12: 9233315-9233315 on Build GRCh38
rs1031717597 Chr.12: 9233591-9233591 on Build GRCh38
rs775896449 Chr.12: 9233379-9233379 on Build GRCh38
rs773985393 Chr.12: 9233425-9233425 on Build GRCh38
rs992917170 Chr.12: 9233623-9233623 on Build GRCh38
rs888320639 Chr.12: 9233744-9233744 on Build GRCh38
rs79617020 Chr.12: 9233771-9233771 on Build GRCh38
rs939670279 Chr.12: 9233700-9233700 on Build GRCh38
rs915754753 Chr.12: 9233639-9233639 on Build GRCh38
rs1003854748 Chr.12: 9233410-9233410 on Build GRCh38
rs909769058 Chr.12: 9233671-9233671 on Build GRCh38
rs116046537 Chr.12: 9233528-9233528 on Build GRCh38
rs760790373 Chr.12: 9233442-9233442 on Build GRCh38
rs186807146 Chr.12: 9233757-9233757 on Build GRCh38
rs183489125 Chr.12: 9233372-9233372 on Build GRCh38
rs748060079 Chr.12: 9233791-9233795 on Build GRCh38
rs765670542 Chr.12: 9233767-9233767 on Build GRCh38
rs762132886 Chr.12: 9233578-9233578 on Build GRCh38
rs933764825 Chr.12: 9233778-9233778 on Build GRCh38
rs977355899 Chr.12: 9233590-9233590 on Build GRCh38
rs948675586 Chr.12: 9233348-9233348 on Build GRCh38
rs749376305 Chr.12: 9233484-9233484 on Build GRCh38
rs899793741 Chr.12: 9233316-9233316 on Build GRCh38
rs1030951631 Chr.12: 9233444-9233444 on Build GRCh38
rs762682455 Chr.12: 9233504-9233504 on Build GRCh38
rs749134760 Chr.12: 9233353-9233353 on Build GRCh38
rs7315222 Chr.12: 9233614-9233614 on Build GRCh38
rs982195652 Chr.12: 9233605-9233605 on Build GRCh38
rs929750083 Chr.12: 9233638-9233638 on Build GRCh38
rs948559775 Chr.12: 9233645-9233645 on Build GRCh38
rs998176364 Chr.12: 9233805-9233805 on Build GRCh38
rs997944026 Chr.12: 9233432-9233432 on Build GRCh38
rs191300651 Chr.12: 9233758-9233758 on Build GRCh38
rs916149143 Chr.12: 9233516-9233516 on Build GRCh38
rs567679154 Chr.12: 9233791-9233791 on Build GRCh38
rs765929054 Chr.12: 9233737-9233737 on Build GRCh38
rs984053568 Chr.12: 9233654-9233654 on Build GRCh38
rs941194292 Chr.12: 9233755-9233755 on Build GRCh38
rs752444774 Chr.12: 9233457-9233457 on Build GRCh38
rs1036790212 Chr.12: 9233414-9233414 on Build GRCh38
rs780199130 Chr.12: 9233500-9233500 on Build GRCh38
rs888133324 Chr.12: 9233386-9233386 on Build GRCh38

View Full Product Details