Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 54608509-54609061 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
GLYCAM1
Entrez Gene ID
644076
Gene Name
glycosylation dependent cell adhesion molecule 1 (pseudogene)
Gene Aliases
-
UniGene
Hs.483519
Gene Chromosome Location
Chr.12: 54608187-54610462 on Build GRCh38
Transcripts
NR_003039.3

SNP Detail

SNP ID Chromosome Location
rs930719637 Chr.12: 54608641-54608641 on Build GRCh38
rs997705477 Chr.12: 54609032-54609032 on Build GRCh38
rs1042911751 Chr.12: 54608981-54608981 on Build GRCh38
rs938621666 Chr.12: 54608554-54608554 on Build GRCh38
rs909265622 Chr.12: 54608866-54608866 on Build GRCh38
rs953575152 Chr.12: 54608804-54608804 on Build GRCh38
rs764325923 Chr.12: 54608780-54608780 on Build GRCh38
rs937309 Chr.12: 54608944-54608944 on Build GRCh38
rs138477792 Chr.12: 54608990-54608990 on Build GRCh38
rs986276079 Chr.12: 54608833-54608833 on Build GRCh38
rs937310 Chr.12: 54608994-54608994 on Build GRCh38
rs559669959 Chr.12: 54608968-54608968 on Build GRCh38
rs1053189773 Chr.12: 54609019-54609019 on Build GRCh38
rs558678335 Chr.12: 54608943-54608943 on Build GRCh38
rs552726109 Chr.12: 54608629-54608629 on Build GRCh38
rs563176332 Chr.12: 54608995-54608995 on Build GRCh38
rs1495353 Chr.12: 54608747-54608747 on Build GRCh38
rs957142170 Chr.12: 54608762-54608762 on Build GRCh38
rs912152610 Chr.12: 54608618-54608618 on Build GRCh38
rs570717459 Chr.12: 54608664-54608664 on Build GRCh38
rs976338189 Chr.12: 54608771-54608771 on Build GRCh38
rs767527651 Chr.12: 54608565-54608565 on Build GRCh38
rs944973689 Chr.12: 54608911-54608911 on Build GRCh38
rs953776626 Chr.12: 54608839-54608839 on Build GRCh38
rs541305832 Chr.12: 54608953-54608953 on Build GRCh38
rs767886925 Chr.12: 54608915-54608915 on Build GRCh38
rs752566437 Chr.12: 54608569-54608569 on Build GRCh38
rs897699253 Chr.12: 54608547-54608547 on Build GRCh38
rs865893448 Chr.12: 54609027-54609027 on Build GRCh38
rs986311090 Chr.12: 54608564-54608564 on Build GRCh38
rs1030453578 Chr.12: 54608779-54608779 on Build GRCh38
rs115754483 Chr.12: 54608699-54608699 on Build GRCh38
rs932183175 Chr.12: 54608543-54608543 on Build GRCh38
rs998901884 Chr.12: 54608737-54608737 on Build GRCh38
rs74091330 Chr.12: 54609030-54609030 on Build GRCh38
rs553270631 Chr.12: 54608733-54608733 on Build GRCh38
rs1021411309 Chr.12: 54608644-54608644 on Build GRCh38
rs894297092 Chr.12: 54608956-54608956 on Build GRCh38
rs944962549 Chr.12: 54608624-54608624 on Build GRCh38
rs775537874 Chr.12: 54608626-54608626 on Build GRCh38
rs10876570 Chr.12: 54608886-54608886 on Build GRCh38
rs1049143531 Chr.12: 54608674-54608674 on Build GRCh38
rs11170988 Chr.12: 54608625-54608625 on Build GRCh38
rs150123718 Chr.12: 54608902-54608902 on Build GRCh38
rs557108337 Chr.12: 54608827-54608827 on Build GRCh38
rs140686299 Chr.12: 54608741-54608741 on Build GRCh38
rs1028066500 Chr.12: 54608834-54608834 on Build GRCh38
rs897808085 Chr.12: 54608639-54608639 on Build GRCh38
rs77141235 Chr.12: 54608974-54608974 on Build GRCh38
rs762447876 Chr.12: 54608894-54608894 on Build GRCh38
rs979056127 Chr.12: 54608820-54608820 on Build GRCh38
rs905270835 Chr.12: 54608683-54608683 on Build GRCh38

View Full Product Details