Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.12: 103883445-103883994 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
TTC41P
Entrez Gene ID
253724
Gene Name
tetratricopeptide repeat domain 41, pseudogene
Gene Aliases
GNN, GNNP
UniGene
Hs.506549
Gene Chromosome Location
Chr.12: 103843749-103930211 on Build GRCh38
Transcripts
NR_027249.1

SNP Detail

SNP ID Chromosome Location
rs1010109873 Chr.12: 103883764-103883764 on Build GRCh38
rs930328466 Chr.12: 103883688-103883688 on Build GRCh38
rs908631114 Chr.12: 103883952-103883952 on Build GRCh38
rs911749744 Chr.12: 103883624-103883624 on Build GRCh38
rs1046611869 Chr.12: 103883603-103883603 on Build GRCh38
rs982859789 Chr.12: 103883861-103883861 on Build GRCh38
rs778534335 Chr.12: 103883807-103883807 on Build GRCh38
rs879099401 Chr.12: 103883815-103883815 on Build GRCh38
rs931332733 Chr.12: 103883847-103883847 on Build GRCh38
rs2583237 Chr.12: 103883794-103883794 on Build GRCh38
rs949597530 Chr.12: 103883513-103883513 on Build GRCh38
rs1020465609 Chr.12: 103883699-103883699 on Build GRCh38
rs892829059 Chr.12: 103883672-103883672 on Build GRCh38
rs967202376 Chr.12: 103883705-103883705 on Build GRCh38
rs988423816 Chr.12: 103883507-103883507 on Build GRCh38
rs1000697462 Chr.12: 103883659-103883659 on Build GRCh38
rs1034000678 Chr.12: 103883665-103883665 on Build GRCh38
rs541788630 Chr.12: 103883887-103883887 on Build GRCh38
rs564768876 Chr.12: 103883962-103883962 on Build GRCh38
rs919170819 Chr.12: 103883660-103883660 on Build GRCh38
rs961135401 Chr.12: 103883493-103883493 on Build GRCh38
rs896660447 Chr.12: 103883756-103883756 on Build GRCh38
rs766935582 Chr.12: 103883532-103883532 on Build GRCh38
rs561695412 Chr.12: 103883841-103883841 on Build GRCh38
rs1000851719 Chr.12: 103883820-103883820 on Build GRCh38
rs887261986 Chr.12: 103883509-103883509 on Build GRCh38
rs1056587613 Chr.12: 103883734-103883734 on Build GRCh38
rs1047614434 Chr.12: 103883482-103883482 on Build GRCh38
rs555988413 Chr.12: 103883745-103883745 on Build GRCh38
rs1036282423 Chr.12: 103883651-103883651 on Build GRCh38
rs183901967 Chr.12: 103883604-103883604 on Build GRCh38
rs1002489200 Chr.12: 103883653-103883653 on Build GRCh38
rs921383521 Chr.12: 103883595-103883595 on Build GRCh38
rs11611305 Chr.12: 103883798-103883798 on Build GRCh38
rs76502009 Chr.12: 103883833-103883833 on Build GRCh38
rs968024500 Chr.12: 103883549-103883549 on Build GRCh38
rs539616950 Chr.12: 103883636-103883636 on Build GRCh38
rs572057093 Chr.12: 103883844-103883844 on Build GRCh38
rs553634488 Chr.12: 103883580-103883580 on Build GRCh38
rs757890185 Chr.12: 103883940-103883941 on Build GRCh38
rs779742168 Chr.12: 103883686-103883686 on Build GRCh38
rs963921169 Chr.12: 103883763-103883763 on Build GRCh38
rs968264845 Chr.12: 103883818-103883818 on Build GRCh38
rs987040617 Chr.12: 103883713-103883713 on Build GRCh38
rs749012667 Chr.12: 103883691-103883691 on Build GRCh38
rs192517119 Chr.12: 103883530-103883530 on Build GRCh38
rs1019482165 Chr.12: 103883754-103883754 on Build GRCh38
rs551987131 Chr.12: 103883647-103883647 on Build GRCh38
rs979399696 Chr.12: 103883588-103883588 on Build GRCh38

View Full Product Details