Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 19263184-19263747 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26P3
Entrez Gene ID
100101938
Gene Name
ankyrin repeat domain 26 pseudogene 3
Gene Aliases
LINC00414
UniGene
Hs.129283
Gene Chromosome Location
Chr.13: 19262800-19344973 on Build GRCh38
Transcripts
NR_027248.3

SNP Detail

SNP ID Chromosome Location
rs369973422 Chr.13: 19263470-19263470 on Build GRCh38
rs922130240 Chr.13: 19263525-19263525 on Build GRCh38
rs181493867 Chr.13: 19263522-19263522 on Build GRCh38
rs536425623 Chr.13: 19263613-19263613 on Build GRCh38
rs983191176 Chr.13: 19263467-19263467 on Build GRCh38
rs768036083 Chr.13: 19263337-19263337 on Build GRCh38
rs994515318 Chr.13: 19263445-19263445 on Build GRCh38
rs374973899 Chr.13: 19263608-19263608 on Build GRCh38
rs539288121 Chr.13: 19263579-19263579 on Build GRCh38
rs937500655 Chr.13: 19263584-19263584 on Build GRCh38
rs745533805 Chr.13: 19263283-19263283 on Build GRCh38
rs188665099 Chr.13: 19263464-19263464 on Build GRCh38
rs508921 Chr.13: 19263307-19263307 on Build GRCh38
rs946018354 Chr.13: 19263554-19263554 on Build GRCh38
rs922070852 Chr.13: 19263237-19263237 on Build GRCh38
rs138310322 Chr.13: 19263381-19263382 on Build GRCh38
rs1034961812 Chr.13: 19263619-19263619 on Build GRCh38
rs879671433 Chr.13: 19263297-19263298 on Build GRCh38
rs141401280 Chr.13: 19263574-19263574 on Build GRCh38
rs764025688 Chr.13: 19263632-19263632 on Build GRCh38
rs781248632 Chr.13: 19263686-19263686 on Build GRCh38
rs940390466 Chr.13: 19263572-19263572 on Build GRCh38
rs1043212176 Chr.13: 19263211-19263211 on Build GRCh38
rs1003462024 Chr.13: 19263431-19263431 on Build GRCh38
rs572624216 Chr.13: 19263250-19263250 on Build GRCh38
rs184765995 Chr.13: 19263217-19263217 on Build GRCh38
rs990081065 Chr.13: 19263550-19263551 on Build GRCh38
rs556097746 Chr.13: 19263703-19263703 on Build GRCh38
rs969600126 Chr.13: 19263404-19263404 on Build GRCh38
rs907399000 Chr.13: 19263338-19263338 on Build GRCh38
rs1009463614 Chr.13: 19263691-19263691 on Build GRCh38
rs1027772213 Chr.13: 19263447-19263447 on Build GRCh38
rs1025880731 Chr.13: 19263689-19263689 on Build GRCh38
rs932104475 Chr.13: 19263532-19263532 on Build GRCh38
rs935879309 Chr.13: 19263544-19263544 on Build GRCh38
rs537446675 Chr.13: 19263666-19263666 on Build GRCh38
rs911758328 Chr.13: 19263252-19263252 on Build GRCh38
rs1034930892 Chr.13: 19263442-19263442 on Build GRCh38
rs765487543 Chr.13: 19263571-19263571 on Build GRCh38
rs530754 Chr.13: 19263360-19263360 on Build GRCh38
rs1036104353 Chr.13: 19263359-19263359 on Build GRCh38
rs766762726 Chr.13: 19263371-19263371 on Build GRCh38
rs544187337 Chr.13: 19263310-19263310 on Build GRCh38
rs533207830 Chr.13: 19263399-19263399 on Build GRCh38
rs755967547 Chr.13: 19263722-19263722 on Build GRCh38
rs2297749 Chr.13: 19263594-19263594 on Build GRCh38
rs751543604 Chr.13: 19263637-19263637 on Build GRCh38
rs776932886 Chr.13: 19263449-19263449 on Build GRCh38
rs944528187 Chr.13: 19263274-19263274 on Build GRCh38
rs565967256 Chr.13: 19263603-19263603 on Build GRCh38
rs573362381 Chr.13: 19263646-19263646 on Build GRCh38
rs745694999 Chr.13: 19263700-19263700 on Build GRCh38
rs965870358 Chr.13: 19263496-19263496 on Build GRCh38
rs757091034 Chr.13: 19263671-19263671 on Build GRCh38
rs575572454 Chr.13: 19263304-19263304 on Build GRCh38
rs508914 Chr.13: 19263298-19263298 on Build GRCh38
rs73440250 Chr.13: 19263479-19263479 on Build GRCh38
rs912138640 Chr.13: 19263563-19263563 on Build GRCh38
rs1023487872 Chr.13: 19263393-19263393 on Build GRCh38
rs913342189 Chr.13: 19263507-19263507 on Build GRCh38
rs1045842419 Chr.13: 19263316-19263316 on Build GRCh38
rs531778580 Chr.13: 19263524-19263524 on Build GRCh38
rs147844929 Chr.13: 19263553-19263553 on Build GRCh38
rs2275230 Chr.13: 19263297-19263297 on Build GRCh38
rs888054625 Chr.13: 19263422-19263422 on Build GRCh38
rs746421326 Chr.13: 19263248-19263248 on Build GRCh38
rs773781273 Chr.13: 19263353-19263353 on Build GRCh38
rs1035971118 Chr.13: 19263573-19263573 on Build GRCh38
rs633378 Chr.13: 19263510-19263510 on Build GRCh38
rs946152527 Chr.13: 19263311-19263311 on Build GRCh38
rs145035750 Chr.13: 19263616-19263616 on Build GRCh38
rs1003368324 Chr.13: 19263609-19263609 on Build GRCh38
rs751507003 Chr.13: 19263595-19263595 on Build GRCh38
rs974652974 Chr.13: 19263296-19263296 on Build GRCh38

View Full Product Details