Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 19296611-19297163 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26P3
Entrez Gene ID
100101938
Gene Name
ankyrin repeat domain 26 pseudogene 3
Gene Aliases
LINC00414
UniGene
Hs.129283
Gene Chromosome Location
Chr.13: 19262800-19344973 on Build GRCh38
Transcripts
NR_027248.3

SNP Detail

SNP ID Chromosome Location
rs966744527 Chr.13: 19296947-19296947 on Build GRCh38
rs890801878 Chr.13: 19296789-19296789 on Build GRCh38
rs576293579 Chr.13: 19296980-19296980 on Build GRCh38
rs187587524 Chr.13: 19296936-19296936 on Build GRCh38
rs990900389 Chr.13: 19297035-19297035 on Build GRCh38
rs777881654 Chr.13: 19296995-19296995 on Build GRCh38
rs191502574 Chr.13: 19296846-19296846 on Build GRCh38
rs984893982 Chr.13: 19297054-19297054 on Build GRCh38
rs781013452 Chr.13: 19297011-19297011 on Build GRCh38
rs756737450 Chr.13: 19296658-19296658 on Build GRCh38
rs149597493 Chr.13: 19296795-19296795 on Build GRCh38
rs979554606 Chr.13: 19296714-19296714 on Build GRCh38
rs564723387 Chr.13: 19296654-19296654 on Build GRCh38
rs978105930 Chr.13: 19296694-19296694 on Build GRCh38
rs532510118 Chr.13: 19296655-19296655 on Build GRCh38
rs940912730 Chr.13: 19297059-19297059 on Build GRCh38
rs776819851 Chr.13: 19297114-19297114 on Build GRCh38
rs1033677317 Chr.13: 19296687-19296687 on Build GRCh38
rs1021066376 Chr.13: 19296946-19296946 on Build GRCh38
rs528512684 Chr.13: 19296744-19296745 on Build GRCh38
rs1036675236 Chr.13: 19296801-19296801 on Build GRCh38
rs142625657 Chr.13: 19297092-19297092 on Build GRCh38
rs564784479 Chr.13: 19297064-19297064 on Build GRCh38
rs945560857 Chr.13: 19296703-19296703 on Build GRCh38
rs1038349349 Chr.13: 19296911-19296911 on Build GRCh38
rs900869765 Chr.13: 19297130-19297130 on Build GRCh38
rs1034990386 Chr.13: 19296986-19296986 on Build GRCh38
rs912778935 Chr.13: 19297038-19297038 on Build GRCh38
rs538745002 Chr.13: 19296854-19296854 on Build GRCh38
rs905812284 Chr.13: 19296866-19296866 on Build GRCh38
rs1001460587 Chr.13: 19296889-19296889 on Build GRCh38
rs17228582 Chr.13: 19296728-19296728 on Build GRCh38
rs796704065 Chr.13: 19296733-19296733 on Build GRCh38
rs896895105 Chr.13: 19296818-19296818 on Build GRCh38
rs1028120175 Chr.13: 19296675-19296675 on Build GRCh38
rs148736622 Chr.13: 19296988-19296988 on Build GRCh38
rs182898631 Chr.13: 19296910-19296910 on Build GRCh38
rs980807341 Chr.13: 19296662-19296662 on Build GRCh38
rs78305686 Chr.13: 19296698-19296698 on Build GRCh38
rs953951534 Chr.13: 19296676-19296676 on Build GRCh38
rs1028067749 Chr.13: 19296872-19296872 on Build GRCh38
rs990952626 Chr.13: 19296719-19296719 on Build GRCh38
rs748517773 Chr.13: 19296955-19296955 on Build GRCh38
rs985750565 Chr.13: 19297024-19297024 on Build GRCh38
rs188462920 Chr.13: 19296693-19296693 on Build GRCh38
rs1036797289 Chr.13: 19296809-19296809 on Build GRCh38
rs908136819 Chr.13: 19297058-19297058 on Build GRCh38
rs559226967 Chr.13: 19296839-19296839 on Build GRCh38
rs989431240 Chr.13: 19296679-19296679 on Build GRCh38
rs760560818 Chr.13: 19297122-19297122 on Build GRCh38
rs771026476 Chr.13: 19297042-19297042 on Build GRCh38
rs554529793 Chr.13: 19296959-19296959 on Build GRCh38
rs144275024 Chr.13: 19296927-19296927 on Build GRCh38
rs182966376 Chr.13: 19296691-19296691 on Build GRCh38
rs960756541 Chr.13: 19296924-19296924 on Build GRCh38
rs747212044 Chr.13: 19297011-19297011 on Build GRCh38
rs978517016 Chr.13: 19296956-19296956 on Build GRCh38
rs570224195 Chr.13: 19296753-19296753 on Build GRCh38

View Full Product Details