Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 19309220-19309788 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26P3
Entrez Gene ID
100101938
Gene Name
ankyrin repeat domain 26 pseudogene 3
Gene Aliases
LINC00414
UniGene
Hs.129283
Gene Chromosome Location
Chr.13: 19262800-19344973 on Build GRCh38
Transcripts
NR_027248.3

SNP Detail

SNP ID Chromosome Location
rs889767884 Chr.13: 19309472-19309472 on Build GRCh38
rs1010981978 Chr.13: 19309473-19309473 on Build GRCh38
rs115946940 Chr.13: 19309317-19309317 on Build GRCh38
rs956569788 Chr.13: 19309325-19309325 on Build GRCh38
rs946683299 Chr.13: 19309734-19309734 on Build GRCh38
rs764078457 Chr.13: 19309389-19309389 on Build GRCh38
rs911966487 Chr.13: 19309461-19309461 on Build GRCh38
rs1037360047 Chr.13: 19309378-19309378 on Build GRCh38
rs572510316 Chr.13: 19309351-19309351 on Build GRCh38
rs755540258 Chr.13: 19309677-19309677 on Build GRCh38
rs558942494 Chr.13: 19309328-19309328 on Build GRCh38
rs529615399 Chr.13: 19309442-19309442 on Build GRCh38
rs1012490685 Chr.13: 19309558-19309558 on Build GRCh38
rs915676459 Chr.13: 19309255-19309255 on Build GRCh38
rs967985202 Chr.13: 19309361-19309361 on Build GRCh38
rs975699985 Chr.13: 19309379-19309379 on Build GRCh38
rs1003266005 Chr.13: 19309295-19309295 on Build GRCh38
rs899444979 Chr.13: 19309514-19309514 on Build GRCh38
rs1003149456 Chr.13: 19309578-19309578 on Build GRCh38
rs1028628298 Chr.13: 19309745-19309745 on Build GRCh38
rs1036529217 Chr.13: 19309486-19309486 on Build GRCh38
rs1022138005 Chr.13: 19309655-19309655 on Build GRCh38
rs1016645229 Chr.13: 19309638-19309638 on Build GRCh38
rs985343922 Chr.13: 19309718-19309718 on Build GRCh38
rs570504281 Chr.13: 19309653-19309653 on Build GRCh38
rs940863076 Chr.13: 19309464-19309464 on Build GRCh38
rs111680332 Chr.13: 19309559-19309559 on Build GRCh38
rs1044200702 Chr.13: 19309735-19309735 on Build GRCh38
rs896710739 Chr.13: 19309545-19309545 on Build GRCh38
rs922522769 Chr.13: 19309384-19309384 on Build GRCh38
rs773827793 Chr.13: 19309346-19309346 on Build GRCh38
rs753113169 Chr.13: 19309753-19309753 on Build GRCh38
rs553469195 Chr.13: 19309738-19309738 on Build GRCh38
rs934008627 Chr.13: 19309386-19309386 on Build GRCh38
rs969823064 Chr.13: 19309259-19309259 on Build GRCh38
rs142381629 Chr.13: 19309722-19309722 on Build GRCh38
rs922416302 Chr.13: 19309679-19309679 on Build GRCh38
rs931950561 Chr.13: 19309406-19309406 on Build GRCh38
rs969704931 Chr.13: 19309565-19309565 on Build GRCh38
rs765733053 Chr.13: 19309704-19309704 on Build GRCh38
rs1056662894 Chr.13: 19309755-19309755 on Build GRCh38
rs1034757549 Chr.13: 19309634-19309634 on Build GRCh38
rs892215786 Chr.13: 19309635-19309635 on Build GRCh38
rs767054819 Chr.13: 19309608-19309608 on Build GRCh38
rs138401083 Chr.13: 19309537-19309537 on Build GRCh38
rs902686825 Chr.13: 19309530-19309530 on Build GRCh38
rs961523275 Chr.13: 19309649-19309649 on Build GRCh38
rs915183883 Chr.13: 19309339-19309339 on Build GRCh38
rs1806658 Chr.13: 19309532-19309532 on Build GRCh38
rs552393305 Chr.13: 19309595-19309595 on Build GRCh38
rs574906314 Chr.13: 19309761-19309761 on Build GRCh38
rs911253734 Chr.13: 19309725-19309725 on Build GRCh38
rs987564741 Chr.13: 19309446-19309446 on Build GRCh38

View Full Product Details