Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 19333151-19333728 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26P3
Entrez Gene ID
100101938
Gene Name
ankyrin repeat domain 26 pseudogene 3
Gene Aliases
LINC00414
UniGene
Hs.129283
Gene Chromosome Location
Chr.13: 19262800-19344973 on Build GRCh38
Transcripts
NR_027248.3

SNP Detail

SNP ID Chromosome Location
rs960516879 Chr.13: 19333494-19333494 on Build GRCh38
rs970256821 Chr.13: 19333563-19333563 on Build GRCh38
rs972237687 Chr.13: 19333386-19333386 on Build GRCh38
rs190760575 Chr.13: 19333694-19333694 on Build GRCh38
rs1043417135 Chr.13: 19333435-19333435 on Build GRCh38
rs908840719 Chr.13: 19333576-19333576 on Build GRCh38
rs972246820 Chr.13: 19333584-19333584 on Build GRCh38
rs767387713 Chr.13: 19333234-19333234 on Build GRCh38
rs1977705 Chr.13: 19333575-19333575 on Build GRCh38
rs904547351 Chr.13: 19333698-19333700 on Build GRCh38
rs528621642 Chr.13: 19333433-19333433 on Build GRCh38
rs369581370 Chr.13: 19333530-19333530 on Build GRCh38
rs947732904 Chr.13: 19333228-19333228 on Build GRCh38
rs904620658 Chr.13: 19333443-19333443 on Build GRCh38
rs56091297 Chr.13: 19333434-19333434 on Build GRCh38
rs185987230 Chr.13: 19333319-19333319 on Build GRCh38
rs933194835 Chr.13: 19333627-19333627 on Build GRCh38
rs960340200 Chr.13: 19333195-19333195 on Build GRCh38
rs757313511 Chr.13: 19333187-19333187 on Build GRCh38
rs375246235 Chr.13: 19333526-19333526 on Build GRCh38
rs755983225 Chr.13: 19333387-19333387 on Build GRCh38
rs141389485 Chr.13: 19333629-19333629 on Build GRCh38
rs1023259267 Chr.13: 19333551-19333551 on Build GRCh38
rs919060355 Chr.13: 19333419-19333419 on Build GRCh38
rs910477949 Chr.13: 19333634-19333634 on Build GRCh38
rs547830310 Chr.13: 19333333-19333333 on Build GRCh38
rs145037955 Chr.13: 19333643-19333643 on Build GRCh38
rs181045986 Chr.13: 19333205-19333205 on Build GRCh38
rs980429315 Chr.13: 19333251-19333251 on Build GRCh38
rs145648530 Chr.13: 19333204-19333204 on Build GRCh38
rs1043082260 Chr.13: 19333675-19333675 on Build GRCh38
rs554226177 Chr.13: 19333238-19333239 on Build GRCh38
rs566052719 Chr.13: 19333341-19333341 on Build GRCh38
rs185949850 Chr.13: 19333586-19333586 on Build GRCh38
rs183083697 Chr.13: 19333529-19333529 on Build GRCh38
rs750416466 Chr.13: 19333285-19333285 on Build GRCh38
rs918999903 Chr.13: 19333612-19333612 on Build GRCh38
rs117875474 Chr.13: 19333448-19333448 on Build GRCh38
rs759619487 Chr.13: 19333651-19333651 on Build GRCh38
rs943262733 Chr.13: 19333665-19333665 on Build GRCh38
rs916154449 Chr.13: 19333215-19333215 on Build GRCh38
rs191231639 Chr.13: 19333430-19333430 on Build GRCh38
rs963177767 Chr.13: 19333583-19333583 on Build GRCh38
rs555012270 Chr.13: 19333653-19333653 on Build GRCh38
rs201205105 Chr.13: 19333480-19333481 on Build GRCh38
rs941740018 Chr.13: 19333336-19333336 on Build GRCh38

View Full Product Details