Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 22897836-22898195 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
BASP1P1
Entrez Gene ID
646201
Gene Name
brain abundant, membrane attached signal protein 1 pseudogene 1
Gene Aliases
-
UniGene
Hs.729338
Gene Chromosome Location
Chr.13: 22897030-22898181 on Build GRCh38
Transcripts
NR_033774.1

SNP Detail

SNP ID Chromosome Location
rs982991726 Chr.13: 22898007-22898007 on Build GRCh38
rs558669889 Chr.13: 22897964-22897964 on Build GRCh38
rs548086729 Chr.13: 22898155-22898155 on Build GRCh38
rs955332424 Chr.13: 22898132-22898132 on Build GRCh38
rs1042789509 Chr.13: 22898115-22898115 on Build GRCh38
rs985646885 Chr.13: 22898096-22898096 on Build GRCh38
rs369002468 Chr.13: 22898017-22898017 on Build GRCh38
rs933986176 Chr.13: 22898003-22898003 on Build GRCh38
rs374809599 Chr.13: 22897944-22897944 on Build GRCh38
rs990018699 Chr.13: 22897949-22897949 on Build GRCh38
rs539316322 Chr.13: 22897862-22897862 on Build GRCh38
rs998722229 Chr.13: 22897899-22897899 on Build GRCh38
rs908809902 Chr.13: 22898010-22898010 on Build GRCh38
rs576569725 Chr.13: 22897925-22897925 on Build GRCh38
rs1033249701 Chr.13: 22898008-22898008 on Build GRCh38
rs1002509343 Chr.13: 22898145-22898145 on Build GRCh38
rs922589475 Chr.13: 22897984-22897984 on Build GRCh38
rs565466399 Chr.13: 22898130-22898130 on Build GRCh38
rs1045491631 Chr.13: 22898051-22898051 on Build GRCh38
rs7328194 Chr.13: 22898129-22898129 on Build GRCh38
rs566062777 Chr.13: 22897865-22897865 on Build GRCh38
rs1050006207 Chr.13: 22897883-22897883 on Build GRCh38
rs879818272 Chr.13: 22898001-22898001 on Build GRCh38
rs866215611 Chr.13: 22897998-22897998 on Build GRCh38
rs536672003 Chr.13: 22897900-22897900 on Build GRCh38
rs150634170 Chr.13: 22898015-22898015 on Build GRCh38
rs767191458 Chr.13: 22897941-22897941 on Build GRCh38
rs964142074 Chr.13: 22897961-22897961 on Build GRCh38
rs894659907 Chr.13: 22898049-22898049 on Build GRCh38
rs998662446 Chr.13: 22897971-22897971 on Build GRCh38
rs555022321 Chr.13: 22897916-22897916 on Build GRCh38
rs892963492 Chr.13: 22898133-22898133 on Build GRCh38
rs559259275 Chr.13: 22898079-22898079 on Build GRCh38
rs1042450745 Chr.13: 22897946-22897946 on Build GRCh38
rs867433604 Chr.13: 22897934-22897934 on Build GRCh38
rs932846691 Chr.13: 22898081-22898081 on Build GRCh38
rs975533969 Chr.13: 22897979-22897979 on Build GRCh38
rs566346463 Chr.13: 22898171-22898171 on Build GRCh38
rs971025575 Chr.13: 22898026-22898026 on Build GRCh38
rs1022866917 Chr.13: 22898143-22898143 on Build GRCh38
rs61958793 Chr.13: 22898050-22898050 on Build GRCh38
rs964935727 Chr.13: 22898108-22898108 on Build GRCh38
rs562961761 Chr.13: 22898107-22898107 on Build GRCh38
rs574073878 Chr.13: 22898101-22898101 on Build GRCh38
rs907124045 Chr.13: 22898144-22898144 on Build GRCh38
rs1026746423 Chr.13: 22898170-22898170 on Build GRCh38
rs530587517 Chr.13: 22898122-22898122 on Build GRCh38
rs532629201 Chr.13: 22898150-22898150 on Build GRCh38
rs1031526412 Chr.13: 22898086-22898086 on Build GRCh38
rs547815162 Chr.13: 22897863-22897863 on Build GRCh38
rs982792256 Chr.13: 22898137-22898137 on Build GRCh38
rs1017883280 Chr.13: 22897959-22897959 on Build GRCh38
rs998480219 Chr.13: 22898080-22898080 on Build GRCh38
rs944001279 Chr.13: 22898112-22898112 on Build GRCh38
rs932897585 Chr.13: 22897866-22897866 on Build GRCh38
rs541398244 Chr.13: 22898102-22898102 on Build GRCh38
rs557062424 Chr.13: 22898048-22898048 on Build GRCh38
rs891111819 Chr.13: 22897931-22897931 on Build GRCh38
rs541312253 Chr.13: 22898059-22898059 on Build GRCh38

View Full Product Details