Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.13: 75238231-75238776 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CTAGE11P
Entrez Gene ID
647288
Gene Name
CTAGE family member 11, pseudogene
Gene Aliases
-
UniGene
Hs.567920
Gene Chromosome Location
Chr.13: 75237753-75240381 on Build GRCh38
Transcripts
NR_027466.1

SNP Detail

SNP ID Chromosome Location
rs941398223 Chr.13: 75238399-75238399 on Build GRCh38
rs945566002 Chr.13: 75238482-75238482 on Build GRCh38
rs564553731 Chr.13: 75238284-75238284 on Build GRCh38
rs950617086 Chr.13: 75238329-75238330 on Build GRCh38
rs565231405 Chr.13: 75238668-75238668 on Build GRCh38
rs533491017 Chr.13: 75238368-75238368 on Build GRCh38
rs1013219962 Chr.13: 75238655-75238655 on Build GRCh38
rs975439880 Chr.13: 75238404-75238404 on Build GRCh38
rs143272034 Chr.13: 75238731-75238731 on Build GRCh38
rs776810544 Chr.13: 75238581-75238581 on Build GRCh38
rs555846 Chr.13: 75238271-75238271 on Build GRCh38
rs909987677 Chr.13: 75238382-75238382 on Build GRCh38
rs1001888172 Chr.13: 75238599-75238599 on Build GRCh38
rs747386959 Chr.13: 75238352-75238352 on Build GRCh38
rs574275311 Chr.13: 75238442-75238442 on Build GRCh38
rs149160873 Chr.13: 75238672-75238672 on Build GRCh38
rs892677879 Chr.13: 75238463-75238463 on Build GRCh38
rs934294045 Chr.13: 75238423-75238423 on Build GRCh38
rs570218764 Chr.13: 75238324-75238324 on Build GRCh38
rs114853755 Chr.13: 75238667-75238667 on Build GRCh38
rs796421824 Chr.13: 75238616-75238616 on Build GRCh38
rs575413629 Chr.13: 75238647-75238647 on Build GRCh38
rs1026183405 Chr.13: 75238273-75238273 on Build GRCh38
rs984585369 Chr.13: 75238329-75238329 on Build GRCh38
rs557241989 Chr.13: 75238630-75238630 on Build GRCh38
rs748574776 Chr.13: 75238583-75238583 on Build GRCh38
rs577454655 Chr.13: 75238669-75238669 on Build GRCh38
rs143373524 Chr.13: 75238374-75238374 on Build GRCh38
rs187032979 Chr.13: 75238619-75238619 on Build GRCh38
rs1016091878 Chr.13: 75238726-75238726 on Build GRCh38
rs552787397 Chr.13: 75238416-75238416 on Build GRCh38
rs866896500 Chr.13: 75238628-75238628 on Build GRCh38
rs111548055 Chr.13: 75238496-75238496 on Build GRCh38
rs558583709 Chr.13: 75238567-75238567 on Build GRCh38

View Full Product Details