Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.14: 70469496-70470041 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ADAM20P1
Entrez Gene ID
317760
Gene Name
ADAM metallopeptidase domain 20 pseudogene 1
Gene Aliases
C14orf55
UniGene
Hs.188369
Gene Chromosome Location
Chr.14: 70468876-70483756 on Build GRCh38
Transcripts
NR_037933.1

SNP Detail

SNP ID Chromosome Location
rs562371947 Chr.14: 70469614-70469614 on Build GRCh38
rs184939319 Chr.14: 70469552-70469552 on Build GRCh38
rs925380195 Chr.14: 70469980-70469980 on Build GRCh38
rs972618174 Chr.14: 70469643-70469643 on Build GRCh38
rs534963915 Chr.14: 70469723-70469723 on Build GRCh38
rs1025503671 Chr.14: 70469865-70469865 on Build GRCh38
rs372023058 Chr.14: 70469862-70469862 on Build GRCh38
rs999743655 Chr.14: 70469561-70469561 on Build GRCh38
rs374718817 Chr.14: 70469517-70469517 on Build GRCh38
rs925466010 Chr.14: 70469761-70469761 on Build GRCh38
rs116498676 Chr.14: 70469615-70469615 on Build GRCh38
rs1016730440 Chr.14: 70469633-70469633 on Build GRCh38
rs1052668353 Chr.14: 70469772-70469772 on Build GRCh38
rs1025556196 Chr.14: 70469647-70469647 on Build GRCh38
rs569094137 Chr.14: 70469543-70469543 on Build GRCh38
rs898332513 Chr.14: 70469843-70469843 on Build GRCh38
rs904022426 Chr.14: 70469550-70469550 on Build GRCh38
rs548823109 Chr.14: 70469934-70469934 on Build GRCh38
rs34620293 Chr.14: 70469999-70470000 on Build GRCh38
rs2293875 Chr.14: 70469917-70469917 on Build GRCh38
rs35905405 Chr.14: 70469780-70469781 on Build GRCh38
rs986848895 Chr.14: 70469666-70469666 on Build GRCh38
rs905532019 Chr.14: 70469801-70469801 on Build GRCh38
rs531851314 Chr.14: 70469590-70469590 on Build GRCh38
rs573961329 Chr.14: 70469529-70469529 on Build GRCh38
rs544092513 Chr.14: 70469591-70469591 on Build GRCh38
rs1001259059 Chr.14: 70469816-70469816 on Build GRCh38
rs955263613 Chr.14: 70469907-70469907 on Build GRCh38
rs987124131 Chr.14: 70469914-70469914 on Build GRCh38
rs138046198 Chr.14: 70469818-70469818 on Build GRCh38
rs565754697 Chr.14: 70469638-70469638 on Build GRCh38
rs949624573 Chr.14: 70469998-70469998 on Build GRCh38
rs993987244 Chr.14: 70469845-70469845 on Build GRCh38
rs572382526 Chr.14: 70470017-70470017 on Build GRCh38
rs533027136 Chr.14: 70469657-70469657 on Build GRCh38
rs535428541 Chr.14: 70469984-70469984 on Build GRCh38
rs569826813 Chr.14: 70469792-70469792 on Build GRCh38
rs911231262 Chr.14: 70469677-70469677 on Build GRCh38
rs1043876474 Chr.14: 70469547-70469547 on Build GRCh38
rs935471746 Chr.14: 70469993-70469993 on Build GRCh38
rs949626967 Chr.14: 70469785-70469785 on Build GRCh38
rs2293873 Chr.14: 70469997-70469997 on Build GRCh38
rs564686333 Chr.14: 70469575-70469575 on Build GRCh38
rs988278014 Chr.14: 70469994-70469994 on Build GRCh38
rs942760943 Chr.14: 70469686-70469686 on Build GRCh38
rs935523963 Chr.14: 70469763-70469763 on Build GRCh38
rs369975147 Chr.14: 70469564-70469564 on Build GRCh38
rs962527188 Chr.14: 70469634-70469634 on Build GRCh38
rs547433280 Chr.14: 70469622-70469622 on Build GRCh38
rs1013951208 Chr.14: 70469609-70469609 on Build GRCh38
rs896767935 Chr.14: 70469774-70469774 on Build GRCh38
rs747340363 Chr.14: 70469967-70469967 on Build GRCh38
rs2293874 Chr.14: 70469955-70469955 on Build GRCh38

View Full Product Details