Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40531572-40532119 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf57
Entrez Gene ID
90416
Gene Name
chromosome 15 open reading frame 57
Gene Aliases
CCDC32
UniGene
Hs.654661
Gene Chromosome Location
Chr.15: 40531341-40565057 on Build GRCh38
Transcripts
NM_001289132.1 NR_002208.1

Gene Symbol
MRPL42P5
Entrez Gene ID
359821
Gene Name
mitochondrial ribosomal protein L42 pseudogene 5
Gene Aliases
-
UniGene
Hs.678598
Gene Chromosome Location
Chr.15: 40531884-40532550 on Build GRCh38
Transcripts
NM_001289132.1 NR_002208.1

SNP Detail

SNP ID Chromosome Location
rs780283966 Chr.15: 40531617-40531617 on Build GRCh38
rs201876664 Chr.15: 40531615-40531617 on Build GRCh38
rs775479584 Chr.15: 40532052-40532052 on Build GRCh38
rs1013928250 Chr.15: 40531901-40531903 on Build GRCh38
rs747319454 Chr.15: 40531633-40531633 on Build GRCh38
rs554500728 Chr.15: 40531717-40531717 on Build GRCh38
rs560822021 Chr.15: 40532065-40532065 on Build GRCh38
rs575400352 Chr.15: 40531973-40531973 on Build GRCh38
rs999249276 Chr.15: 40532043-40532043 on Build GRCh38
rs934462476 Chr.15: 40531883-40531884 on Build GRCh38
rs576265656 Chr.15: 40531716-40531716 on Build GRCh38
rs963554980 Chr.15: 40532006-40532006 on Build GRCh38
rs530039593 Chr.15: 40532072-40532072 on Build GRCh38
rs1053079974 Chr.15: 40531884-40531884 on Build GRCh38
rs4923875 Chr.15: 40531719-40531719 on Build GRCh38
rs540656797 Chr.15: 40532021-40532021 on Build GRCh38
rs930050072 Chr.15: 40531857-40531857 on Build GRCh38
rs190440947 Chr.15: 40531805-40531805 on Build GRCh38
rs564875767 Chr.15: 40531926-40531926 on Build GRCh38
rs911088564 Chr.15: 40531804-40531804 on Build GRCh38
rs1022746517 Chr.15: 40531647-40531647 on Build GRCh38
rs1005289532 Chr.15: 40531946-40531946 on Build GRCh38
rs907463785 Chr.15: 40531636-40531636 on Build GRCh38
rs558821504 Chr.15: 40531604-40531604 on Build GRCh38
rs903612537 Chr.15: 40532028-40532028 on Build GRCh38
rs770329502 Chr.15: 40531772-40531772 on Build GRCh38
rs1044412986 Chr.15: 40531942-40531942 on Build GRCh38
rs969743625 Chr.15: 40531654-40531654 on Build GRCh38
rs113509274 Chr.15: 40531886-40531887 on Build GRCh38
rs74335200 Chr.15: 40531818-40531818 on Build GRCh38
rs986677139 Chr.15: 40531803-40531803 on Build GRCh38
rs987845347 Chr.15: 40531801-40531801 on Build GRCh38
rs978178850 Chr.15: 40531736-40531736 on Build GRCh38
rs920024952 Chr.15: 40531829-40531829 on Build GRCh38
rs577398279 Chr.15: 40531854-40531854 on Build GRCh38
rs537897638 Chr.15: 40531606-40531606 on Build GRCh38
rs1012034635 Chr.15: 40531688-40531688 on Build GRCh38
rs758831786 Chr.15: 40531685-40531685 on Build GRCh38
rs866954746 Chr.15: 40531928-40531928 on Build GRCh38
rs938968107 Chr.15: 40531894-40531894 on Build GRCh38
rs905634619 Chr.15: 40531945-40531945 on Build GRCh38
rs748836928 Chr.15: 40531724-40531724 on Build GRCh38
rs2034653 Chr.15: 40531883-40531883 on Build GRCh38
rs894673333 Chr.15: 40531977-40531977 on Build GRCh38
rs569314756 Chr.15: 40531605-40531605 on Build GRCh38
rs919500435 Chr.15: 40531784-40531784 on Build GRCh38
rs1030637787 Chr.15: 40531788-40531788 on Build GRCh38
rs1016359629 Chr.15: 40531696-40531696 on Build GRCh38
rs894642161 Chr.15: 40531687-40531687 on Build GRCh38
rs558367660 Chr.15: 40531799-40531799 on Build GRCh38
rs942524937 Chr.15: 40531808-40531808 on Build GRCh38
rs1003094792 Chr.15: 40531663-40531663 on Build GRCh38
rs1047515801 Chr.15: 40531882-40531882 on Build GRCh38
rs554819247 Chr.15: 40531640-40531640 on Build GRCh38
rs183389793 Chr.15: 40532017-40532017 on Build GRCh38
rs964694107 Chr.15: 40532064-40532064 on Build GRCh38
rs755538221 Chr.15: 40531668-40531668 on Build GRCh38
rs895712378 Chr.15: 40531884-40531884 on Build GRCh38

View Full Product Details