Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 40531993-40532542 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
C15orf57
Entrez Gene ID
90416
Gene Name
chromosome 15 open reading frame 57
Gene Aliases
CCDC32
UniGene
Hs.654661
Gene Chromosome Location
Chr.15: 40531341-40565057 on Build GRCh38
Transcripts
NM_001289132.1 NR_002208.1

Gene Symbol
MRPL42P5
Entrez Gene ID
359821
Gene Name
mitochondrial ribosomal protein L42 pseudogene 5
Gene Aliases
-
UniGene
Hs.678598
Gene Chromosome Location
Chr.15: 40531884-40532550 on Build GRCh38
Transcripts
NM_001289132.1 NR_002208.1

SNP Detail

SNP ID Chromosome Location
rs973892700 Chr.15: 40532149-40532149 on Build GRCh38
rs775479584 Chr.15: 40532052-40532052 on Build GRCh38
rs1018130251 Chr.15: 40532114-40532114 on Build GRCh38
rs757367605 Chr.15: 40532461-40532461 on Build GRCh38
rs560822021 Chr.15: 40532065-40532065 on Build GRCh38
rs999249276 Chr.15: 40532043-40532043 on Build GRCh38
rs1027275409 Chr.15: 40532499-40532499 on Build GRCh38
rs778633312 Chr.15: 40532251-40532251 on Build GRCh38
rs963939387 Chr.15: 40532444-40532444 on Build GRCh38
rs532238431 Chr.15: 40532213-40532213 on Build GRCh38
rs761760035 Chr.15: 40532430-40532430 on Build GRCh38
rs568077418 Chr.15: 40532319-40532319 on Build GRCh38
rs779751086 Chr.15: 40532272-40532272 on Build GRCh38
rs560383322 Chr.15: 40532121-40532121 on Build GRCh38
rs949980838 Chr.15: 40532166-40532166 on Build GRCh38
rs954996298 Chr.15: 40532381-40532381 on Build GRCh38
rs530039593 Chr.15: 40532072-40532072 on Build GRCh38
rs554195963 Chr.15: 40532355-40532355 on Build GRCh38
rs760538311 Chr.15: 40532140-40532140 on Build GRCh38
rs1009590448 Chr.15: 40532390-40532390 on Build GRCh38
rs878949054 Chr.15: 40532505-40532505 on Build GRCh38
rs867156174 Chr.15: 40532397-40532397 on Build GRCh38
rs539684520 Chr.15: 40532396-40532396 on Build GRCh38
rs56046472 Chr.15: 40532375-40532375 on Build GRCh38
rs540656797 Chr.15: 40532021-40532021 on Build GRCh38
rs113542465 Chr.15: 40532515-40532515 on Build GRCh38
rs930056240 Chr.15: 40532210-40532210 on Build GRCh38
rs75323018 Chr.15: 40532113-40532113 on Build GRCh38
rs919737642 Chr.15: 40532200-40532200 on Build GRCh38
rs903612537 Chr.15: 40532028-40532028 on Build GRCh38
rs999716668 Chr.15: 40532341-40532341 on Build GRCh38
rs567665497 Chr.15: 40532225-40532225 on Build GRCh38
rs192354843 Chr.15: 40532337-40532337 on Build GRCh38
rs896876847 Chr.15: 40532343-40532343 on Build GRCh38
rs768749972 Chr.15: 40532306-40532306 on Build GRCh38
rs995346843 Chr.15: 40532451-40532451 on Build GRCh38
rs758110110 Chr.15: 40532262-40532262 on Build GRCh38
rs75117134 Chr.15: 40532288-40532288 on Build GRCh38
rs566802902 Chr.15: 40532472-40532472 on Build GRCh38
rs575415201 Chr.15: 40532514-40532514 on Build GRCh38
rs8029208 Chr.15: 40532511-40532511 on Build GRCh38
rs746495288 Chr.15: 40532291-40532291 on Build GRCh38
rs569051842 Chr.15: 40532230-40532230 on Build GRCh38
rs796345392 Chr.15: 40532245-40532245 on Build GRCh38
rs903311387 Chr.15: 40532320-40532320 on Build GRCh38
rs745460671 Chr.15: 40532253-40532253 on Build GRCh38
rs964694107 Chr.15: 40532064-40532064 on Build GRCh38
rs144060153 Chr.15: 40532219-40532219 on Build GRCh38
rs926022595 Chr.15: 40532102-40532102 on Build GRCh38
rs765262738 Chr.15: 40532463-40532463 on Build GRCh38
rs947589929 Chr.15: 40532310-40532310 on Build GRCh38
rs189049717 Chr.15: 40532265-40532265 on Build GRCh38

View Full Product Details