Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 43875769-43876324 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
FRMD5
Entrez Gene ID
84978
Gene Name
FERM domain containing 5
Gene Aliases
-
UniGene
Hs.578544
Gene Chromosome Location
Chr.15: 43870761-44195294 on Build GRCh38
Transcripts
NM_001286490.1 NM_001286491.1 NM_001322949.1 NM_001322950.1 NM_001322951.1 NM_032892.4 NR_104455.1 XM_005254730.3 NR_003571.1

Gene Symbol
PIN4P1
Entrez Gene ID
728758
Gene Name
peptidylprolyl cis/trans isomerase, NIMA-interacting 4 pseudogene 1
Gene Aliases
-
UniGene
Hs.658099
Gene Chromosome Location
Chr.15: 43875806-43878171 on Build GRCh38
Transcripts
NM_001286490.1 NM_001286491.1 NM_001322949.1 NM_001322950.1 NM_001322951.1 NM_032892.4 NR_104455.1 XM_005254730.3 NR_003571.1

SNP Detail

SNP ID Chromosome Location
rs1018120379 Chr.15: 43876212-43876212 on Build GRCh38
rs879096491 Chr.15: 43875856-43875856 on Build GRCh38
rs1025618843 Chr.15: 43875805-43875812 on Build GRCh38
rs760877318 Chr.15: 43875905-43875905 on Build GRCh38
rs1050759775 Chr.15: 43876195-43876195 on Build GRCh38
rs878996051 Chr.15: 43876035-43876035 on Build GRCh38
rs559155917 Chr.15: 43876183-43876183 on Build GRCh38
rs530014409 Chr.15: 43876004-43876004 on Build GRCh38
rs367918994 Chr.15: 43875828-43875828 on Build GRCh38
rs1002110828 Chr.15: 43875977-43875977 on Build GRCh38
rs981930180 Chr.15: 43876282-43876282 on Build GRCh38
rs981493004 Chr.15: 43875972-43875972 on Build GRCh38
rs778781823 Chr.15: 43875975-43875975 on Build GRCh38
rs1018773028 Chr.15: 43876063-43876063 on Build GRCh38
rs1029732295 Chr.15: 43875879-43875879 on Build GRCh38
rs942519571 Chr.15: 43875827-43875827 on Build GRCh38
rs879134901 Chr.15: 43876197-43876197 on Build GRCh38
rs868633489 Chr.15: 43875815-43875815 on Build GRCh38
rs911086253 Chr.15: 43875809-43875809 on Build GRCh38
rs539322559 Chr.15: 43876158-43876158 on Build GRCh38
rs935216401 Chr.15: 43875882-43875882 on Build GRCh38
rs1022008882 Chr.15: 43875822-43875822 on Build GRCh38
rs893315650 Chr.15: 43876108-43876108 on Build GRCh38
rs1006769949 Chr.15: 43876209-43876209 on Build GRCh38
rs201111977 Chr.15: 43875828-43875828 on Build GRCh38
rs59125307 Chr.15: 43875827-43875828 on Build GRCh38
rs756553148 Chr.15: 43875804-43875805 on Build GRCh38
rs976736368 Chr.15: 43875842-43875842 on Build GRCh38
rs763781986 Chr.15: 43876296-43876296 on Build GRCh38
rs962881993 Chr.15: 43876222-43876222 on Build GRCh38
rs878875445 Chr.15: 43876110-43876110 on Build GRCh38
rs575926633 Chr.15: 43876196-43876196 on Build GRCh38
rs374698927 Chr.15: 43875807-43875808 on Build GRCh38
rs746474755 Chr.15: 43876210-43876210 on Build GRCh38
rs535006682 Chr.15: 43875822-43875823 on Build GRCh38
rs999677309 Chr.15: 43876168-43876168 on Build GRCh38
rs902290796 Chr.15: 43876164-43876164 on Build GRCh38
rs773541354 Chr.15: 43875794-43875794 on Build GRCh38
rs143606979 Chr.15: 43875928-43875928 on Build GRCh38
rs375846572 Chr.15: 43876074-43876074 on Build GRCh38
rs906067197 Chr.15: 43875955-43875955 on Build GRCh38
rs554735603 Chr.15: 43876239-43876239 on Build GRCh38
rs184424372 Chr.15: 43875995-43875995 on Build GRCh38
rs866882530 Chr.15: 43875813-43875813 on Build GRCh38
rs536284238 Chr.15: 43875970-43875970 on Build GRCh38
rs947549219 Chr.15: 43876131-43876131 on Build GRCh38
rs976999838 Chr.15: 43876104-43876104 on Build GRCh38
rs930033169 Chr.15: 43875958-43875958 on Build GRCh38
rs955326737 Chr.15: 43875902-43875902 on Build GRCh38
rs201864727 Chr.15: 43875812-43875813 on Build GRCh38
rs972735298 Chr.15: 43875798-43875798 on Build GRCh38
rs952790992 Chr.15: 43875805-43875816 on Build GRCh38
rs139491002 Chr.15: 43876240-43876240 on Build GRCh38
rs984254875 Chr.15: 43876046-43876046 on Build GRCh38
rs879222258 Chr.15: 43876005-43876005 on Build GRCh38
rs878872791 Chr.15: 43876040-43876040 on Build GRCh38
rs748592713 Chr.15: 43876292-43876292 on Build GRCh38
rs999226277 Chr.15: 43875878-43875878 on Build GRCh38
rs922412266 Chr.15: 43876108-43876109 on Build GRCh38
rs1042262035 Chr.15: 43876137-43876137 on Build GRCh38
rs118074011 Chr.15: 43875946-43875946 on Build GRCh38
rs988003190 Chr.15: 43876201-43876201 on Build GRCh38
rs12442297 Chr.15: 43876272-43876272 on Build GRCh38
rs1035826233 Chr.15: 43875979-43875979 on Build GRCh38
rs560093841 Chr.15: 43876275-43876275 on Build GRCh38
rs937562885 Chr.15: 43875805-43875805 on Build GRCh38
rs902188388 Chr.15: 43875877-43875877 on Build GRCh38
rs922514422 Chr.15: 43875868-43875870 on Build GRCh38
rs754337278 Chr.15: 43876096-43876096 on Build GRCh38
rs878976721 Chr.15: 43875938-43875938 on Build GRCh38
rs879169999 Chr.15: 43875851-43875851 on Build GRCh38
rs779581541 Chr.15: 43876130-43876130 on Build GRCh38
rs146312339 Chr.15: 43876204-43876204 on Build GRCh38
rs796522665 Chr.15: 43875813-43875815 on Build GRCh38
rs946380105 Chr.15: 43876225-43876226 on Build GRCh38
rs962485704 Chr.15: 43875931-43875931 on Build GRCh38
rs1025674254 Chr.15: 43876036-43876036 on Build GRCh38
rs397744718 Chr.15: 43875805-43875805 on Build GRCh38
rs752632495 Chr.15: 43876288-43876288 on Build GRCh38
rs893304444 Chr.15: 43875814-43875814 on Build GRCh38
rs34063043 Chr.15: 43875827-43875827 on Build GRCh38
rs878954016 Chr.15: 43876003-43876003 on Build GRCh38

View Full Product Details