Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 50945732-50946312 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
AP4E1
Entrez Gene ID
23431
Gene Name
adaptor related protein complex 4 epsilon 1 subunit
Gene Aliases
CPSQ4, SPG51, STUT1
UniGene
Hs.413366
Gene Chromosome Location
Chr.15: 50908569-51005900 on Build GRCh38
Transcripts
NM_001252127.1 NM_007347.4 XM_005254264.3 XM_006720447.3 XM_017022042.1 NR_027642.1

Gene Symbol
DCAF13P3
Entrez Gene ID
100132724
Gene Name
DDB1 and CUL4 associated factor 13 pseudogene 3
Gene Aliases
-
UniGene
Hs.730493
Gene Chromosome Location
Chr.15: 50944129-50946573 on Build GRCh38
Transcripts
NM_001252127.1 NM_007347.4 XM_005254264.3 XM_006720447.3 XM_017022042.1 NR_027642.1

SNP Detail

SNP ID Chromosome Location
rs902811912 Chr.15: 50945763-50945763 on Build GRCh38
rs537688835 Chr.15: 50945840-50945840 on Build GRCh38
rs1026217809 Chr.15: 50946233-50946233 on Build GRCh38
rs146366966 Chr.15: 50946260-50946260 on Build GRCh38
rs566449137 Chr.15: 50945808-50945808 on Build GRCh38
rs540573650 Chr.15: 50946140-50946140 on Build GRCh38
rs1026291707 Chr.15: 50945902-50945902 on Build GRCh38
rs945614106 Chr.15: 50946081-50946081 on Build GRCh38
rs573162522 Chr.15: 50946028-50946028 on Build GRCh38
rs3784304 Chr.15: 50945863-50945863 on Build GRCh38
rs370714293 Chr.15: 50945964-50945964 on Build GRCh38
rs768447620 Chr.15: 50945886-50945886 on Build GRCh38
rs74015237 Chr.15: 50946154-50946154 on Build GRCh38
rs1051975778 Chr.15: 50946159-50946159 on Build GRCh38
rs879119644 Chr.15: 50945901-50945901 on Build GRCh38
rs190529794 Chr.15: 50946285-50946285 on Build GRCh38
rs533861790 Chr.15: 50945815-50945815 on Build GRCh38
rs879172436 Chr.15: 50945907-50945907 on Build GRCh38
rs993388625 Chr.15: 50946197-50946197 on Build GRCh38
rs113244470 Chr.15: 50945959-50945959 on Build GRCh38
rs879055345 Chr.15: 50945883-50945883 on Build GRCh38
rs997694808 Chr.15: 50945810-50945810 on Build GRCh38
rs879171826 Chr.15: 50945784-50945784 on Build GRCh38
rs888861766 Chr.15: 50946166-50946169 on Build GRCh38
rs868850748 Chr.15: 50945904-50945904 on Build GRCh38
rs924286959 Chr.15: 50946129-50946129 on Build GRCh38
rs898978663 Chr.15: 50946184-50946184 on Build GRCh38
rs756048451 Chr.15: 50946201-50946201 on Build GRCh38
rs529603620 Chr.15: 50946168-50946168 on Build GRCh38
rs781187761 Chr.15: 50945923-50945923 on Build GRCh38
rs771121888 Chr.15: 50946105-50946105 on Build GRCh38
rs879116330 Chr.15: 50945771-50945771 on Build GRCh38
rs776616338 Chr.15: 50946170-50946173 on Build GRCh38
rs888978237 Chr.15: 50945836-50945836 on Build GRCh38
rs1022491867 Chr.15: 50946276-50946276 on Build GRCh38
rs17599981 Chr.15: 50945819-50945819 on Build GRCh38
rs1955 Chr.15: 50945898-50945898 on Build GRCh38
rs183206961 Chr.15: 50945905-50945905 on Build GRCh38
rs139726527 Chr.15: 50946266-50946266 on Build GRCh38
rs35783233 Chr.15: 50946269-50946269 on Build GRCh38
rs7162209 Chr.15: 50945978-50945978 on Build GRCh38
rs766323156 Chr.15: 50945861-50945861 on Build GRCh38
rs1033750655 Chr.15: 50945925-50945925 on Build GRCh38
rs920375198 Chr.15: 50945842-50945869 on Build GRCh38
rs989526012 Chr.15: 50946273-50946273 on Build GRCh38
rs188493858 Chr.15: 50946264-50946264 on Build GRCh38
rs879048808 Chr.15: 50945864-50945864 on Build GRCh38
rs148802645 Chr.15: 50945833-50945833 on Build GRCh38

View Full Product Details