Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99791423-99791972 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1P46
Entrez Gene ID
196968
Gene Name
dynamin 1 pseudogene 46
Gene Aliases
C15orf51, DNM1DN14.2, DNM1DN14@
UniGene
Hs.567763
Gene Chromosome Location
Chr.15: 99790156-99806927 on Build GRCh38
Transcripts
NR_003260.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
6141673 c.90G>T p.W30C Chr.15: 99791632-99791632 on Build GRCh38
3499668 c.45G>A p.E15E Chr.15: 99791587-99791587 on Build GRCh38
3499669 c.61C>T p.P21S Chr.15: 99791603-99791603 on Build GRCh38
3499670 c.76G>A p.D26N Chr.15: 99791618-99791618 on Build GRCh38
6265148 c.98C>T p.T33M Chr.15: 99791640-99791640 on Build GRCh38
View More

SNP Detail

SNP ID Chromosome Location
rs60883741 Chr.15: 99791748-99791748 on Build GRCh38
rs374487396 Chr.15: 99791931-99791931 on Build GRCh38
rs770357536 Chr.15: 99791652-99791652 on Build GRCh38
rs992889802 Chr.15: 99791701-99791701 on Build GRCh38
rs758003609 Chr.15: 99791540-99791540 on Build GRCh38
rs755420932 Chr.15: 99791624-99791624 on Build GRCh38
rs377107927 Chr.15: 99791803-99791803 on Build GRCh38
rs2002316 Chr.15: 99791459-99791459 on Build GRCh38
rs755513222 Chr.15: 99791811-99791811 on Build GRCh38
rs759310754 Chr.15: 99791520-99791520 on Build GRCh38
rs1809156 Chr.15: 99791756-99791756 on Build GRCh38
rs745475224 Chr.15: 99791755-99791755 on Build GRCh38
rs752018342 Chr.15: 99791740-99791740 on Build GRCh38
rs200930880 Chr.15: 99791483-99791483 on Build GRCh38
rs57196130 Chr.15: 99791832-99791832 on Build GRCh38
rs775801014 Chr.15: 99791852-99791852 on Build GRCh38
rs150563178 Chr.15: 99791762-99791762 on Build GRCh38
rs769717363 Chr.15: 99791849-99791849 on Build GRCh38
rs776919924 Chr.15: 99791773-99791773 on Build GRCh38
rs113026694 Chr.15: 99791819-99791819 on Build GRCh38
rs574525248 Chr.15: 99791582-99791582 on Build GRCh38
rs751644802 Chr.15: 99791462-99791462 on Build GRCh38
rs746062024 Chr.15: 99791481-99791481 on Build GRCh38
rs376191987 Chr.15: 99791694-99791694 on Build GRCh38
rs779788782 Chr.15: 99791814-99791814 on Build GRCh38
rs778697283 Chr.15: 99791884-99791884 on Build GRCh38
rs370226845 Chr.15: 99791749-99791749 on Build GRCh38
rs754637697 Chr.15: 99791820-99791820 on Build GRCh38
rs773223552 Chr.15: 99791847-99791847 on Build GRCh38
rs775311239 Chr.15: 99791599-99791599 on Build GRCh38
rs764982118 Chr.15: 99791946-99791946 on Build GRCh38
rs757312633 Chr.15: 99791538-99791538 on Build GRCh38
rs753351886 Chr.15: 99791944-99791944 on Build GRCh38
rs371418106 Chr.15: 99791915-99791915 on Build GRCh38
rs760371857 Chr.15: 99791517-99791517 on Build GRCh38
rs757299199 Chr.15: 99791463-99791463 on Build GRCh38
rs768018124 Chr.15: 99791867-99791867 on Build GRCh38
rs140463284 Chr.15: 99791769-99791769 on Build GRCh38
rs747617021 Chr.15: 99791507-99791507 on Build GRCh38
rs781095676 Chr.15: 99791699-99791699 on Build GRCh38
rs779323944 Chr.15: 99791572-99791572 on Build GRCh38
rs747954907 Chr.15: 99791824-99791824 on Build GRCh38
rs184216297 Chr.15: 99791641-99791641 on Build GRCh38
rs970289090 Chr.15: 99791860-99791860 on Build GRCh38
rs777882463 Chr.15: 99791902-99791902 on Build GRCh38
rs778895418 Chr.15: 99791823-99791823 on Build GRCh38
rs779389551 Chr.15: 99791572-99791574 on Build GRCh38
rs779386655 Chr.15: 99791633-99791633 on Build GRCh38
rs746071 Chr.15: 99791547-99791547 on Build GRCh38
rs762994735 Chr.15: 99791605-99791605 on Build GRCh38
rs746514909 Chr.15: 99791591-99791591 on Build GRCh38
rs976511675 Chr.15: 99791932-99791932 on Build GRCh38
rs148063894 Chr.15: 99791640-99791640 on Build GRCh38
rs36116585 Chr.15: 99791773-99791774 on Build GRCh38
rs750001080 Chr.15: 99791527-99791527 on Build GRCh38
rs554402538 Chr.15: 99791853-99791853 on Build GRCh38
rs993006017 Chr.15: 99791546-99791546 on Build GRCh38
rs548874848 Chr.15: 99791678-99791678 on Build GRCh38
rs386787233 Chr.15: 99791459-99791460 on Build GRCh38
rs368202979 Chr.15: 99791511-99791511 on Build GRCh38
rs745962088 Chr.15: 99791848-99791848 on Build GRCh38
rs529670264 Chr.15: 99791938-99791938 on Build GRCh38
rs559511977 Chr.15: 99791919-99791919 on Build GRCh38
rs773605253 Chr.15: 99791933-99791933 on Build GRCh38
rs750447254 Chr.15: 99791548-99791548 on Build GRCh38
rs922347381 Chr.15: 99791936-99791936 on Build GRCh38
rs774366663 Chr.15: 99791663-99791663 on Build GRCh38
rs776475187 Chr.15: 99791518-99791518 on Build GRCh38
rs746675846 Chr.15: 99791579-99791581 on Build GRCh38
rs755328928 Chr.15: 99791565-99791565 on Build GRCh38
rs2958994 Chr.15: 99791614-99791614 on Build GRCh38
rs760422237 Chr.15: 99791454-99791454 on Build GRCh38
rs970615673 Chr.15: 99791746-99791746 on Build GRCh38
rs774498098 Chr.15: 99791922-99791922 on Build GRCh38
rs765170210 Chr.15: 99791523-99791523 on Build GRCh38
rs532231725 Chr.15: 99791623-99791623 on Build GRCh38
rs769591146 Chr.15: 99791596-99791596 on Build GRCh38
rs754269404 Chr.15: 99791561-99791561 on Build GRCh38
rs535100307 Chr.15: 99791775-99791775 on Build GRCh38
rs202013273 Chr.15: 99791797-99791797 on Build GRCh38
rs897142090 Chr.15: 99791837-99791837 on Build GRCh38
rs771342870 Chr.15: 99791464-99791464 on Build GRCh38
rs762150447 Chr.15: 99791865-99791865 on Build GRCh38
rs778125889 Chr.15: 99791644-99791644 on Build GRCh38
rs746533223 Chr.15: 99791645-99791645 on Build GRCh38
rs780850878 Chr.15: 99791654-99791654 on Build GRCh38
rs751885913 Chr.15: 99791683-99791684 on Build GRCh38
rs553063958 Chr.15: 99791776-99791776 on Build GRCh38
rs763905135 Chr.15: 99791613-99791613 on Build GRCh38
rs752556562 Chr.15: 99791893-99791893 on Build GRCh38
rs765377132 Chr.15: 99791686-99791686 on Build GRCh38
rs529730144 Chr.15: 99791656-99791656 on Build GRCh38
rs773159791 Chr.15: 99791875-99791875 on Build GRCh38
rs192182901 Chr.15: 99791558-99791558 on Build GRCh38
rs142799822 Chr.15: 99791907-99791907 on Build GRCh38
rs777385713 Chr.15: 99791589-99791589 on Build GRCh38
rs372284341 Chr.15: 99791594-99791594 on Build GRCh38
rs763402384 Chr.15: 99791788-99791788 on Build GRCh38
rs764663439 Chr.15: 99791724-99791724 on Build GRCh38
rs779516738 Chr.15: 99791751-99791751 on Build GRCh38
rs749667695 Chr.15: 99791492-99791492 on Build GRCh38
rs2004046 Chr.15: 99791619-99791619 on Build GRCh38
rs760255485 Chr.15: 99791872-99791872 on Build GRCh38
rs370279132 Chr.15: 99791731-99791731 on Build GRCh38
rs576275616 Chr.15: 99791871-99791871 on Build GRCh38
rs752608674 Chr.15: 99791527-99791527 on Build GRCh38
rs577278863 Chr.15: 99791494-99791494 on Build GRCh38
rs770695628 Chr.15: 99791642-99791642 on Build GRCh38
rs769776958 Chr.15: 99791836-99791839 on Build GRCh38
rs147495914 Chr.15: 99791914-99791914 on Build GRCh38
rs774864535 Chr.15: 99791506-99791506 on Build GRCh38
rs2077254 Chr.15: 99791460-99791460 on Build GRCh38
rs763228327 Chr.15: 99791945-99791945 on Build GRCh38
rs764390401 Chr.15: 99791791-99791791 on Build GRCh38
rs768652928 Chr.15: 99791661-99791661 on Build GRCh38
rs753360822 Chr.15: 99791881-99791881 on Build GRCh38
rs535459515 Chr.15: 99791845-99791845 on Build GRCh38
rs748643961 Chr.15: 99791574-99791574 on Build GRCh38
rs762197633 Chr.15: 99791925-99791925 on Build GRCh38
rs756781049 Chr.15: 99791743-99791743 on Build GRCh38
rs754747780 Chr.15: 99791883-99791883 on Build GRCh38
rs750005621 Chr.15: 99791805-99791805 on Build GRCh38
rs774761924 Chr.15: 99791864-99791864 on Build GRCh38
rs758629791 Chr.15: 99791719-99791719 on Build GRCh38
rs765857139 Chr.15: 99791876-99791876 on Build GRCh38
rs760869248 Chr.15: 99791671-99791671 on Build GRCh38
rs146847301 Chr.15: 99791659-99791659 on Build GRCh38
rs780612351 Chr.15: 99791490-99791490 on Build GRCh38
rs766116025 Chr.15: 99791458-99791458 on Build GRCh38
rs762649360 Chr.15: 99791659-99791659 on Build GRCh38
rs747929597 Chr.15: 99791742-99791743 on Build GRCh38
rs781275442 Chr.15: 99791471-99791471 on Build GRCh38
rs543612334 Chr.15: 99791618-99791618 on Build GRCh38
rs748843547 Chr.15: 99791818-99791818 on Build GRCh38
rs2004045 Chr.15: 99791595-99791595 on Build GRCh38
rs772765153 Chr.15: 99791515-99791515 on Build GRCh38
rs543397099 Chr.15: 99791895-99791895 on Build GRCh38
rs139955858 Chr.15: 99791937-99791937 on Build GRCh38

View Full Product Details