Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.15: 99799711-99800300 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DNM1P46
Entrez Gene ID
196968
Gene Name
dynamin 1 pseudogene 46
Gene Aliases
C15orf51, DNM1DN14.2, DNM1DN14@
UniGene
Hs.567763
Gene Chromosome Location
Chr.15: 99790156-99806927 on Build GRCh38
Transcripts
NR_003260.1

SNP Detail

SNP ID Chromosome Location
rs761322545 Chr.15: 99799744-99799744 on Build GRCh38
rs753682321 Chr.15: 99799954-99799960 on Build GRCh38
rs765040844 Chr.15: 99799969-99799970 on Build GRCh38
rs764140771 Chr.15: 99800056-99800103 on Build GRCh38
rs374404821 Chr.15: 99800047-99800047 on Build GRCh38
rs771884012 Chr.15: 99800197-99800197 on Build GRCh38
rs779175896 Chr.15: 99800081-99800081 on Build GRCh38
rs757260139 Chr.15: 99799878-99799879 on Build GRCh38
rs879275021 Chr.15: 99800034-99800036 on Build GRCh38
rs572152598 Chr.15: 99800209-99800209 on Build GRCh38
rs759617689 Chr.15: 99800144-99800147 on Build GRCh38
rs200285387 Chr.15: 99799765-99799765 on Build GRCh38
rs201033523 Chr.15: 99800038-99800038 on Build GRCh38
rs764616241 Chr.15: 99799938-99799938 on Build GRCh38
rs759303503 Chr.15: 99799880-99799880 on Build GRCh38
rs776000909 Chr.15: 99800023-99800064 on Build GRCh38
rs766787873 Chr.15: 99799804-99799805 on Build GRCh38
rs770125875 Chr.15: 99799888-99799888 on Build GRCh38
rs750230015 Chr.15: 99800129-99800129 on Build GRCh38
rs760495321 Chr.15: 99800159-99800159 on Build GRCh38
rs2924766 Chr.15: 99799769-99799769 on Build GRCh38
rs776019883 Chr.15: 99800148-99800148 on Build GRCh38
rs577492113 Chr.15: 99800181-99800181 on Build GRCh38
rs201397519 Chr.15: 99800200-99800200 on Build GRCh38
rs202212587 Chr.15: 99800035-99800035 on Build GRCh38
rs769319990 Chr.15: 99799882-99799882 on Build GRCh38
rs200691929 Chr.15: 99799970-99799970 on Build GRCh38
rs747473616 Chr.15: 99800035-99800051 on Build GRCh38
rs767030576 Chr.15: 99799935-99799968 on Build GRCh38
rs532901507 Chr.15: 99800113-99800113 on Build GRCh38
rs775021538 Chr.15: 99799987-99799989 on Build GRCh38
rs771124218 Chr.15: 99800141-99800141 on Build GRCh38
rs200762313 Chr.15: 99800035-99800035 on Build GRCh38
rs77574325 Chr.15: 99800279-99800279 on Build GRCh38
rs761974786 Chr.15: 99800196-99800203 on Build GRCh38
rs372299313 Chr.15: 99799860-99799860 on Build GRCh38
rs757854653 Chr.15: 99799939-99799939 on Build GRCh38
rs576374785 Chr.15: 99800008-99800008 on Build GRCh38
rs746757593 Chr.15: 99800055-99800055 on Build GRCh38
rs757491120 Chr.15: 99800018-99800023 on Build GRCh38
rs777605084 Chr.15: 99799876-99799878 on Build GRCh38
rs760089784 Chr.15: 99800115-99800115 on Build GRCh38
rs768488525 Chr.15: 99799916-99799916 on Build GRCh38
rs200568726 Chr.15: 99800214-99800214 on Build GRCh38
rs765116474 Chr.15: 99799811-99799811 on Build GRCh38
rs763321753 Chr.15: 99799983-99799983 on Build GRCh38
rs538591648 Chr.15: 99800204-99800204 on Build GRCh38
rs560908236 Chr.15: 99800081-99800081 on Build GRCh38
rs376517074 Chr.15: 99800210-99800210 on Build GRCh38
rs746213991 Chr.15: 99800019-99800026 on Build GRCh38
rs879100596 Chr.15: 99800117-99800117 on Build GRCh38
rs371070889 Chr.15: 99799751-99799751 on Build GRCh38
rs776371934 Chr.15: 99799810-99799810 on Build GRCh38
rs200156126 Chr.15: 99799867-99799867 on Build GRCh38
rs752434231 Chr.15: 99799816-99799816 on Build GRCh38
rs149253821 Chr.15: 99799994-99799994 on Build GRCh38
rs750563405 Chr.15: 99800099-99800100 on Build GRCh38
rs761058826 Chr.15: 99800067-99800067 on Build GRCh38
rs759494210 Chr.15: 99799929-99799929 on Build GRCh38
rs770706722 Chr.15: 99800244-99800245 on Build GRCh38
rs780254957 Chr.15: 99800186-99800186 on Build GRCh38
rs765240769 Chr.15: 99800246-99800246 on Build GRCh38
rs368425453 Chr.15: 99799918-99799920 on Build GRCh38
rs71114059 Chr.15: 99799754-99799755 on Build GRCh38
rs543954261 Chr.15: 99800174-99800174 on Build GRCh38
rs75484537 Chr.15: 99800208-99800208 on Build GRCh38
rs757831913 Chr.15: 99799901-99799901 on Build GRCh38
rs4965538 Chr.15: 99800103-99800103 on Build GRCh38
rs748392248 Chr.15: 99799887-99799888 on Build GRCh38
rs200874874 Chr.15: 99800026-99800026 on Build GRCh38
rs753916161 Chr.15: 99800148-99800149 on Build GRCh38
rs772859697 Chr.15: 99799806-99799806 on Build GRCh38
rs878909727 Chr.15: 99799833-99799833 on Build GRCh38
rs867538109 Chr.15: 99800002-99800002 on Build GRCh38
rs763705377 Chr.15: 99799951-99799951 on Build GRCh38
rs954745200 Chr.15: 99799887-99799887 on Build GRCh38
rs763570063 Chr.15: 99799803-99799819 on Build GRCh38
rs760388200 Chr.15: 99799948-99799948 on Build GRCh38
rs781476850 Chr.15: 99799985-99799986 on Build GRCh38
rs781107302 Chr.15: 99799761-99799761 on Build GRCh38
rs771811242 Chr.15: 99800003-99800003 on Build GRCh38
rs757660317 Chr.15: 99799990-99799990 on Build GRCh38
rs202053487 Chr.15: 99800012-99800012 on Build GRCh38
rs151282233 Chr.15: 99800158-99800158 on Build GRCh38
rs756117533 Chr.15: 99800098-99800098 on Build GRCh38
rs577378383 Chr.15: 99800060-99800060 on Build GRCh38
rs745842130 Chr.15: 99800095-99800095 on Build GRCh38
rs780815667 Chr.15: 99799903-99799903 on Build GRCh38
rs752869211 Chr.15: 99799995-99800000 on Build GRCh38
rs375097857 Chr.15: 99799870-99799870 on Build GRCh38
rs772213794 Chr.15: 99800109-99800109 on Build GRCh38
rs113166086 Chr.15: 99799973-99799973 on Build GRCh38
rs879935282 Chr.15: 99800028-99800039 on Build GRCh38
rs76795743 Chr.15: 99800091-99800091 on Build GRCh38
rs766383253 Chr.15: 99799994-99800016 on Build GRCh38
rs749687390 Chr.15: 99799800-99799800 on Build GRCh38
rs774969370 Chr.15: 99799913-99799913 on Build GRCh38
rs554444882 Chr.15: 99799943-99799943 on Build GRCh38
rs778372400 Chr.15: 99800048-99800048 on Build GRCh38
rs770781260 Chr.15: 99800056-99800056 on Build GRCh38
rs370763277 Chr.15: 99800118-99800118 on Build GRCh38
rs200037202 Chr.15: 99799974-99799974 on Build GRCh38
rs750509068 Chr.15: 99799830-99799830 on Build GRCh38
rs536373915 Chr.15: 99799956-99799956 on Build GRCh38
rs766344096 Chr.15: 99799748-99799748 on Build GRCh38
rs572544601 Chr.15: 99800137-99800140 on Build GRCh38
rs781117087 Chr.15: 99800092-99800092 on Build GRCh38
rs750216146 Chr.15: 99799989-99799991 on Build GRCh38
rs375545524 Chr.15: 99799743-99799743 on Build GRCh38
rs774331949 Chr.15: 99800257-99800258 on Build GRCh38
rs1017763313 Chr.15: 99799920-99799920 on Build GRCh38
rs374942363 Chr.15: 99799907-99799907 on Build GRCh38
rs780329756 Chr.15: 99800111-99800116 on Build GRCh38
rs367968117 Chr.15: 99800215-99800215 on Build GRCh38
rs2958978 Chr.15: 99800188-99800188 on Build GRCh38
rs8027467 Chr.15: 99799747-99799747 on Build GRCh38
rs765514936 Chr.15: 99799930-99799930 on Build GRCh38
rs779213678 Chr.15: 99800019-99800025 on Build GRCh38
rs775728667 Chr.15: 99800015-99800015 on Build GRCh38
rs769495112 Chr.15: 99799909-99799909 on Build GRCh38
rs550514942 Chr.15: 99799926-99799926 on Build GRCh38
rs375825638 Chr.15: 99800080-99800080 on Build GRCh38
rs375152504 Chr.15: 99799752-99799752 on Build GRCh38
rs746250932 Chr.15: 99799732-99799732 on Build GRCh38
rs765191583 Chr.15: 99800212-99800213 on Build GRCh38
rs780072225 Chr.15: 99800132-99800132 on Build GRCh38
rs539242114 Chr.15: 99799942-99799942 on Build GRCh38
rs777937616 Chr.15: 99800012-99800012 on Build GRCh38
rs1043478138 Chr.15: 99799754-99799754 on Build GRCh38
rs200145736 Chr.15: 99800036-99800036 on Build GRCh38
rs531995335 Chr.15: 99799917-99799917 on Build GRCh38
rs868308613 Chr.15: 99800137-99800137 on Build GRCh38
rs566160472 Chr.15: 99799951-99799951 on Build GRCh38
rs755239712 Chr.15: 99799890-99799890 on Build GRCh38
rs12905745 Chr.15: 99799791-99799791 on Build GRCh38
rs963990956 Chr.15: 99799960-99799960 on Build GRCh38
rs779193744 Chr.15: 99800104-99800104 on Build GRCh38
rs774864971 Chr.15: 99800119-99800119 on Build GRCh38
rs368158096 Chr.15: 99799757-99799757 on Build GRCh38
rs753626079 Chr.15: 99799877-99799878 on Build GRCh38
rs878869443 Chr.15: 99799958-99799958 on Build GRCh38
rs760024569 Chr.15: 99799932-99799933 on Build GRCh38
rs757309702 Chr.15: 99799759-99799759 on Build GRCh38
rs775892886 Chr.15: 99799768-99799768 on Build GRCh38
rs745770087 Chr.15: 99799881-99799882 on Build GRCh38
rs770318928 Chr.15: 99800147-99800147 on Build GRCh38
rs780525538 Chr.15: 99800217-99800217 on Build GRCh38
rs752187776 Chr.15: 99800084-99800084 on Build GRCh38
rs772617183 Chr.15: 99800020-99800024 on Build GRCh38
rs777683213 Chr.15: 99800138-99800138 on Build GRCh38
rs751345777 Chr.15: 99800088-99800088 on Build GRCh38
rs12904401 Chr.15: 99799755-99799755 on Build GRCh38
rs745937058 Chr.15: 99799795-99799795 on Build GRCh38
rs767486827 Chr.15: 99800126-99800126 on Build GRCh38
rs28696056 Chr.15: 99800114-99800114 on Build GRCh38
rs775633500 Chr.15: 99799982-99799982 on Build GRCh38
rs750797162 Chr.15: 99800043-99800043 on Build GRCh38
rs773775999 Chr.15: 99799897-99799912 on Build GRCh38
rs746161419 Chr.15: 99800145-99800145 on Build GRCh38
rs759208290 Chr.15: 99800235-99800235 on Build GRCh38
rs773926423 Chr.15: 99800189-99800189 on Build GRCh38
rs199920642 Chr.15: 99799996-99799996 on Build GRCh38
rs774968591 Chr.15: 99799947-99799948 on Build GRCh38
rs777589636 Chr.15: 99800009-99800009 on Build GRCh38
rs536716167 Chr.15: 99800156-99800156 on Build GRCh38
rs367692336 Chr.15: 99800107-99800107 on Build GRCh38
rs775804993 Chr.15: 99799735-99799735 on Build GRCh38
rs762085962 Chr.15: 99799734-99799736 on Build GRCh38
rs769211112 Chr.15: 99800219-99800219 on Build GRCh38
rs878995148 Chr.15: 99799947-99799947 on Build GRCh38
rs776869172 Chr.15: 99799731-99799731 on Build GRCh38
rs780278848 Chr.15: 99799832-99799832 on Build GRCh38
rs756147840 Chr.15: 99799831-99799831 on Build GRCh38
rs373620902 Chr.15: 99800157-99800157 on Build GRCh38
rs202001736 Chr.15: 99800039-99800039 on Build GRCh38
rs74183881 Chr.15: 99800225-99800225 on Build GRCh38
rs771208428 Chr.15: 99799986-99799987 on Build GRCh38
rs762848663 Chr.15: 99800270-99800270 on Build GRCh38
rs753852423 Chr.15: 99800161-99800161 on Build GRCh38
rs4965539 Chr.15: 99800111-99800111 on Build GRCh38
rs767017262 Chr.15: 99799825-99799867 on Build GRCh38
rs750330801 Chr.15: 99799974-99799974 on Build GRCh38
rs757023677 Chr.15: 99799967-99799968 on Build GRCh38
rs760460413 Chr.15: 99799782-99799782 on Build GRCh38
rs112848112 Chr.15: 99799734-99799734 on Build GRCh38
rs766063801 Chr.15: 99800160-99800160 on Build GRCh38
rs759493231 Chr.15: 99800052-99800053 on Build GRCh38
rs748202888 Chr.15: 99800106-99800106 on Build GRCh38
rs12910758 Chr.15: 99799954-99799954 on Build GRCh38
rs149988221 Chr.15: 99800178-99800178 on Build GRCh38
rs770612488 Chr.15: 99799879-99799879 on Build GRCh38
rs756612955 Chr.15: 99799993-99799993 on Build GRCh38
rs775457512 Chr.15: 99800264-99800264 on Build GRCh38
rs544659325 Chr.15: 99799826-99799826 on Build GRCh38
rs200645530 Chr.15: 99799892-99799892 on Build GRCh38
rs779390590 Chr.15: 99799857-99799857 on Build GRCh38
rs531208583 Chr.15: 99800086-99800086 on Build GRCh38
rs756297319 Chr.15: 99800012-99800013 on Build GRCh38
rs201682562 Chr.15: 99799971-99799971 on Build GRCh38
rs747869414 Chr.15: 99799957-99799957 on Build GRCh38
rs559964164 Chr.15: 99800074-99800074 on Build GRCh38
rs377747541 Chr.15: 99800064-99800064 on Build GRCh38
rs767654052 Chr.15: 99800277-99800277 on Build GRCh38
rs761342014 Chr.15: 99800196-99800196 on Build GRCh38
rs767383923 Chr.15: 99799924-99799924 on Build GRCh38
rs776146872 Chr.15: 99800036-99800037 on Build GRCh38
rs768294228 Chr.15: 99799955-99799955 on Build GRCh38
rs755126070 Chr.15: 99799847-99799847 on Build GRCh38
rs768647404 Chr.15: 99799883-99799885 on Build GRCh38
rs201459191 Chr.15: 99800061-99800061 on Build GRCh38
rs201407168 Chr.15: 99799792-99799792 on Build GRCh38
rs748197474 Chr.15: 99800136-99800136 on Build GRCh38
rs764653067 Chr.15: 99799738-99799738 on Build GRCh38
rs762592010 Chr.15: 99799884-99799884 on Build GRCh38
rs769376891 Chr.15: 99800142-99800143 on Build GRCh38
rs374490983 Chr.15: 99800177-99800177 on Build GRCh38
rs560837787 Chr.15: 99800261-99800261 on Build GRCh38
rs766069488 Chr.15: 99799783-99799783 on Build GRCh38
rs749488198 Chr.15: 99800018-99800059 on Build GRCh38
rs200405906 Chr.15: 99799797-99799797 on Build GRCh38
rs548963102 Chr.15: 99799897-99799897 on Build GRCh38
rs562308080 Chr.15: 99799805-99799805 on Build GRCh38
rs749009399 Chr.15: 99800241-99800242 on Build GRCh38
rs753561195 Chr.15: 99800206-99800206 on Build GRCh38
rs773046048 Chr.15: 99800199-99800199 on Build GRCh38
rs12905752 Chr.15: 99799801-99799801 on Build GRCh38
rs755707937 Chr.15: 99799859-99799871 on Build GRCh38
rs760462369 Chr.15: 99800228-99800228 on Build GRCh38
rs758061676 Chr.15: 99800054-99800054 on Build GRCh38
rs770285567 Chr.15: 99799755-99799769 on Build GRCh38
rs773651483 Chr.15: 99799764-99799770 on Build GRCh38
rs370958921 Chr.15: 99800073-99800073 on Build GRCh38
rs758296849 Chr.15: 99799984-99799985 on Build GRCh38
rs556044398 Chr.15: 99800058-99800058 on Build GRCh38
rs770427765 Chr.15: 99800233-99800233 on Build GRCh38
rs774669465 Chr.15: 99800023-99800023 on Build GRCh38
rs765269054 Chr.15: 99799977-99799977 on Build GRCh38
rs775373859 Chr.15: 99799883-99799883 on Build GRCh38
rs780631073 Chr.15: 99800130-99800158 on Build GRCh38
rs772936522 Chr.15: 99800143-99800144 on Build GRCh38
rs2924765 Chr.15: 99800170-99800170 on Build GRCh38
rs769136855 Chr.15: 99800117-99800148 on Build GRCh38
rs750047936 Chr.15: 99799746-99799746 on Build GRCh38
rs202160044 Chr.15: 99800077-99800077 on Build GRCh38
rs566176186 Chr.15: 99800153-99800153 on Build GRCh38
rs763521795 Chr.15: 99799737-99799737 on Build GRCh38
rs764356623 Chr.15: 99799989-99799989 on Build GRCh38
rs369789350 Chr.15: 99799999-99799999 on Build GRCh38
rs2958979 Chr.15: 99799961-99799961 on Build GRCh38
rs774104034 Chr.15: 99799919-99799919 on Build GRCh38
rs554933545 Chr.15: 99799998-99799998 on Build GRCh38
rs369493668 Chr.15: 99799893-99799893 on Build GRCh38
rs201721951 Chr.15: 99799749-99799749 on Build GRCh38
rs781352484 Chr.15: 99799883-99799884 on Build GRCh38
rs751550874 Chr.15: 99800046-99800059 on Build GRCh38
rs772326813 Chr.15: 99800063-99800063 on Build GRCh38
rs199701716 Chr.15: 99799794-99799794 on Build GRCh38
rs764050857 Chr.15: 99799885-99799885 on Build GRCh38
rs751277871 Chr.15: 99800124-99800124 on Build GRCh38
rs767812690 Chr.15: 99800037-99800037 on Build GRCh38
rs763544599 Chr.15: 99799987-99799988 on Build GRCh38
rs761429360 Chr.15: 99800037-99800043 on Build GRCh38
rs147955712 Chr.15: 99800234-99800234 on Build GRCh38
rs752384002 Chr.15: 99800053-99800053 on Build GRCh38
rs771746937 Chr.15: 99800222-99800222 on Build GRCh38
rs747526462 Chr.15: 99800115-99800116 on Build GRCh38
rs762816063 Chr.15: 99800040-99800041 on Build GRCh38
rs763193623 Chr.15: 99799915-99799916 on Build GRCh38
rs556408353 Chr.15: 99800016-99800016 on Build GRCh38
rs777092917 Chr.15: 99800240-99800241 on Build GRCh38
rs776990222 Chr.15: 99800144-99800144 on Build GRCh38
rs368528624 Chr.15: 99799859-99799859 on Build GRCh38
rs751116581 Chr.15: 99799805-99799806 on Build GRCh38
rs368349456 Chr.15: 99800046-99800046 on Build GRCh38
rs748232580 Chr.15: 99799986-99799993 on Build GRCh38
rs780452493 Chr.15: 99800213-99800214 on Build GRCh38
rs748641606 Chr.15: 99800220-99800220 on Build GRCh38
rs761806640 Chr.15: 99799923-99799923 on Build GRCh38
rs749131606 Chr.15: 99799915-99799915 on Build GRCh38
rs770003487 Chr.15: 99799766-99799766 on Build GRCh38
rs753687254 Chr.15: 99799787-99799787 on Build GRCh38
rs758474323 Chr.15: 99799991-99799992 on Build GRCh38
rs778693812 Chr.15: 99799882-99799883 on Build GRCh38
rs763349973 Chr.15: 99800152-99800152 on Build GRCh38
rs769480673 Chr.15: 99800039-99800040 on Build GRCh38
rs760194856 Chr.15: 99799808-99799808 on Build GRCh38
rs751449259 Chr.15: 99799886-99799886 on Build GRCh38
rs12905599 Chr.15: 99799733-99799733 on Build GRCh38
rs762487215 Chr.15: 99800122-99800122 on Build GRCh38
rs374367546 Chr.15: 99799799-99799799 on Build GRCh38
rs202154374 Chr.15: 99799779-99799779 on Build GRCh38
rs778380527 Chr.15: 99799863-99799863 on Build GRCh38
rs781488890 Chr.15: 99799940-99799940 on Build GRCh38
rs757462081 Chr.15: 99800077-99800078 on Build GRCh38
rs201248730 Chr.15: 99799856-99799856 on Build GRCh38
rs527360773 Chr.15: 99800075-99800075 on Build GRCh38
rs752908227 Chr.15: 99799935-99799935 on Build GRCh38
rs12910573 Chr.15: 99799825-99799825 on Build GRCh38
rs752353459 Chr.15: 99799827-99799829 on Build GRCh38
rs766223784 Chr.15: 99800044-99800045 on Build GRCh38
rs200975818 Chr.15: 99799981-99799981 on Build GRCh38
rs774756598 Chr.15: 99799918-99799927 on Build GRCh38
rs754141261 Chr.15: 99799844-99799844 on Build GRCh38
rs544213650 Chr.15: 99799802-99799802 on Build GRCh38
rs571820682 Chr.15: 99799931-99799931 on Build GRCh38
rs772077597 Chr.15: 99799962-99799962 on Build GRCh38
rs758793446 Chr.15: 99800102-99800104 on Build GRCh38
rs755531423 Chr.15: 99799948-99799948 on Build GRCh38
rs866613224 Chr.15: 99800071-99800071 on Build GRCh38
rs761718593 Chr.15: 99799888-99799888 on Build GRCh38
rs753951009 Chr.15: 99800073-99800074 on Build GRCh38
rs756135822 Chr.15: 99800130-99800130 on Build GRCh38
rs139432801 Chr.15: 99800059-99800059 on Build GRCh38

View Full Product Details