Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 2425459-2426008 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCA17P
Entrez Gene ID
650655
Gene Name
ATP binding cassette subfamily A member 17, pseudogene
Gene Aliases
ABCA17
UniGene
Hs.376281
Gene Chromosome Location
Chr.16: 2340922-2426699 on Build GRCh38
Transcripts
NR_003574.1 XM_017023822.1 XM_017023823.1

Gene Symbol
CCNF
Entrez Gene ID
899
Gene Name
cyclin F
Gene Aliases
FBX1, FBXO1
UniGene
Hs.1973
Gene Chromosome Location
Chr.16: 2425438-2458858 on Build GRCh38
Transcripts
NR_003574.1 XM_017023822.1 XM_017023823.1

SNP Detail

SNP ID Chromosome Location
rs765021966 Chr.16: 2425903-2425904 on Build GRCh38
rs527756471 Chr.16: 2425853-2425853 on Build GRCh38
rs546371907 Chr.16: 2425513-2425513 on Build GRCh38
rs574296738 Chr.16: 2425742-2425742 on Build GRCh38
rs747428907 Chr.16: 2425875-2425875 on Build GRCh38
rs1043587750 Chr.16: 2425747-2425747 on Build GRCh38
rs866442461 Chr.16: 2425594-2425594 on Build GRCh38
rs1053072739 Chr.16: 2425776-2425776 on Build GRCh38
rs902125573 Chr.16: 2425832-2425832 on Build GRCh38
rs4238910 Chr.16: 2425786-2425786 on Build GRCh38
rs971361164 Chr.16: 2425948-2425948 on Build GRCh38
rs535355918 Chr.16: 2425632-2425632 on Build GRCh38
rs951665219 Chr.16: 2425654-2425654 on Build GRCh38
rs117268559 Chr.16: 2425722-2425722 on Build GRCh38
rs950132207 Chr.16: 2425503-2425503 on Build GRCh38
rs529914512 Chr.16: 2425658-2425658 on Build GRCh38
rs544991934 Chr.16: 2425796-2425796 on Build GRCh38
rs576075240 Chr.16: 2425790-2425790 on Build GRCh38
rs975816598 Chr.16: 2425917-2425917 on Build GRCh38
rs542918443 Chr.16: 2425784-2425784 on Build GRCh38
rs143261271 Chr.16: 2425647-2425647 on Build GRCh38
rs866732629 Chr.16: 2425608-2425608 on Build GRCh38
rs921824718 Chr.16: 2425684-2425684 on Build GRCh38
rs778591269 Chr.16: 2425674-2425674 on Build GRCh38
rs546408838 Chr.16: 2425900-2425900 on Build GRCh38
rs1014628067 Chr.16: 2425908-2425908 on Build GRCh38
rs529194287 Chr.16: 2425980-2425980 on Build GRCh38
rs950196851 Chr.16: 2425795-2425795 on Build GRCh38
rs1029819262 Chr.16: 2425653-2425653 on Build GRCh38
rs1023297651 Chr.16: 2425939-2425939 on Build GRCh38
rs1050806686 Chr.16: 2425867-2425867 on Build GRCh38
rs899498994 Chr.16: 2425648-2425648 on Build GRCh38
rs886835884 Chr.16: 2425869-2425869 on Build GRCh38
rs750661946 Chr.16: 2425964-2425964 on Build GRCh38
rs1012743529 Chr.16: 2425918-2425918 on Build GRCh38
rs557933006 Chr.16: 2425667-2425667 on Build GRCh38
rs4318208 Chr.16: 2425669-2425669 on Build GRCh38
rs1007827656 Chr.16: 2425585-2425585 on Build GRCh38
rs998247664 Chr.16: 2425839-2425839 on Build GRCh38
rs771519832 Chr.16: 2425941-2425941 on Build GRCh38
rs915874989 Chr.16: 2425692-2425696 on Build GRCh38
rs1054650562 Chr.16: 2425563-2425563 on Build GRCh38
rs976315994 Chr.16: 2425675-2425675 on Build GRCh38
rs1001717208 Chr.16: 2425873-2425873 on Build GRCh38
rs1014818603 Chr.16: 2425625-2425625 on Build GRCh38
rs4414494 Chr.16: 2425650-2425650 on Build GRCh38
rs1000158835 Chr.16: 2425548-2425548 on Build GRCh38
rs921833992 Chr.16: 2425482-2425482 on Build GRCh38
rs1049712728 Chr.16: 2425528-2425528 on Build GRCh38
rs891896251 Chr.16: 2425613-2425613 on Build GRCh38
rs935979575 Chr.16: 2425773-2425773 on Build GRCh38
rs1029165213 Chr.16: 2425922-2425922 on Build GRCh38
rs991376199 Chr.16: 2425685-2425685 on Build GRCh38
rs565071474 Chr.16: 2425798-2425798 on Build GRCh38
rs376487824 Chr.16: 2425721-2425721 on Build GRCh38
rs953185142 Chr.16: 2425953-2425953 on Build GRCh38
rs904491031 Chr.16: 2425536-2425536 on Build GRCh38
rs1054597773 Chr.16: 2425785-2425785 on Build GRCh38
rs771508213 Chr.16: 2425777-2425777 on Build GRCh38
rs532999839 Chr.16: 2425478-2425478 on Build GRCh38
rs962522400 Chr.16: 2425913-2425913 on Build GRCh38
rs1053103382 Chr.16: 2425580-2425580 on Build GRCh38
rs567268819 Chr.16: 2425901-2425901 on Build GRCh38
rs966187295 Chr.16: 2425915-2425915 on Build GRCh38
rs943561730 Chr.16: 2425793-2425793 on Build GRCh38
rs905998325 Chr.16: 2425850-2425850 on Build GRCh38
rs560377283 Chr.16: 2425777-2425777 on Build GRCh38
rs764274407 Chr.16: 2425508-2425508 on Build GRCh38
rs747808132 Chr.16: 2425763-2425763 on Build GRCh38
rs186906013 Chr.16: 2425535-2425535 on Build GRCh38
rs1004539772 Chr.16: 2425905-2425905 on Build GRCh38
rs565137842 Chr.16: 2425660-2425660 on Build GRCh38
rs374408148 Chr.16: 2425633-2425633 on Build GRCh38

View Full Product Details