Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 18596269-18596831 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ABCC6P1
Entrez Gene ID
653190
Gene Name
ATP binding cassette subfamily C member 6 pseudogene 1
Gene Aliases
-
UniGene
Hs.13188
Gene Chromosome Location
Chr.16: 18571248-18598289 on Build GRCh38
Transcripts
NR_003569.1

SNP Detail

SNP ID Chromosome Location
rs976321777 Chr.16: 18596352-18596352 on Build GRCh38
rs1019299473 Chr.16: 18596625-18596625 on Build GRCh38
rs1034251352 Chr.16: 18596304-18596304 on Build GRCh38
rs1029204990 Chr.16: 18596643-18596643 on Build GRCh38
rs985053199 Chr.16: 18596682-18596682 on Build GRCh38
rs967569648 Chr.16: 18596325-18596325 on Build GRCh38
rs113791780 Chr.16: 18596739-18596739 on Build GRCh38
rs375761635 Chr.16: 18596384-18596384 on Build GRCh38
rs1021375741 Chr.16: 18596324-18596324 on Build GRCh38
rs985157280 Chr.16: 18596414-18596415 on Build GRCh38
rs1031157432 Chr.16: 18596332-18596332 on Build GRCh38
rs1055674019 Chr.16: 18596792-18596792 on Build GRCh38
rs113156023 Chr.16: 18596315-18596315 on Build GRCh38
rs907061818 Chr.16: 18596503-18596503 on Build GRCh38
rs894355895 Chr.16: 18596557-18596557 on Build GRCh38
rs144127725 Chr.16: 18596489-18596489 on Build GRCh38
rs573628669 Chr.16: 18596511-18596511 on Build GRCh38
rs144682630 Chr.16: 18596544-18596544 on Build GRCh38
rs1011488804 Chr.16: 18596579-18596579 on Build GRCh38
rs575617480 Chr.16: 18596488-18596488 on Build GRCh38
rs940685520 Chr.16: 18596724-18596724 on Build GRCh38
rs868835874 Chr.16: 18596543-18596543 on Build GRCh38
rs966193602 Chr.16: 18596631-18596631 on Build GRCh38
rs368898670 Chr.16: 18596488-18596488 on Build GRCh38
rs976187871 Chr.16: 18596639-18596639 on Build GRCh38
rs928677325 Chr.16: 18596755-18596755 on Build GRCh38
rs1011623387 Chr.16: 18596318-18596318 on Build GRCh38
rs539917870 Chr.16: 18596778-18596778 on Build GRCh38
rs776120823 Chr.16: 18596580-18596580 on Build GRCh38
rs866849959 Chr.16: 18596781-18596781 on Build GRCh38
rs938496328 Chr.16: 18596763-18596763 on Build GRCh38
rs953798532 Chr.16: 18596668-18596668 on Build GRCh38
rs909235941 Chr.16: 18596701-18596701 on Build GRCh38
rs909213424 Chr.16: 18596466-18596466 on Build GRCh38
rs376801033 Chr.16: 18596303-18596303 on Build GRCh38
rs931992205 Chr.16: 18596411-18596411 on Build GRCh38
rs563516289 Chr.16: 18596488-18596489 on Build GRCh38
rs866526636 Chr.16: 18596526-18596526 on Build GRCh38
rs1036437938 Chr.16: 18596499-18596499 on Build GRCh38
rs1055735076 Chr.16: 18596549-18596549 on Build GRCh38
rs550919589 Chr.16: 18596624-18596624 on Build GRCh38
rs754784380 Chr.16: 18596370-18596371 on Build GRCh38
rs902833305 Chr.16: 18596612-18596612 on Build GRCh38
rs564690356 Chr.16: 18596335-18596335 on Build GRCh38
rs977188221 Chr.16: 18596329-18596329 on Build GRCh38

View Full Product Details