Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46479304-46479729 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26P1
Entrez Gene ID
124149
Gene Name
ankyrin repeat domain 26 pseudogene 1
Gene Aliases
-
UniGene
Hs.97414
Gene Chromosome Location
Chr.16: 46469337-46569097 on Build GRCh38
Transcripts
NR_026556.1

SNP Detail

SNP ID Chromosome Location
rs144318450 Chr.16: 46479331-46479331 on Build GRCh38
rs545022780 Chr.16: 46479367-46479367 on Build GRCh38
rs1021930975 Chr.16: 46479506-46479506 on Build GRCh38
rs184786474 Chr.16: 46479341-46479341 on Build GRCh38
rs968014611 Chr.16: 46479535-46479535 on Build GRCh38
rs1049331428 Chr.16: 46479625-46479625 on Build GRCh38
rs933051406 Chr.16: 46479403-46479403 on Build GRCh38
rs981220379 Chr.16: 46479570-46479570 on Build GRCh38
rs901566076 Chr.16: 46479655-46479655 on Build GRCh38
rs745493268 Chr.16: 46479390-46479390 on Build GRCh38
rs528284912 Chr.16: 46479641-46479641 on Build GRCh38
rs919414503 Chr.16: 46479655-46479656 on Build GRCh38
rs1036537144 Chr.16: 46479369-46479369 on Build GRCh38
rs140952383 Chr.16: 46479491-46479491 on Build GRCh38
rs188483726 Chr.16: 46479654-46479654 on Build GRCh38
rs990030885 Chr.16: 46479490-46479490 on Build GRCh38
rs999052633 Chr.16: 46479527-46479527 on Build GRCh38
rs865841723 Chr.16: 46479536-46479536 on Build GRCh38
rs955934894 Chr.16: 46479489-46479489 on Build GRCh38
rs929510472 Chr.16: 46479624-46479624 on Build GRCh38
rs770665983 Chr.16: 46479371-46479371 on Build GRCh38
rs563319810 Chr.16: 46479373-46479373 on Build GRCh38
rs770551137 Chr.16: 46479560-46479560 on Build GRCh38
rs1051542072 Chr.16: 46479422-46479422 on Build GRCh38
rs1025036805 Chr.16: 46479593-46479593 on Build GRCh38
rs888066390 Chr.16: 46479637-46479637 on Build GRCh38
rs1031937597 Chr.16: 46479473-46479473 on Build GRCh38
rs919427640 Chr.16: 46479616-46479616 on Build GRCh38
rs561387663 Chr.16: 46479520-46479520 on Build GRCh38
rs972343766 Chr.16: 46479627-46479629 on Build GRCh38
rs771622987 Chr.16: 46479430-46479430 on Build GRCh38
rs371774326 Chr.16: 46479495-46479495 on Build GRCh38
rs1051489504 Chr.16: 46479692-46479692 on Build GRCh38
rs911212195 Chr.16: 46479518-46479519 on Build GRCh38
rs959898568 Chr.16: 46479550-46479550 on Build GRCh38
rs1007207030 Chr.16: 46479651-46479651 on Build GRCh38
rs1056610333 Chr.16: 46479602-46479602 on Build GRCh38
rs926711695 Chr.16: 46479573-46479573 on Build GRCh38
rs1013610510 Chr.16: 46479558-46479558 on Build GRCh38
rs939461653 Chr.16: 46479596-46479596 on Build GRCh38
rs753362423 Chr.16: 46479632-46479632 on Build GRCh38
rs575725923 Chr.16: 46479377-46479377 on Build GRCh38
rs980804956 Chr.16: 46479611-46479612 on Build GRCh38
rs933501434 Chr.16: 46479682-46479682 on Build GRCh38

View Full Product Details