Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 46517945-46518507 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
ANKRD26P1
Entrez Gene ID
124149
Gene Name
ankyrin repeat domain 26 pseudogene 1
Gene Aliases
-
UniGene
Hs.97414
Gene Chromosome Location
Chr.16: 46469337-46569097 on Build GRCh38
Transcripts
NR_026556.1

Cosmic Detail

Cosmic ID Mutation AA Transcript Location
2920645 c.924T>C p.I308I Chr.16: 46518347-46518347 on Build GRCh38

SNP Detail

SNP ID Chromosome Location
rs868573532 Chr.16: 46518356-46518356 on Build GRCh38
rs1038357652 Chr.16: 46518220-46518220 on Build GRCh38
rs376668020 Chr.16: 46518087-46518090 on Build GRCh38
rs201717893 Chr.16: 46518472-46518473 on Build GRCh38
rs932699678 Chr.16: 46518480-46518480 on Build GRCh38
rs987080047 Chr.16: 46518008-46518008 on Build GRCh38
rs988979989 Chr.16: 46518248-46518248 on Build GRCh38
rs990569089 Chr.16: 46518370-46518370 on Build GRCh38
rs1028414826 Chr.16: 46518259-46518259 on Build GRCh38
rs891298425 Chr.16: 46518261-46518261 on Build GRCh38
rs770212345 Chr.16: 46518055-46518055 on Build GRCh38
rs941980724 Chr.16: 46518394-46518394 on Build GRCh38
rs184038308 Chr.16: 46518149-46518149 on Build GRCh38
rs879311065 Chr.16: 46518161-46518161 on Build GRCh38
rs372321803 Chr.16: 46518107-46518107 on Build GRCh38
rs919935889 Chr.16: 46518437-46518437 on Build GRCh38
rs996920407 Chr.16: 46518230-46518230 on Build GRCh38
rs375867786 Chr.16: 46518136-46518137 on Build GRCh38
rs926223166 Chr.16: 46518175-46518175 on Build GRCh38
rs1038326221 Chr.16: 46518407-46518407 on Build GRCh38
rs990642481 Chr.16: 46518081-46518081 on Build GRCh38
rs56227132 Chr.16: 46518222-46518222 on Build GRCh38
rs575892131 Chr.16: 46518136-46518136 on Build GRCh38
rs565360714 Chr.16: 46518286-46518286 on Build GRCh38
rs369692904 Chr.16: 46518046-46518046 on Build GRCh38
rs941051590 Chr.16: 46518337-46518337 on Build GRCh38
rs187726140 Chr.16: 46518301-46518301 on Build GRCh38
rs779610426 Chr.16: 46518358-46518358 on Build GRCh38
rs907766066 Chr.16: 46518392-46518392 on Build GRCh38
rs887078356 Chr.16: 46518192-46518192 on Build GRCh38
rs189893994 Chr.16: 46518348-46518348 on Build GRCh38
rs369587847 Chr.16: 46518036-46518036 on Build GRCh38
rs566151323 Chr.16: 46518454-46518454 on Build GRCh38
rs954796129 Chr.16: 46518086-46518089 on Build GRCh38
rs369476942 Chr.16: 46518459-46518462 on Build GRCh38
rs925013300 Chr.16: 46518065-46518065 on Build GRCh38
rs983717858 Chr.16: 46518386-46518386 on Build GRCh38
rs965115215 Chr.16: 46518151-46518151 on Build GRCh38
rs1008874023 Chr.16: 46518275-46518275 on Build GRCh38
rs142378856 Chr.16: 46518372-46518372 on Build GRCh38
rs150880221 Chr.16: 46518374-46518374 on Build GRCh38
rs937770623 Chr.16: 46518067-46518067 on Build GRCh38
rs117165801 Chr.16: 46518289-46518289 on Build GRCh38
rs80247277 Chr.16: 46518405-46518405 on Build GRCh38
rs987843890 Chr.16: 46518102-46518103 on Build GRCh38
rs528462812 Chr.16: 46518204-46518204 on Build GRCh38
rs148769591 Chr.16: 46518153-46518153 on Build GRCh38
rs71380959 Chr.16: 46518084-46518084 on Build GRCh38
rs753914927 Chr.16: 46517971-46517971 on Build GRCh38
rs534231843 Chr.16: 46518180-46518180 on Build GRCh38
rs775676298 Chr.16: 46518156-46518159 on Build GRCh38
rs1021000798 Chr.16: 46518018-46518018 on Build GRCh38
rs979076163 Chr.16: 46518324-46518324 on Build GRCh38
rs867484379 Chr.16: 46518360-46518360 on Build GRCh38

View Full Product Details