Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55724486-55725052 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES1P2
Entrez Gene ID
390732
Gene Name
carboxylesterase 1 pseudogene 2
Gene Aliases
-
UniGene
Hs.554599
Gene Chromosome Location
Chr.16: 55724925-55750211 on Build GRCh38
Transcripts
NR_033740.1

SNP Detail

SNP ID Chromosome Location
rs377598156 Chr.16: 55724658-55724658 on Build GRCh38
rs770560345 Chr.16: 55724800-55724800 on Build GRCh38
rs776822667 Chr.16: 55724927-55724927 on Build GRCh38
rs559590352 Chr.16: 55724769-55724769 on Build GRCh38
rs117053960 Chr.16: 55724586-55724586 on Build GRCh38
rs573326275 Chr.16: 55724585-55724585 on Build GRCh38
rs756084887 Chr.16: 55724628-55724628 on Build GRCh38
rs115761517 Chr.16: 55724838-55724838 on Build GRCh38
rs748124490 Chr.16: 55724584-55724584 on Build GRCh38
rs557966201 Chr.16: 55724553-55724553 on Build GRCh38
rs775425443 Chr.16: 55724642-55724642 on Build GRCh38
rs879729495 Chr.16: 55724857-55724857 on Build GRCh38
rs769088261 Chr.16: 55724923-55724923 on Build GRCh38
rs748796934 Chr.16: 55724776-55724776 on Build GRCh38
rs187712 Chr.16: 55724974-55724974 on Build GRCh38
rs769849545 Chr.16: 55724634-55724634 on Build GRCh38
rs765716920 Chr.16: 55724940-55724940 on Build GRCh38
rs535560476 Chr.16: 55725002-55725002 on Build GRCh38
rs386791005 Chr.16: 55724567-55724578 on Build GRCh38
rs774062164 Chr.16: 55724870-55724870 on Build GRCh38
rs370278348 Chr.16: 55724741-55724741 on Build GRCh38
rs553117504 Chr.16: 55724930-55724930 on Build GRCh38
rs8052177 Chr.16: 55724929-55724929 on Build GRCh38
rs745453200 Chr.16: 55724873-55724873 on Build GRCh38
rs777477620 Chr.16: 55724732-55724732 on Build GRCh38

View Full Product Details