Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.16: 55740292-55740841 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CES1P2
Entrez Gene ID
390732
Gene Name
carboxylesterase 1 pseudogene 2
Gene Aliases
-
UniGene
Hs.554599
Gene Chromosome Location
Chr.16: 55724925-55750211 on Build GRCh38
Transcripts
NR_033740.1

SNP Detail

SNP ID Chromosome Location
rs377020116 Chr.16: 55740422-55740422 on Build GRCh38
rs12926148 Chr.16: 55740516-55740516 on Build GRCh38
rs529657538 Chr.16: 55740341-55740341 on Build GRCh38
rs562939153 Chr.16: 55740606-55740606 on Build GRCh38
rs756245912 Chr.16: 55740757-55740757 on Build GRCh38
rs148151026 Chr.16: 55740651-55740651 on Build GRCh38
rs138995347 Chr.16: 55740454-55740454 on Build GRCh38
rs866102949 Chr.16: 55740352-55740352 on Build GRCh38
rs8051055 Chr.16: 55740671-55740671 on Build GRCh38
rs571805630 Chr.16: 55740750-55740750 on Build GRCh38
rs796971423 Chr.16: 55740376-55740376 on Build GRCh38
rs112121334 Chr.16: 55740346-55740346 on Build GRCh38
rs193278381 Chr.16: 55740504-55740504 on Build GRCh38
rs370052013 Chr.16: 55740339-55740339 on Build GRCh38
rs374575624 Chr.16: 55740742-55740742 on Build GRCh38
rs114925447 Chr.16: 55740637-55740637 on Build GRCh38
rs376399887 Chr.16: 55740662-55740662 on Build GRCh38
rs558085438 Chr.16: 55740532-55740532 on Build GRCh38
rs763618001 Chr.16: 55740700-55740700 on Build GRCh38
rs775232407 Chr.16: 55740607-55740607 on Build GRCh38
rs867845672 Chr.16: 55740653-55740653 on Build GRCh38
rs369593339 Chr.16: 55740481-55740481 on Build GRCh38
rs200251929 Chr.16: 55740380-55740380 on Build GRCh38
rs755933074 Chr.16: 55740333-55740333 on Build GRCh38
rs12444861 Chr.16: 55740648-55740648 on Build GRCh38
rs535038448 Chr.16: 55740765-55740765 on Build GRCh38
rs556599388 Chr.16: 55740456-55740456 on Build GRCh38
rs142250750 Chr.16: 55740543-55740543 on Build GRCh38
rs529298441 Chr.16: 55740553-55740553 on Build GRCh38
rs771427283 Chr.16: 55740576-55740576 on Build GRCh38
rs573420254 Chr.16: 55740534-55740534 on Build GRCh38
rs372803458 Chr.16: 55740344-55740344 on Build GRCh38
rs146364302 Chr.16: 55740552-55740552 on Build GRCh38
rs544618196 Chr.16: 55740554-55740554 on Build GRCh38
rs745388572 Chr.16: 55740530-55740530 on Build GRCh38
rs546480325 Chr.16: 55740690-55740690 on Build GRCh38
rs71393095 Chr.16: 55740501-55740501 on Build GRCh38

View Full Product Details