Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18582388-18582979 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC144B
Entrez Gene ID
284047
Gene Name
coiled-coil domain containing 144B (pseudogene)
Gene Aliases
-
UniGene
Hs.448012
Gene Chromosome Location
Chr.17: 18537800-18625617 on Build GRCh38
Transcripts
NR_036647.1

SNP Detail

SNP ID Chromosome Location
rs1000195087 Chr.17: 18582886-18582886 on Build GRCh38
rs752497213 Chr.17: 18582524-18582525 on Build GRCh38
rs957948444 Chr.17: 18582526-18582526 on Build GRCh38
rs563869152 Chr.17: 18582870-18582870 on Build GRCh38
rs1014817339 Chr.17: 18582847-18582847 on Build GRCh38
rs1032978000 Chr.17: 18582888-18582888 on Build GRCh38
rs1000137509 Chr.17: 18582519-18582530 on Build GRCh38
rs2649311 Chr.17: 18582706-18582706 on Build GRCh38
rs1056420622 Chr.17: 18582805-18582805 on Build GRCh38
rs570167156 Chr.17: 18582953-18582953 on Build GRCh38
rs530324717 Chr.17: 18582774-18582774 on Build GRCh38
rs544094379 Chr.17: 18582804-18582804 on Build GRCh38
rs1020761904 Chr.17: 18582517-18582518 on Build GRCh38
rs1041833074 Chr.17: 18582856-18582856 on Build GRCh38
rs200603812 Chr.17: 18582521-18582522 on Build GRCh38
rs909508689 Chr.17: 18582760-18582760 on Build GRCh38
rs1048084124 Chr.17: 18582707-18582707 on Build GRCh38
rs879194900 Chr.17: 18582520-18582521 on Build GRCh38
rs967426987 Chr.17: 18582519-18582522 on Build GRCh38
rs1055970818 Chr.17: 18582410-18582410 on Build GRCh38
rs561429553 Chr.17: 18582734-18582734 on Build GRCh38
rs200549049 Chr.17: 18582790-18582790 on Build GRCh38
rs911066940 Chr.17: 18582560-18582560 on Build GRCh38
rs896002096 Chr.17: 18582810-18582810 on Build GRCh38
rs2649313 Chr.17: 18582564-18582564 on Build GRCh38
rs1014353572 Chr.17: 18582458-18582458 on Build GRCh38
rs918313993 Chr.17: 18582677-18582677 on Build GRCh38
rs59250245 Chr.17: 18582558-18582559 on Build GRCh38
rs985307843 Chr.17: 18582537-18582537 on Build GRCh38
rs2649312 Chr.17: 18582604-18582604 on Build GRCh38
rs541960183 Chr.17: 18582574-18582574 on Build GRCh38
rs895936383 Chr.17: 18582428-18582428 on Build GRCh38
rs745311017 Chr.17: 18582689-18582689 on Build GRCh38
rs142405065 Chr.17: 18582521-18582524 on Build GRCh38

View Full Product Details