Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18594591-18595185 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC144B
Entrez Gene ID
284047
Gene Name
coiled-coil domain containing 144B (pseudogene)
Gene Aliases
-
UniGene
Hs.448012
Gene Chromosome Location
Chr.17: 18537800-18625617 on Build GRCh38
Transcripts
NR_036647.1

SNP Detail

SNP ID Chromosome Location
rs563491268 Chr.17: 18595017-18595017 on Build GRCh38
rs928368349 Chr.17: 18595116-18595116 on Build GRCh38
rs2649251 Chr.17: 18595150-18595150 on Build GRCh38
rs760693628 Chr.17: 18595149-18595150 on Build GRCh38
rs532417555 Chr.17: 18595033-18595033 on Build GRCh38
rs150054123 Chr.17: 18595055-18595055 on Build GRCh38
rs201440757 Chr.17: 18595128-18595128 on Build GRCh38
rs969848269 Chr.17: 18595071-18595071 on Build GRCh38
rs2649253 Chr.17: 18595074-18595074 on Build GRCh38
rs962957325 Chr.17: 18594960-18594960 on Build GRCh38
rs981188472 Chr.17: 18595072-18595072 on Build GRCh38

View Full Product Details