Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 18625377-18625932 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CCDC144B
Entrez Gene ID
284047
Gene Name
coiled-coil domain containing 144B (pseudogene)
Gene Aliases
-
UniGene
Hs.448012
Gene Chromosome Location
Chr.17: 18537800-18625617 on Build GRCh38
Transcripts
NR_036647.1

SNP Detail

SNP ID Chromosome Location
rs145581464 Chr.17: 18625816-18625816 on Build GRCh38
rs2936648 Chr.17: 18625610-18625610 on Build GRCh38
rs576991381 Chr.17: 18625678-18625678 on Build GRCh38
rs778712558 Chr.17: 18625868-18625868 on Build GRCh38
rs779883111 Chr.17: 18625408-18625408 on Build GRCh38
rs765381686 Chr.17: 18625479-18625479 on Build GRCh38
rs532564610 Chr.17: 18625832-18625832 on Build GRCh38
rs954140442 Chr.17: 18625552-18625552 on Build GRCh38
rs754187873 Chr.17: 18625476-18625476 on Build GRCh38
rs879374888 Chr.17: 18625726-18625726 on Build GRCh38
rs1037254708 Chr.17: 18625583-18625583 on Build GRCh38
rs376073632 Chr.17: 18625464-18625464 on Build GRCh38
rs543592682 Chr.17: 18625637-18625637 on Build GRCh38
rs2936647 Chr.17: 18625789-18625789 on Build GRCh38
rs371732809 Chr.17: 18625704-18625704 on Build GRCh38
rs377040870 Chr.17: 18625898-18625898 on Build GRCh38
rs1056767759 Chr.17: 18625856-18625856 on Build GRCh38
rs745947688 Chr.17: 18625397-18625397 on Build GRCh38
rs746912684 Chr.17: 18625410-18625410 on Build GRCh38
rs570202036 Chr.17: 18625438-18625438 on Build GRCh38
rs757559528 Chr.17: 18625478-18625478 on Build GRCh38
rs563494253 Chr.17: 18625663-18625663 on Build GRCh38
rs11652268 Chr.17: 18625820-18625820 on Build GRCh38
rs968414157 Chr.17: 18625527-18625527 on Build GRCh38
rs912710865 Chr.17: 18625569-18625569 on Build GRCh38
rs376030483 Chr.17: 18625423-18625423 on Build GRCh38
rs764175636 Chr.17: 18625860-18625860 on Build GRCh38
rs562993027 Chr.17: 18625521-18625521 on Build GRCh38
rs761142060 Chr.17: 18625444-18625444 on Build GRCh38
rs528414009 Chr.17: 18625738-18625738 on Build GRCh38
rs1003071143 Chr.17: 18625487-18625487 on Build GRCh38
rs184213625 Chr.17: 18625741-18625741 on Build GRCh38
rs552376322 Chr.17: 18625853-18625853 on Build GRCh38
rs531723943 Chr.17: 18625591-18625591 on Build GRCh38
rs375892325 Chr.17: 18625415-18625415 on Build GRCh38
rs868809785 Chr.17: 18625762-18625762 on Build GRCh38
rs550343311 Chr.17: 18625416-18625416 on Build GRCh38
rs1056829348 Chr.17: 18625656-18625656 on Build GRCh38
rs954201970 Chr.17: 18625743-18625743 on Build GRCh38
rs980785867 Chr.17: 18625842-18625842 on Build GRCh38
rs758436484 Chr.17: 18625407-18625407 on Build GRCh38
rs866719099 Chr.17: 18625594-18625594 on Build GRCh38
rs775912105 Chr.17: 18625440-18625440 on Build GRCh38
rs955897407 Chr.17: 18625502-18625502 on Build GRCh38
rs1021452873 Chr.17: 18625695-18625695 on Build GRCh38
rs758425957 Chr.17: 18625486-18625486 on Build GRCh38
rs561709113 Chr.17: 18625768-18625768 on Build GRCh38
rs768617326 Chr.17: 18625411-18625411 on Build GRCh38
rs776566173 Chr.17: 18625414-18625414 on Build GRCh38
rs1010078283 Chr.17: 18625511-18625511 on Build GRCh38
rs996523545 Chr.17: 18625710-18625711 on Build GRCh38
rs905555193 Chr.17: 18625593-18625593 on Build GRCh38
rs2936649 Chr.17: 18625395-18625395 on Build GRCh38
rs530630422 Chr.17: 18625774-18625774 on Build GRCh38
rs931568778 Chr.17: 18625841-18625841 on Build GRCh38
rs915663392 Chr.17: 18625560-18625578 on Build GRCh38
rs898128723 Chr.17: 18625424-18625424 on Build GRCh38
rs796668539 Chr.17: 18625876-18625876 on Build GRCh38
rs766520400 Chr.17: 18625621-18625621 on Build GRCh38
rs780157472 Chr.17: 18625496-18625496 on Build GRCh38
rs559650285 Chr.17: 18625731-18625731 on Build GRCh38
rs940235460 Chr.17: 18625419-18625419 on Build GRCh38
rs1035474907 Chr.17: 18625489-18625489 on Build GRCh38
rs546061094 Chr.17: 18625703-18625703 on Build GRCh38
rs991272513 Chr.17: 18625537-18625560 on Build GRCh38
rs931598857 Chr.17: 18625590-18625590 on Build GRCh38
rs555379937 Chr.17: 18625884-18625884 on Build GRCh38
rs554547755 Chr.17: 18625585-18625585 on Build GRCh38
rs750615408 Chr.17: 18625484-18625484 on Build GRCh38
rs565985839 Chr.17: 18625874-18625874 on Build GRCh38
rs891596017 Chr.17: 18625665-18625665 on Build GRCh38
rs1010027433 Chr.17: 18625687-18625687 on Build GRCh38
rs565377948 Chr.17: 18625756-18625757 on Build GRCh38
rs558858157 Chr.17: 18625513-18625513 on Build GRCh38
rs995192852 Chr.17: 18625435-18625435 on Build GRCh38
rs905610205 Chr.17: 18625470-18625470 on Build GRCh38
rs920194901 Chr.17: 18625590-18625590 on Build GRCh38
rs772858145 Chr.17: 18625436-18625436 on Build GRCh38
rs940186355 Chr.17: 18625576-18625576 on Build GRCh38
rs369207123 Chr.17: 18625575-18625575 on Build GRCh38

View Full Product Details