Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.17: 30974995-30975553 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
DPRXP4
Entrez Gene ID
503645
Gene Name
divergent-paired related homeobox pseudogene 4
Gene Aliases
-
UniGene
Hs.683578
Gene Chromosome Location
Chr.17: 30975335-30976004 on Build GRCh38
Transcripts
NR_002221.1 NM_001184992.1 NM_032322.3 NM_197939.1 XM_005258043.4 XM_017025223.1

Gene Symbol
RNF135
Entrez Gene ID
84282
Gene Name
ring finger protein 135
Gene Aliases
L13, MMFD, REUL, Riplet
UniGene
Hs.29874
Gene Chromosome Location
Chr.17: 30968642-30999911 on Build GRCh38
Transcripts
NR_002221.1 NM_001184992.1 NM_032322.3 NM_197939.1 XM_005258043.4 XM_017025223.1

SNP Detail

SNP ID Chromosome Location
rs991395869 Chr.17: 30975157-30975157 on Build GRCh38
rs1011712322 Chr.17: 30975346-30975346 on Build GRCh38
rs1053410000 Chr.17: 30975436-30975436 on Build GRCh38
rs893410511 Chr.17: 30975301-30975301 on Build GRCh38
rs1020357121 Chr.17: 30975121-30975121 on Build GRCh38
rs7208589 Chr.17: 30975032-30975032 on Build GRCh38
rs957553239 Chr.17: 30975118-30975118 on Build GRCh38
rs1049612946 Chr.17: 30975391-30975391 on Build GRCh38
rs113573287 Chr.17: 30975021-30975021 on Build GRCh38
rs547984066 Chr.17: 30975511-30975511 on Build GRCh38
rs984103784 Chr.17: 30975260-30975260 on Build GRCh38
rs891814371 Chr.17: 30975473-30975473 on Build GRCh38
rs905981112 Chr.17: 30975519-30975519 on Build GRCh38
rs928666892 Chr.17: 30975147-30975147 on Build GRCh38
rs1030421533 Chr.17: 30975030-30975032 on Build GRCh38
rs530451880 Chr.17: 30975247-30975247 on Build GRCh38
rs944002947 Chr.17: 30975407-30975407 on Build GRCh38
rs909096212 Chr.17: 30975490-30975490 on Build GRCh38
rs540491977 Chr.17: 30975248-30975248 on Build GRCh38
rs1018433412 Chr.17: 30975074-30975075 on Build GRCh38
rs960321425 Chr.17: 30975254-30975254 on Build GRCh38
rs1035683600 Chr.17: 30975389-30975389 on Build GRCh38
rs909148518 Chr.17: 30975157-30975157 on Build GRCh38
rs543181819 Chr.17: 30975216-30975216 on Build GRCh38
rs963275220 Chr.17: 30975494-30975494 on Build GRCh38
rs991469612 Chr.17: 30975480-30975480 on Build GRCh38
rs557617932 Chr.17: 30975168-30975168 on Build GRCh38
rs1007590463 Chr.17: 30975067-30975067 on Build GRCh38
rs539221565 Chr.17: 30975103-30975103 on Build GRCh38
rs967485864 Chr.17: 30975360-30975360 on Build GRCh38
rs1039949264 Chr.17: 30975253-30975253 on Build GRCh38
rs1011764445 Chr.17: 30975120-30975121 on Build GRCh38
rs775301634 Chr.17: 30975231-30975231 on Build GRCh38
rs761056746 Chr.17: 30975393-30975393 on Build GRCh38
rs891782650 Chr.17: 30975207-30975207 on Build GRCh38
rs908211176 Chr.17: 30975278-30975278 on Build GRCh38
rs1036158483 Chr.17: 30975169-30975169 on Build GRCh38
rs971866273 Chr.17: 30975500-30975500 on Build GRCh38
rs981458522 Chr.17: 30975137-30975143 on Build GRCh38
rs550337796 Chr.17: 30975382-30975382 on Build GRCh38
rs891074782 Chr.17: 30975154-30975154 on Build GRCh38
rs111710995 Chr.17: 30975217-30975217 on Build GRCh38
rs1008130150 Chr.17: 30975156-30975156 on Build GRCh38
rs1038494521 Chr.17: 30975036-30975036 on Build GRCh38
rs183227868 Chr.17: 30975211-30975211 on Build GRCh38
rs1020471949 Chr.17: 30975347-30975347 on Build GRCh38
rs532305773 Chr.17: 30975317-30975317 on Build GRCh38
rs919644426 Chr.17: 30975378-30975378 on Build GRCh38
rs967869114 Chr.17: 30975121-30975121 on Build GRCh38
rs1011963864 Chr.17: 30975477-30975477 on Build GRCh38
rs566434964 Chr.17: 30975523-30975523 on Build GRCh38
rs1031710731 Chr.17: 30975194-30975194 on Build GRCh38
rs898332938 Chr.17: 30975045-30975045 on Build GRCh38
rs1049684020 Chr.17: 30975110-30975110 on Build GRCh38
rs529701933 Chr.17: 30975507-30975507 on Build GRCh38
rs973865545 Chr.17: 30975337-30975337 on Build GRCh38
rs912396509 Chr.17: 30975401-30975401 on Build GRCh38
rs562384743 Chr.17: 30975402-30975402 on Build GRCh38
rs565047596 Chr.17: 30975270-30975270 on Build GRCh38
rs546276874 Chr.17: 30975316-30975316 on Build GRCh38
rs1003393297 Chr.17: 30975377-30975377 on Build GRCh38
rs888801603 Chr.17: 30975218-30975218 on Build GRCh38

View Full Product Details