Product Details

Primer Sequences
Log in to view
Primer Chromosome Location
Chr.18: 14337406-14337953 on Build GRCh38
Format
Each primer will be delivered in a single tube, dried-down
Quick Reference Guide
Primer Designer Tool Quick Reference
Ampliseq ID's

Gene Details

Gene Symbol
CYP4F35P
Entrez Gene ID
284233
Gene Name
cytochrome P450 family 4 subfamily F member 35, pseudogene
Gene Aliases
-
UniGene
Hs.607579
Gene Chromosome Location
Chr.18: 14337423-14342524 on Build GRCh38
Transcripts
NR_026756.1

SNP Detail

SNP ID Chromosome Location
rs144017813 Chr.18: 14337444-14337444 on Build GRCh38
rs536731793 Chr.18: 14337533-14337533 on Build GRCh38
rs533537001 Chr.18: 14337446-14337446 on Build GRCh38
rs556612530 Chr.18: 14337536-14337536 on Build GRCh38
rs923907496 Chr.18: 14337663-14337663 on Build GRCh38
rs1007881642 Chr.18: 14337549-14337549 on Build GRCh38
rs896832534 Chr.18: 14337552-14337552 on Build GRCh38
rs564523353 Chr.18: 14337724-14337724 on Build GRCh38
rs570218539 Chr.18: 14337560-14337560 on Build GRCh38
rs147571293 Chr.18: 14337755-14337755 on Build GRCh38
rs199789840 Chr.18: 14337697-14337697 on Build GRCh38
rs749938789 Chr.18: 14337532-14337532 on Build GRCh38
rs1023787813 Chr.18: 14337779-14337779 on Build GRCh38
rs889740110 Chr.18: 14337547-14337547 on Build GRCh38
rs1027142972 Chr.18: 14337760-14337760 on Build GRCh38
rs943908748 Chr.18: 14337736-14337736 on Build GRCh38
rs990993580 Chr.18: 14337436-14337436 on Build GRCh38
rs953472048 Chr.18: 14337875-14337875 on Build GRCh38
rs148677275 Chr.18: 14337498-14337498 on Build GRCh38
rs185744851 Chr.18: 14337764-14337764 on Build GRCh38
rs12455096 Chr.18: 14337442-14337442 on Build GRCh38
rs994300802 Chr.18: 14337568-14337568 on Build GRCh38
rs369209099 Chr.18: 14337578-14337578 on Build GRCh38
rs79822489 Chr.18: 14337430-14337430 on Build GRCh38
rs952849432 Chr.18: 14337579-14337579 on Build GRCh38
rs990982162 Chr.18: 14337613-14337613 on Build GRCh38
rs1053888014 Chr.18: 14337503-14337503 on Build GRCh38
rs547218120 Chr.18: 14337471-14337471 on Build GRCh38
rs1031252786 Chr.18: 14337818-14337818 on Build GRCh38
rs748957921 Chr.18: 14337622-14337622 on Build GRCh38
rs889696233 Chr.18: 14337733-14337733 on Build GRCh38
rs564580468 Chr.18: 14337528-14337528 on Build GRCh38
rs935363262 Chr.18: 14337664-14337664 on Build GRCh38
rs986228470 Chr.18: 14337881-14337881 on Build GRCh38
rs1027539822 Chr.18: 14337433-14337433 on Build GRCh38
rs1012350718 Chr.18: 14337767-14337767 on Build GRCh38
rs553175939 Chr.18: 14337611-14337611 on Build GRCh38
rs761140783 Chr.18: 14337476-14337476 on Build GRCh38
rs879085253 Chr.18: 14337571-14337571 on Build GRCh38
rs896776008 Chr.18: 14337751-14337751 on Build GRCh38
rs949379534 Chr.18: 14337452-14337452 on Build GRCh38
rs892934999 Chr.18: 14337531-14337531 on Build GRCh38
rs924031246 Chr.18: 14337482-14337482 on Build GRCh38
rs952901303 Chr.18: 14337434-14337434 on Build GRCh38
rs1004179547 Chr.18: 14337817-14337817 on Build GRCh38
rs982121034 Chr.18: 14337643-14337643 on Build GRCh38
rs1019729487 Chr.18: 14337550-14337550 on Build GRCh38
rs956883513 Chr.18: 14337837-14337837 on Build GRCh38
rs540459863 Chr.18: 14337729-14337729 on Build GRCh38
rs12962258 Chr.18: 14337725-14337725 on Build GRCh38
rs763680710 Chr.18: 14337687-14337687 on Build GRCh38
rs571579311 Chr.18: 14337614-14337614 on Build GRCh38
rs970882113 Chr.18: 14337804-14337804 on Build GRCh38
rs568073995 Chr.18: 14337516-14337516 on Build GRCh38
rs73960803 Chr.18: 14337673-14337673 on Build GRCh38
rs575675906 Chr.18: 14337690-14337690 on Build GRCh38
rs529546350 Chr.18: 14337906-14337906 on Build GRCh38
rs796293012 Chr.18: 14337923-14337923 on Build GRCh38
rs888235693 Chr.18: 14337761-14337761 on Build GRCh38
rs915323416 Chr.18: 14337850-14337850 on Build GRCh38
rs116672683 Chr.18: 14337479-14337479 on Build GRCh38
rs865897920 Chr.18: 14337507-14337507 on Build GRCh38
rs1040987473 Chr.18: 14337749-14337749 on Build GRCh38
rs34001884 Chr.18: 14337671-14337671 on Build GRCh38
rs796605362 Chr.18: 14337917-14337917 on Build GRCh38
rs1023840323 Chr.18: 14337618-14337618 on Build GRCh38
rs547871593 Chr.18: 14337504-14337504 on Build GRCh38
rs140377143 Chr.18: 14337847-14337847 on Build GRCh38

View Full Product Details